Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121822428-121822917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL11
Entrez Gene ID
158135
Gene Name
tubulin tyrosine ligase like 11
Gene Aliases
C9orf20, bA244O19.1
UniGene
Hs.438937
Gene Chromosome Location
Chr.9: 121815674-122093606 on Build GRCh38
Transcripts
NM_001139442.1 XM_005251728.2 XM_005251729.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6013620 c.2262G>A p.R754R Chr.9: 121822728-121822728 on Build GRCh38
4620855 c.2246G>A p.R749H Chr.9: 121822744-121822744 on Build GRCh38
5677536 c.2271G>C p.P757P Chr.9: 121822719-121822719 on Build GRCh38
6530695 c.2244A>G p.K748K Chr.9: 121822746-121822746 on Build GRCh38
1104790 c.2176C>T p.P726S Chr.9: 121822814-121822814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756329198 Chr.9: 121822857-121822857 on Build GRCh38
rs983461021 Chr.9: 121822627-121822627 on Build GRCh38
rs974632187 Chr.9: 121822853-121822853 on Build GRCh38
rs748432964 Chr.9: 121822464-121822464 on Build GRCh38
rs753390416 Chr.9: 121822733-121822733 on Build GRCh38
rs557371956 Chr.9: 121822637-121822637 on Build GRCh38
rs749331174 Chr.9: 121822860-121822860 on Build GRCh38
rs766983167 Chr.9: 121822776-121822776 on Build GRCh38
rs906787371 Chr.9: 121822702-121822702 on Build GRCh38
rs905303174 Chr.9: 121822447-121822447 on Build GRCh38
rs182523001 Chr.9: 121822814-121822814 on Build GRCh38
rs529641246 Chr.9: 121822850-121822850 on Build GRCh38
rs936894416 Chr.9: 121822815-121822815 on Build GRCh38
rs755309273 Chr.9: 121822771-121822771 on Build GRCh38
rs545750235 Chr.9: 121822719-121822719 on Build GRCh38
rs1009627337 Chr.9: 121822863-121822863 on Build GRCh38
rs905321808 Chr.9: 121822671-121822671 on Build GRCh38
rs997172416 Chr.9: 121822893-121822893 on Build GRCh38
rs749206323 Chr.9: 121822696-121822696 on Build GRCh38
rs1054107219 Chr.9: 121822513-121822513 on Build GRCh38
rs769902746 Chr.9: 121822642-121822642 on Build GRCh38
rs1009817152 Chr.9: 121822692-121822692 on Build GRCh38
rs781405964 Chr.9: 121822635-121822635 on Build GRCh38
rs572971380 Chr.9: 121822729-121822729 on Build GRCh38
rs777976702 Chr.9: 121822859-121822859 on Build GRCh38
rs557898227 Chr.9: 121822487-121822487 on Build GRCh38
rs879087283 Chr.9: 121822494-121822494 on Build GRCh38
rs550824818 Chr.9: 121822762-121822762 on Build GRCh38
rs973920059 Chr.9: 121822580-121822580 on Build GRCh38
rs931072871 Chr.9: 121822576-121822576 on Build GRCh38
rs949624758 Chr.9: 121822645-121822645 on Build GRCh38
rs544169233 Chr.9: 121822564-121822564 on Build GRCh38
rs539395059 Chr.9: 121822463-121822463 on Build GRCh38
rs752061350 Chr.9: 121822764-121822764 on Build GRCh38
rs901137020 Chr.9: 121822885-121822885 on Build GRCh38
rs774750796 Chr.9: 121822730-121822730 on Build GRCh38
rs1053908987 Chr.9: 121822699-121822699 on Build GRCh38
rs961092812 Chr.9: 121822755-121822755 on Build GRCh38
rs970956122 Chr.9: 121822555-121822555 on Build GRCh38
rs956699987 Chr.9: 121822493-121822493 on Build GRCh38
rs563986013 Chr.9: 121822782-121822782 on Build GRCh38
rs564254181 Chr.9: 121822724-121822724 on Build GRCh38
rs771039843 Chr.9: 121822697-121822697 on Build GRCh38
rs763432013 Chr.9: 121822752-121822752 on Build GRCh38
rs752957915 Chr.9: 121822809-121822809 on Build GRCh38
rs759336197 Chr.9: 121822711-121822711 on Build GRCh38
rs916885484 Chr.9: 121822767-121822767 on Build GRCh38
rs763416030 Chr.9: 121822562-121822562 on Build GRCh38
rs778063545 Chr.9: 121822644-121822644 on Build GRCh38
rs1033843446 Chr.9: 121822571-121822571 on Build GRCh38
rs1009597569 Chr.9: 121822715-121822715 on Build GRCh38
rs983847300 Chr.9: 121822582-121822582 on Build GRCh38
rs543092374 Chr.9: 121822884-121822884 on Build GRCh38
rs750086288 Chr.9: 121822641-121822642 on Build GRCh38
rs984350967 Chr.9: 121822750-121822750 on Build GRCh38
rs908324793 Chr.9: 121822753-121822753 on Build GRCh38
rs755324706 Chr.9: 121822621-121822621 on Build GRCh38
rs1045652579 Chr.9: 121822647-121822647 on Build GRCh38
rs188581291 Chr.9: 121822530-121822530 on Build GRCh38
rs1045411569 Chr.9: 121822697-121822699 on Build GRCh38
rs1033790962 Chr.9: 121822716-121822716 on Build GRCh38
rs771770451 Chr.9: 121822714-121822714 on Build GRCh38
rs566540719 Chr.9: 121822506-121822506 on Build GRCh38
rs200414996 Chr.9: 121822708-121822708 on Build GRCh38
rs748328447 Chr.9: 121822636-121822636 on Build GRCh38
rs747321692 Chr.9: 121822605-121822605 on Build GRCh38
rs555251865 Chr.9: 121822539-121822539 on Build GRCh38
rs1056721646 Chr.9: 121822638-121822638 on Build GRCh38
rs965303542 Chr.9: 121822720-121822720 on Build GRCh38
rs192800946 Chr.9: 121822554-121822554 on Build GRCh38
rs771166459 Chr.9: 121822862-121822862 on Build GRCh38

View Full Product Details