Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 109541349-109541897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXT2
Entrez Gene ID
55916
Gene Name
nuclear transport factor 2 like export factor 2
Gene Aliases
P15-2
UniGene
Hs.25010
Gene Chromosome Location
Chr.X: 109535781-109544698 on Build GRCh38
Transcripts
NM_001242617.1 NM_001242618.1 NM_018698.4 XM_011530989.2 XM_017029648.1

SNP Detail

SNP ID Chromosome Location
rs766088998 Chr.X: 109541529-109541529 on Build GRCh38
rs1009234131 Chr.X: 109541645-109541645 on Build GRCh38
rs180794856 Chr.X: 109541463-109541463 on Build GRCh38
rs952404334 Chr.X: 109541763-109541763 on Build GRCh38
rs201313535 Chr.X: 109541442-109541442 on Build GRCh38
rs1054910354 Chr.X: 109541389-109541389 on Build GRCh38
rs749975378 Chr.X: 109541489-109541489 on Build GRCh38
rs1017744833 Chr.X: 109541866-109541866 on Build GRCh38
rs779570229 Chr.X: 109541647-109541647 on Build GRCh38
rs5903337 Chr.X: 109541688-109541689 on Build GRCh38
rs774095718 Chr.X: 109541437-109541437 on Build GRCh38
rs1048034683 Chr.X: 109541537-109541537 on Build GRCh38
rs375502259 Chr.X: 109541667-109541667 on Build GRCh38
rs996792601 Chr.X: 109541690-109541690 on Build GRCh38
rs1021541767 Chr.X: 109541833-109541833 on Build GRCh38
rs372211276 Chr.X: 109541475-109541475 on Build GRCh38
rs760308245 Chr.X: 109541504-109541504 on Build GRCh38
rs768368643 Chr.X: 109541801-109541801 on Build GRCh38
rs748678082 Chr.X: 109541735-109541735 on Build GRCh38
rs753108630 Chr.X: 109541657-109541657 on Build GRCh38
rs767534868 Chr.X: 109541832-109541832 on Build GRCh38
rs759647848 Chr.X: 109541797-109541797 on Build GRCh38
rs751735844 Chr.X: 109541681-109541681 on Build GRCh38
rs139869080 Chr.X: 109541609-109541609 on Build GRCh38
rs753534626 Chr.X: 109541546-109541546 on Build GRCh38

View Full Product Details