Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 124376126-124376663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TENM1
Entrez Gene ID
10178
Gene Name
teneurin transmembrane protein 1
Gene Aliases
ODZ1, ODZ3, TEN-M1, TNM, TNM1
UniGene
Hs.23796
Gene Chromosome Location
Chr.X: 124374680-125204305 on Build GRCh38
Transcripts
NM_001163278.1 NM_001163279.1 NM_014253.3 XM_011531230.2 XM_011531236.2 XM_011531237.2 XM_017029208.1 XM_017029209.1 XM_017029210.1 XM_017029211.1 XM_017029212.1 XM_017029213.1 XM_017029214.1

SNP Detail

SNP ID Chromosome Location
rs775635520 Chr.X: 124376329-124376329 on Build GRCh38
rs760424190 Chr.X: 124376624-124376624 on Build GRCh38
rs181447625 Chr.X: 124376461-124376461 on Build GRCh38
rs929387978 Chr.X: 124376430-124376430 on Build GRCh38
rs1045714392 Chr.X: 124376175-124376175 on Build GRCh38
rs73544875 Chr.X: 124376498-124376498 on Build GRCh38
rs150975707 Chr.X: 124376628-124376628 on Build GRCh38
rs974674493 Chr.X: 124376483-124376486 on Build GRCh38
rs1056480879 Chr.X: 124376257-124376257 on Build GRCh38
rs748958901 Chr.X: 124376388-124376388 on Build GRCh38
rs1001809263 Chr.X: 124376239-124376239 on Build GRCh38
rs1056469192 Chr.X: 124376387-124376387 on Build GRCh38
rs772222317 Chr.X: 124376155-124376155 on Build GRCh38
rs900419761 Chr.X: 124376598-124376598 on Build GRCh38
rs1017447168 Chr.X: 124376571-124376571 on Build GRCh38
rs772162053 Chr.X: 124376545-124376545 on Build GRCh38
rs938258682 Chr.X: 124376337-124376337 on Build GRCh38
rs188735355 Chr.X: 124376398-124376398 on Build GRCh38
rs775472106 Chr.X: 124376614-124376614 on Build GRCh38
rs985061101 Chr.X: 124376588-124376588 on Build GRCh38
rs777356814 Chr.X: 124376221-124376221 on Build GRCh38
rs999153479 Chr.X: 124376405-124376405 on Build GRCh38
rs1047818968 Chr.X: 124376438-124376438 on Build GRCh38
rs926927635 Chr.X: 124376277-124376277 on Build GRCh38
rs979554648 Chr.X: 124376251-124376251 on Build GRCh38
rs190603773 Chr.X: 124376637-124376637 on Build GRCh38

View Full Product Details