Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87727179-87727690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAPSS2
Entrez Gene ID
9060
Gene Name
3'-phosphoadenosine 5'-phosphosulfate synthase 2
Gene Aliases
ATPSK2, BCYM4, SK2
UniGene
Hs.524491
Gene Chromosome Location
Chr.10: 87659719-87747705 on Build GRCh38
Transcripts
NM_001015880.1 NM_004670.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5678771 c.984A>G p.L328L Chr.10: 87727402-87727402 on Build GRCh38
5126441 c.1030C>T p.R344C Chr.10: 87727448-87727448 on Build GRCh38
4016443 c.991G>A p.A331T Chr.10: 87727409-87727409 on Build GRCh38
6737581 c.985C>T p.R329* Chr.10: 87727403-87727403 on Build GRCh38
5553256 c.931G>T p.E311* Chr.10: 87727349-87727349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763447246 Chr.10: 87727522-87727522 on Build GRCh38
rs199576612 Chr.10: 87727343-87727343 on Build GRCh38
rs45467596 Chr.10: 87727289-87727289 on Build GRCh38
rs750923679 Chr.10: 87727404-87727404 on Build GRCh38
rs544558219 Chr.10: 87727286-87727286 on Build GRCh38
rs911657151 Chr.10: 87727397-87727397 on Build GRCh38
rs549724201 Chr.10: 87727594-87727594 on Build GRCh38
rs769771964 Chr.10: 87727267-87727267 on Build GRCh38
rs143480125 Chr.10: 87727523-87727523 on Build GRCh38
rs747947708 Chr.10: 87727346-87727346 on Build GRCh38
rs17127057 Chr.10: 87727214-87727214 on Build GRCh38
rs752982930 Chr.10: 87727440-87727440 on Build GRCh38
rs532144990 Chr.10: 87727443-87727443 on Build GRCh38
rs757872733 Chr.10: 87727239-87727239 on Build GRCh38
rs911725771 Chr.10: 87727535-87727535 on Build GRCh38
rs148986860 Chr.10: 87727381-87727381 on Build GRCh38
rs113435066 Chr.10: 87727413-87727413 on Build GRCh38
rs771777618 Chr.10: 87727479-87727479 on Build GRCh38
rs183111777 Chr.10: 87727408-87727408 on Build GRCh38
rs1038897478 Chr.10: 87727625-87727625 on Build GRCh38
rs747274873 Chr.10: 87727256-87727256 on Build GRCh38
rs368206390 Chr.10: 87727287-87727287 on Build GRCh38
rs758477460 Chr.10: 87727321-87727321 on Build GRCh38
rs765764640 Chr.10: 87727310-87727310 on Build GRCh38
rs267602608 Chr.10: 87727306-87727306 on Build GRCh38
rs560333289 Chr.10: 87727385-87727385 on Build GRCh38
rs754348801 Chr.10: 87727235-87727235 on Build GRCh38
rs900486111 Chr.10: 87727278-87727278 on Build GRCh38
rs915012214 Chr.10: 87727213-87727213 on Build GRCh38
rs759284893 Chr.10: 87727288-87727288 on Build GRCh38
rs775344235 Chr.10: 87727390-87727390 on Build GRCh38
rs772537897 Chr.10: 87727274-87727274 on Build GRCh38
rs900432702 Chr.10: 87727627-87727627 on Build GRCh38
rs895906554 Chr.10: 87727574-87727574 on Build GRCh38
rs867033588 Chr.10: 87727444-87727444 on Build GRCh38
rs548810141 Chr.10: 87727419-87727419 on Build GRCh38
rs148202527 Chr.10: 87727393-87727393 on Build GRCh38
rs74146361 Chr.10: 87727650-87727650 on Build GRCh38
rs944568982 Chr.10: 87727584-87727584 on Build GRCh38
rs754039934 Chr.10: 87727326-87727326 on Build GRCh38
rs770150344 Chr.10: 87727505-87727505 on Build GRCh38
rs768759177 Chr.10: 87727294-87727294 on Build GRCh38
rs200320883 Chr.10: 87727258-87727258 on Build GRCh38
rs376867519 Chr.10: 87727259-87727259 on Build GRCh38
rs780323088 Chr.10: 87727254-87727254 on Build GRCh38
rs201534761 Chr.10: 87727423-87727423 on Build GRCh38
rs774894555 Chr.10: 87727539-87727539 on Build GRCh38
rs775255860 Chr.10: 87727499-87727499 on Build GRCh38
rs751727686 Chr.10: 87727334-87727334 on Build GRCh38
rs762088120 Chr.10: 87727322-87727322 on Build GRCh38
rs773476234 Chr.10: 87727519-87727519 on Build GRCh38
rs985738480 Chr.10: 87727366-87727366 on Build GRCh38
rs770452479 Chr.10: 87727371-87727371 on Build GRCh38
rs1055774415 Chr.10: 87727536-87727536 on Build GRCh38
rs777759625 Chr.10: 87727352-87727352 on Build GRCh38
rs745360414 Chr.10: 87727466-87727466 on Build GRCh38
rs749223667 Chr.10: 87727370-87727370 on Build GRCh38
rs141138435 Chr.10: 87727449-87727449 on Build GRCh38
rs749210631 Chr.10: 87727446-87727446 on Build GRCh38
rs1048966989 Chr.10: 87727277-87727277 on Build GRCh38
rs778792401 Chr.10: 87727242-87727242 on Build GRCh38
rs147682410 Chr.10: 87727386-87727386 on Build GRCh38
rs769534289 Chr.10: 87727347-87727347 on Build GRCh38
rs953188794 Chr.10: 87727353-87727353 on Build GRCh38
rs750532009 Chr.10: 87727245-87727245 on Build GRCh38
rs17127065 Chr.10: 87727250-87727250 on Build GRCh38
rs121908952 Chr.10: 87727403-87727403 on Build GRCh38
rs780895988 Chr.10: 87727344-87727344 on Build GRCh38
rs766402310 Chr.10: 87727409-87727409 on Build GRCh38
rs767711277 Chr.10: 87727439-87727439 on Build GRCh38

View Full Product Details