Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97714868-97715370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARVELD1
Entrez Gene ID
83742
Gene Name
MARVEL domain containing 1
Gene Aliases
GB14, MARVD1, MRVLDC1, bA548K23.8
UniGene
Hs.744073
Gene Chromosome Location
Chr.10: 97713708-97718152 on Build GRCh38
Transcripts
NM_031484.3

SNP Detail

SNP ID Chromosome Location
rs955001454 Chr.10: 97715285-97715285 on Build GRCh38
rs896464974 Chr.10: 97715109-97715109 on Build GRCh38
rs955598714 Chr.10: 97715147-97715147 on Build GRCh38
rs1013644778 Chr.10: 97715151-97715151 on Build GRCh38
rs962598434 Chr.10: 97714900-97714900 on Build GRCh38
rs925264083 Chr.10: 97715053-97715053 on Build GRCh38
rs539383750 Chr.10: 97715012-97715012 on Build GRCh38
rs148497407 Chr.10: 97715310-97715310 on Build GRCh38
rs911008034 Chr.10: 97715332-97715332 on Build GRCh38
rs938378010 Chr.10: 97715304-97715304 on Build GRCh38
rs980383519 Chr.10: 97715258-97715258 on Build GRCh38
rs567514376 Chr.10: 97714912-97714912 on Build GRCh38
rs530618931 Chr.10: 97714999-97714999 on Build GRCh38
rs900390733 Chr.10: 97715083-97715083 on Build GRCh38
rs1047973704 Chr.10: 97715051-97715051 on Build GRCh38
rs1016840381 Chr.10: 97715228-97715228 on Build GRCh38
rs938435734 Chr.10: 97715041-97715041 on Build GRCh38
rs567513903 Chr.10: 97715239-97715239 on Build GRCh38
rs1016072825 Chr.10: 97715082-97715082 on Build GRCh38
rs113320735 Chr.10: 97715248-97715248 on Build GRCh38
rs934847901 Chr.10: 97714921-97714921 on Build GRCh38
rs144673082 Chr.10: 97714970-97714970 on Build GRCh38
rs550366431 Chr.10: 97715181-97715181 on Build GRCh38
rs541189162 Chr.10: 97715256-97715256 on Build GRCh38
rs73322600 Chr.10: 97715283-97715283 on Build GRCh38
rs752447929 Chr.10: 97715219-97715232 on Build GRCh38
rs986556643 Chr.10: 97715306-97715306 on Build GRCh38
rs547444572 Chr.10: 97714977-97714977 on Build GRCh38
rs964132603 Chr.10: 97715341-97715341 on Build GRCh38
rs986886485 Chr.10: 97715148-97715148 on Build GRCh38
rs558090760 Chr.10: 97715080-97715080 on Build GRCh38
rs1043943901 Chr.10: 97715016-97715016 on Build GRCh38
rs576356883 Chr.10: 97715154-97715154 on Build GRCh38
rs924801558 Chr.10: 97714920-97714920 on Build GRCh38
rs914744134 Chr.10: 97715243-97715243 on Build GRCh38
rs1028272865 Chr.10: 97715105-97715105 on Build GRCh38
rs1016480040 Chr.10: 97714892-97714893 on Build GRCh38
rs886262615 Chr.10: 97715052-97715052 on Build GRCh38
rs955136817 Chr.10: 97714976-97714976 on Build GRCh38
rs77370940 Chr.10: 97715159-97715159 on Build GRCh38
rs996057671 Chr.10: 97715092-97715092 on Build GRCh38
rs746424382 Chr.10: 97715313-97715313 on Build GRCh38
rs1044043883 Chr.10: 97714979-97714979 on Build GRCh38
rs990387209 Chr.10: 97714945-97714945 on Build GRCh38

View Full Product Details