Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97717498-97718008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARVELD1
Entrez Gene ID
83742
Gene Name
MARVEL domain containing 1
Gene Aliases
GB14, MARVD1, MRVLDC1, bA548K23.8
UniGene
Hs.744073
Gene Chromosome Location
Chr.10: 97713708-97718152 on Build GRCh38
Transcripts
NM_031484.3

SNP Detail

SNP ID Chromosome Location
rs976292838 Chr.10: 97717774-97717774 on Build GRCh38
rs769332685 Chr.10: 97717598-97717599 on Build GRCh38
rs11551585 Chr.10: 97717601-97717601 on Build GRCh38
rs372675686 Chr.10: 97717674-97717674 on Build GRCh38
rs1014716922 Chr.10: 97717946-97717946 on Build GRCh38
rs11551584 Chr.10: 97717973-97717973 on Build GRCh38
rs199831072 Chr.10: 97717599-97717599 on Build GRCh38
rs944898461 Chr.10: 97717769-97717769 on Build GRCh38
rs560969498 Chr.10: 97717826-97717826 on Build GRCh38
rs535853163 Chr.10: 97717822-97717822 on Build GRCh38
rs1001960775 Chr.10: 97717632-97717632 on Build GRCh38
rs975965199 Chr.10: 97717975-97717975 on Build GRCh38
rs1013482507 Chr.10: 97717667-97717667 on Build GRCh38
rs568700469 Chr.10: 97717902-97717902 on Build GRCh38
rs896346533 Chr.10: 97717655-97717655 on Build GRCh38
rs373708244 Chr.10: 97717782-97717783 on Build GRCh38
rs937771314 Chr.10: 97717819-97717819 on Build GRCh38
rs1003546418 Chr.10: 97717696-97717696 on Build GRCh38
rs900508803 Chr.10: 97717600-97717600 on Build GRCh38
rs796100147 Chr.10: 97717918-97717918 on Build GRCh38
rs759431497 Chr.10: 97717621-97717621 on Build GRCh38
rs1040604291 Chr.10: 97717778-97717778 on Build GRCh38
rs893115286 Chr.10: 97717834-97717834 on Build GRCh38
rs1027613793 Chr.10: 97717707-97717707 on Build GRCh38
rs920242632 Chr.10: 97717731-97717731 on Build GRCh38
rs8231 Chr.10: 97717932-97717932 on Build GRCh38
rs386746857 Chr.10: 97717599-97717601 on Build GRCh38
rs1043379896 Chr.10: 97717808-97717808 on Build GRCh38
rs951655460 Chr.10: 97717930-97717930 on Build GRCh38
rs532176643 Chr.10: 97717867-97717867 on Build GRCh38
rs995418839 Chr.10: 97717874-97717874 on Build GRCh38
rs1010278366 Chr.10: 97717846-97717846 on Build GRCh38
rs931873686 Chr.10: 97717799-97717799 on Build GRCh38
rs1043904 Chr.10: 97717956-97717956 on Build GRCh38
rs138161579 Chr.10: 97717794-97717794 on Build GRCh38
rs542930961 Chr.10: 97717745-97717745 on Build GRCh38
rs944975637 Chr.10: 97717525-97717525 on Build GRCh38
rs546891599 Chr.10: 97717839-97717839 on Build GRCh38
rs764196451 Chr.10: 97717681-97717681 on Build GRCh38
rs966174514 Chr.10: 97717937-97717937 on Build GRCh38
rs952123066 Chr.10: 97717732-97717732 on Build GRCh38
rs944522646 Chr.10: 97717772-97717772 on Build GRCh38
rs990527110 Chr.10: 97717729-97717729 on Build GRCh38
rs369457290 Chr.10: 97717964-97717964 on Build GRCh38
rs60930000 Chr.10: 97717599-97717599 on Build GRCh38
rs535915664 Chr.10: 97717768-97717768 on Build GRCh38
rs369921661 Chr.10: 97717787-97717787 on Build GRCh38
rs1013809005 Chr.10: 97717864-97717864 on Build GRCh38
rs1027343551 Chr.10: 97717920-97717920 on Build GRCh38
rs189937422 Chr.10: 97717858-97717858 on Build GRCh38
rs972015008 Chr.10: 97717699-97717699 on Build GRCh38
rs557045711 Chr.10: 97717913-97717913 on Build GRCh38
rs1027610466 Chr.10: 97717899-97717899 on Build GRCh38
rs149947527 Chr.10: 97717781-97717782 on Build GRCh38
rs896304484 Chr.10: 97717857-97717857 on Build GRCh38
rs375424830 Chr.10: 97717747-97717747 on Build GRCh38
rs961288785 Chr.10: 97717682-97717682 on Build GRCh38
rs150470272 Chr.10: 97717550-97717550 on Build GRCh38
rs180685262 Chr.10: 97717868-97717868 on Build GRCh38
rs573138966 Chr.10: 97717532-97717532 on Build GRCh38
rs1033508672 Chr.10: 97717640-97717640 on Build GRCh38
rs944997415 Chr.10: 97717570-97717570 on Build GRCh38
rs1041749091 Chr.10: 97717863-97717863 on Build GRCh38
rs983173521 Chr.10: 97717767-97717767 on Build GRCh38
rs944637522 Chr.10: 97717564-97717567 on Build GRCh38
rs971499809 Chr.10: 97717685-97717685 on Build GRCh38
rs542343810 Chr.10: 97717816-97717816 on Build GRCh38
rs1055305057 Chr.10: 97717849-97717849 on Build GRCh38
rs1043854 Chr.10: 97717547-97717547 on Build GRCh38
rs528049519 Chr.10: 97717831-97717831 on Build GRCh38
rs893246092 Chr.10: 97717631-97717631 on Build GRCh38
rs1043855 Chr.10: 97717548-97717548 on Build GRCh38
rs773919895 Chr.10: 97717609-97717609 on Build GRCh38
rs577522581 Chr.10: 97717586-97717586 on Build GRCh38

View Full Product Details