Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101131280-101131778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLX1
Entrez Gene ID
3195
Gene Name
T-cell leukemia homeobox 1
Gene Aliases
HOX11, TCL3
UniGene
Hs.89583
Gene Chromosome Location
Chr.10: 101130773-101137789 on Build GRCh38
Transcripts
NM_001195517.1 NM_005521.3 XM_011539744.2 XM_011539745.2 NR_130724.1

Gene Symbol
TLX1NB
Entrez Gene ID
100038246
Gene Name
TLX1 neighbor
Gene Aliases
APT-B7, TD1, TDI
UniGene
Hs.368784
Gene Chromosome Location
Chr.10: 101089321-101141266 on Build GRCh38
Transcripts
NM_001195517.1 NM_005521.3 XM_011539744.2 XM_011539745.2 NR_130724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5658683 c.37C>G p.H13D Chr.10: 101131578-101131578 on Build GRCh38
6551253 c.175G>T p.G59C Chr.10: 101131716-101131716 on Build GRCh38
7393964 c.92G>A p.G31D Chr.10: 101131633-101131633 on Build GRCh38
4011109 c.116G>A p.R39H Chr.10: 101131657-101131657 on Build GRCh38
6551252 c.114G>T p.S38S Chr.10: 101131655-101131655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020366741 Chr.10: 101131606-101131606 on Build GRCh38
rs1040218564 Chr.10: 101131529-101131529 on Build GRCh38
rs369093136 Chr.10: 101131621-101131621 on Build GRCh38
rs780244330 Chr.10: 101131734-101131734 on Build GRCh38
rs1046805140 Chr.10: 101131481-101131481 on Build GRCh38
rs756282883 Chr.10: 101131731-101131731 on Build GRCh38
rs992155351 Chr.10: 101131753-101131753 on Build GRCh38
rs377362073 Chr.10: 101131317-101131317 on Build GRCh38
rs780415955 Chr.10: 101131495-101131495 on Build GRCh38
rs965667097 Chr.10: 101131352-101131352 on Build GRCh38
rs1018575610 Chr.10: 101131332-101131332 on Build GRCh38
rs769004487 Chr.10: 101131504-101131504 on Build GRCh38
rs770195363 Chr.10: 101131492-101131492 on Build GRCh38
rs901399812 Chr.10: 101131653-101131653 on Build GRCh38
rs759034658 Chr.10: 101131559-101131559 on Build GRCh38
rs150815977 Chr.10: 101131595-101131595 on Build GRCh38
rs967515735 Chr.10: 101131402-101131402 on Build GRCh38
rs11599711 Chr.10: 101131703-101131703 on Build GRCh38
rs755280568 Chr.10: 101131741-101131741 on Build GRCh38
rs751280290 Chr.10: 101131583-101131583 on Build GRCh38
rs984981055 Chr.10: 101131505-101131505 on Build GRCh38
rs749671795 Chr.10: 101131500-101131500 on Build GRCh38
rs763630862 Chr.10: 101131700-101131700 on Build GRCh38
rs529590604 Chr.10: 101131485-101131485 on Build GRCh38
rs768935688 Chr.10: 101131624-101131624 on Build GRCh38
rs878941357 Chr.10: 101131437-101131437 on Build GRCh38
rs769237202 Chr.10: 101131676-101131676 on Build GRCh38
rs772430343 Chr.10: 101131518-101131518 on Build GRCh38
rs1032683928 Chr.10: 101131756-101131756 on Build GRCh38
rs773704342 Chr.10: 101131645-101131645 on Build GRCh38
rs547819047 Chr.10: 101131576-101131576 on Build GRCh38
rs756193245 Chr.10: 101131614-101131614 on Build GRCh38
rs200315484 Chr.10: 101131592-101131592 on Build GRCh38
rs564545434 Chr.10: 101131327-101131327 on Build GRCh38
rs937214339 Chr.10: 101131420-101131420 on Build GRCh38
rs142285605 Chr.10: 101131563-101131563 on Build GRCh38
rs774744622 Chr.10: 101131508-101131508 on Build GRCh38
rs751429042 Chr.10: 101131702-101131702 on Build GRCh38
rs940528700 Chr.10: 101131527-101131528 on Build GRCh38
rs545226571 Chr.10: 101131309-101131309 on Build GRCh38
rs780434498 Chr.10: 101131616-101131616 on Build GRCh38
rs199727081 Chr.10: 101131696-101131696 on Build GRCh38
rs893110223 Chr.10: 101131328-101131328 on Build GRCh38
rs930036609 Chr.10: 101131498-101131498 on Build GRCh38
rs7100938 Chr.10: 101131430-101131430 on Build GRCh38
rs201643256 Chr.10: 101131698-101131698 on Build GRCh38
rs531859900 Chr.10: 101131413-101131413 on Build GRCh38
rs976797739 Chr.10: 101131358-101131358 on Build GRCh38
rs779744037 Chr.10: 101131535-101131535 on Build GRCh38
rs568390501 Chr.10: 101131472-101131472 on Build GRCh38
rs776170968 Chr.10: 101131558-101131558 on Build GRCh38
rs1020332070 Chr.10: 101131399-101131399 on Build GRCh38
rs999899225 Chr.10: 101131740-101131740 on Build GRCh38
rs764936594 Chr.10: 101131566-101131566 on Build GRCh38
rs372550407 Chr.10: 101131598-101131598 on Build GRCh38
rs140202357 Chr.10: 101131633-101131633 on Build GRCh38
rs998472586 Chr.10: 101131657-101131657 on Build GRCh38
rs557943782 Chr.10: 101131757-101131757 on Build GRCh38
rs763908094 Chr.10: 101131580-101131580 on Build GRCh38
rs2742015 Chr.10: 101131752-101131752 on Build GRCh38
rs750325726 Chr.10: 101131729-101131729 on Build GRCh38
rs771871097 Chr.10: 101131714-101131715 on Build GRCh38
rs991284649 Chr.10: 101131452-101131452 on Build GRCh38
rs748007830 Chr.10: 101131524-101131524 on Build GRCh38
rs1003026900 Chr.10: 101131644-101131644 on Build GRCh38
rs752281847 Chr.10: 101131573-101131573 on Build GRCh38
rs1048542164 Chr.10: 101131514-101131514 on Build GRCh38
rs1012076974 Chr.10: 101131395-101131395 on Build GRCh38
rs1019968633 Chr.10: 101131389-101131389 on Build GRCh38
rs376987512 Chr.10: 101131677-101131677 on Build GRCh38
rs1027204962 Chr.10: 101131754-101131754 on Build GRCh38
rs775152441 Chr.10: 101131690-101131690 on Build GRCh38
rs550133755 Chr.10: 101131460-101131460 on Build GRCh38
rs759053139 Chr.10: 101131656-101131656 on Build GRCh38
rs867450849 Chr.10: 101131643-101131643 on Build GRCh38
rs753834322 Chr.10: 101131735-101131735 on Build GRCh38
rs950009721 Chr.10: 101131473-101131473 on Build GRCh38
rs762344186 Chr.10: 101131517-101131517 on Build GRCh38
rs145612659 Chr.10: 101131687-101131687 on Build GRCh38
rs925782818 Chr.10: 101131417-101131417 on Build GRCh38
rs772694205 Chr.10: 101131635-101131635 on Build GRCh38

View Full Product Details