Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103087895-103088393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NT5C2
Entrez Gene ID
22978
Gene Name
5'-nucleotidase, cytosolic II
Gene Aliases
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
UniGene
Hs.97439
Gene Chromosome Location
Chr.10: 103088017-103193306 on Build GRCh38
Transcripts
NM_001134373.2 NM_012229.4 XM_005269632.4 XM_005269633.4 XM_005269634.4 XM_005269635.4 XM_005269636.4 XM_005269637.4 XM_005269638.4 XM_005269639.4 XM_005269640.4 XM_005269641.4 XM_005269642.4 XM_005269643.4 XM_005269644.4 XM_005269645.4 XM_005269646.4 XM_006717721.3 XM_006717723.3 XM_011539537.2 XM_017015947.1 XM_017015948.1 XM_017015949.1 XM_017015950.1 XM_017015951.1 XM_017015952.1 XM_017015953.1 XM_017015954.1 XM_017015955.1 XM_017015956.1 XM_017015957.1 XM_017015958.1 XM_017015959.1 XM_017015960.1 XM_017015961.1 XM_017015962.1 XM_017015963.1 XM_017015964.1 XM_017015965.1 XM_017015966.1 XM_017015967.1 XM_017015968.1 XM_017015969.1 XM_017015970.1 XM_017015971.1 XM_017015972.1 XM_017015973.1 XM_017015974.1 XM_017015975.1 XM_017015976.1

SNP Detail

SNP ID Chromosome Location
rs553218262 Chr.10: 103088046-103088046 on Build GRCh38
rs529790614 Chr.10: 103088033-103088034 on Build GRCh38
rs773034894 Chr.10: 103088102-103088102 on Build GRCh38
rs752976301 Chr.10: 103088048-103088049 on Build GRCh38
rs965070285 Chr.10: 103088013-103088013 on Build GRCh38
rs188000434 Chr.10: 103088208-103088208 on Build GRCh38
rs749261653 Chr.10: 103088036-103088036 on Build GRCh38
rs8139 Chr.10: 103088366-103088366 on Build GRCh38
rs780661319 Chr.10: 103088268-103088271 on Build GRCh38
rs773511007 Chr.10: 103088028-103088029 on Build GRCh38
rs143048391 Chr.10: 103088128-103088128 on Build GRCh38
rs748828956 Chr.10: 103088025-103088026 on Build GRCh38
rs902730159 Chr.10: 103088277-103088277 on Build GRCh38
rs998492206 Chr.10: 103088000-103088000 on Build GRCh38
rs938057122 Chr.10: 103088152-103088152 on Build GRCh38
rs545125517 Chr.10: 103088048-103088048 on Build GRCh38
rs184769844 Chr.10: 103088007-103088007 on Build GRCh38
rs928049718 Chr.10: 103088149-103088149 on Build GRCh38
rs1052507047 Chr.10: 103087950-103087950 on Build GRCh38
rs979125620 Chr.10: 103088126-103088126 on Build GRCh38
rs749688116 Chr.10: 103088305-103088305 on Build GRCh38
rs573985912 Chr.10: 103088027-103088027 on Build GRCh38
rs993676263 Chr.10: 103088026-103088027 on Build GRCh38
rs142058693 Chr.10: 103087956-103087956 on Build GRCh38
rs547558133 Chr.10: 103088348-103088348 on Build GRCh38
rs536447923 Chr.10: 103088342-103088342 on Build GRCh38
rs996653683 Chr.10: 103088024-103088024 on Build GRCh38
rs1017714921 Chr.10: 103088070-103088070 on Build GRCh38
rs955838123 Chr.10: 103088129-103088129 on Build GRCh38
rs181636365 Chr.10: 103087985-103087985 on Build GRCh38
rs577782151 Chr.10: 103088047-103088047 on Build GRCh38
rs563129336 Chr.10: 103088069-103088069 on Build GRCh38
rs905431664 Chr.10: 103088002-103088002 on Build GRCh38
rs890194727 Chr.10: 103088300-103088300 on Build GRCh38
rs954460492 Chr.10: 103088035-103088036 on Build GRCh38
rs1040665611 Chr.10: 103088361-103088361 on Build GRCh38
rs756722265 Chr.10: 103088087-103088087 on Build GRCh38
rs934333576 Chr.10: 103088329-103088329 on Build GRCh38
rs1019330626 Chr.10: 103088011-103088011 on Build GRCh38
rs368943200 Chr.10: 103088048-103088051 on Build GRCh38
rs762673886 Chr.10: 103088123-103088123 on Build GRCh38
rs915403352 Chr.10: 103088217-103088217 on Build GRCh38
rs1046365586 Chr.10: 103088207-103088207 on Build GRCh38
rs952691014 Chr.10: 103088049-103088049 on Build GRCh38
rs938270721 Chr.10: 103088215-103088215 on Build GRCh38
rs557846344 Chr.10: 103087971-103087971 on Build GRCh38
rs1015636247 Chr.10: 103088081-103088081 on Build GRCh38
rs899365070 Chr.10: 103088015-103088015 on Build GRCh38
rs539569203 Chr.10: 103087967-103087967 on Build GRCh38
rs762876378 Chr.10: 103088258-103088259 on Build GRCh38
rs1047936997 Chr.10: 103088296-103088296 on Build GRCh38
rs1008673587 Chr.10: 103088363-103088363 on Build GRCh38
rs555679394 Chr.10: 103088026-103088026 on Build GRCh38
rs984437783 Chr.10: 103088065-103088065 on Build GRCh38
rs1038327287 Chr.10: 103088227-103088227 on Build GRCh38
rs913960452 Chr.10: 103088153-103088153 on Build GRCh38
rs375244612 Chr.10: 103088199-103088199 on Build GRCh38
rs936766994 Chr.10: 103087921-103087921 on Build GRCh38
rs777070202 Chr.10: 103088039-103088040 on Build GRCh38
rs988622712 Chr.10: 103088142-103088142 on Build GRCh38
rs541184140 Chr.10: 103088028-103088028 on Build GRCh38
rs529558966 Chr.10: 103088298-103088298 on Build GRCh38
rs899415109 Chr.10: 103088247-103088250 on Build GRCh38
rs759641696 Chr.10: 103088046-103088047 on Build GRCh38
rs1028095851 Chr.10: 103088033-103088033 on Build GRCh38
rs530577492 Chr.10: 103088099-103088099 on Build GRCh38
rs1043035022 Chr.10: 103088259-103088259 on Build GRCh38
rs553066733 Chr.10: 103087981-103087981 on Build GRCh38
rs949501280 Chr.10: 103088154-103088154 on Build GRCh38
rs947027349 Chr.10: 103088251-103088251 on Build GRCh38

View Full Product Details