Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122186002-122186421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC43
Entrez Gene ID
254050
Gene Name
leucine rich repeat containing 43
Gene Aliases
-
UniGene
Hs.374856
Gene Chromosome Location
Chr.12: 122167719-122203471 on Build GRCh38
Transcripts
NM_001098519.1 NM_152759.4 XM_017019124.1 XM_017019125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1213858 c.479G>A p.R160Q Chr.12: 122186257-122186257 on Build GRCh38
7375604 c.462G>T p.L154F Chr.12: 122186240-122186240 on Build GRCh38
4412513 c.412-9C>T p.? Chr.12: 122186181-122186181 on Build GRCh38
290491 c.456G>A p.E152E Chr.12: 122186234-122186234 on Build GRCh38
4439109 c.430G>C p.D144H Chr.12: 122186208-122186208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767324469 Chr.12: 122186221-122186221 on Build GRCh38
rs986141659 Chr.12: 122186134-122186134 on Build GRCh38
rs1033631375 Chr.12: 122186316-122186316 on Build GRCh38
rs745471870 Chr.12: 122186340-122186340 on Build GRCh38
rs535003286 Chr.12: 122186141-122186141 on Build GRCh38
rs768615954 Chr.12: 122186291-122186292 on Build GRCh38
rs753418820 Chr.12: 122186339-122186339 on Build GRCh38
rs371216115 Chr.12: 122186398-122186398 on Build GRCh38
rs368112879 Chr.12: 122186072-122186072 on Build GRCh38
rs114357090 Chr.12: 122186160-122186160 on Build GRCh38
rs779461164 Chr.12: 122186314-122186314 on Build GRCh38
rs755365274 Chr.12: 122186313-122186313 on Build GRCh38
rs930699881 Chr.12: 122186130-122186130 on Build GRCh38
rs79734505 Chr.12: 122186393-122186393 on Build GRCh38
rs779330004 Chr.12: 122186186-122186186 on Build GRCh38
rs780501018 Chr.12: 122186162-122186162 on Build GRCh38
rs769462417 Chr.12: 122186344-122186344 on Build GRCh38
rs373790801 Chr.12: 122186309-122186309 on Build GRCh38
rs746524600 Chr.12: 122186188-122186188 on Build GRCh38
rs138068954 Chr.12: 122186278-122186278 on Build GRCh38
rs757449027 Chr.12: 122186154-122186154 on Build GRCh38
rs553742228 Chr.12: 122186142-122186142 on Build GRCh38
rs1015431714 Chr.12: 122186070-122186070 on Build GRCh38
rs774218011 Chr.12: 122186257-122186257 on Build GRCh38
rs764782648 Chr.12: 122186377-122186377 on Build GRCh38
rs755489169 Chr.12: 122186181-122186181 on Build GRCh38
rs756895394 Chr.12: 122186366-122186366 on Build GRCh38
rs945519767 Chr.12: 122186107-122186107 on Build GRCh38
rs920699010 Chr.12: 122186110-122186110 on Build GRCh38
rs201842898 Chr.12: 122186240-122186240 on Build GRCh38
rs986174403 Chr.12: 122186364-122186364 on Build GRCh38
rs982052434 Chr.12: 122186035-122186035 on Build GRCh38
rs892916770 Chr.12: 122186243-122186243 on Build GRCh38
rs370859960 Chr.12: 122186291-122186291 on Build GRCh38
rs780764974 Chr.12: 122186209-122186209 on Build GRCh38
rs767713442 Chr.12: 122186274-122186274 on Build GRCh38
rs377255154 Chr.12: 122186334-122186334 on Build GRCh38
rs761930560 Chr.12: 122186268-122186268 on Build GRCh38
rs775386171 Chr.12: 122186231-122186231 on Build GRCh38
rs757817564 Chr.12: 122186266-122186266 on Build GRCh38
rs184399723 Chr.12: 122186249-122186249 on Build GRCh38
rs767842983 Chr.12: 122186155-122186155 on Build GRCh38
rs555459952 Chr.12: 122186091-122186091 on Build GRCh38
rs970583270 Chr.12: 122186025-122186025 on Build GRCh38
rs747474468 Chr.12: 122186120-122186120 on Build GRCh38
rs1022574152 Chr.12: 122186050-122186050 on Build GRCh38
rs749711177 Chr.12: 122186164-122186164 on Build GRCh38
rs966068501 Chr.12: 122186391-122186391 on Build GRCh38
rs370162492 Chr.12: 122186254-122186254 on Build GRCh38
rs750691595 Chr.12: 122186157-122186157 on Build GRCh38
rs531526199 Chr.12: 122186250-122186250 on Build GRCh38
rs189232351 Chr.12: 122186351-122186351 on Build GRCh38
rs753215919 Chr.12: 122186315-122186315 on Build GRCh38
rs146960575 Chr.12: 122186106-122186106 on Build GRCh38
rs1012560289 Chr.12: 122186031-122186031 on Build GRCh38
rs182225221 Chr.12: 122186194-122186194 on Build GRCh38
rs374355093 Chr.12: 122186276-122186276 on Build GRCh38
rs971303453 Chr.12: 122186058-122186058 on Build GRCh38
rs981745071 Chr.12: 122186067-122186067 on Build GRCh38

View Full Product Details