Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67406108-67406604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC1
Entrez Gene ID
29800
Gene Name
zinc finger DHHC-type containing 1
Gene Aliases
C16orf1, HSU90653, ZNF377
UniGene
Hs.624541
Gene Chromosome Location
Chr.16: 67394419-67416494 on Build GRCh38
Transcripts
NM_001323627.1 NM_013304.2 XM_005255915.1 XM_006721185.1 XM_011523058.1 XM_011523059.1 XM_011523060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6826058 c.249C>T p.Y83Y Chr.16: 67406203-67406203 on Build GRCh38
471945 c.156G>T p.Q52H Chr.16: 67406296-67406296 on Build GRCh38
3888765 c.141C>T p.P47P Chr.16: 67406311-67406311 on Build GRCh38
5043986 c.250G>C p.A84P Chr.16: 67406202-67406202 on Build GRCh38
4630236 c.196G>T p.G66C Chr.16: 67406256-67406256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769069403 Chr.16: 67406410-67406410 on Build GRCh38
rs780259886 Chr.16: 67406283-67406283 on Build GRCh38
rs770545062 Chr.16: 67406212-67406212 on Build GRCh38
rs931890362 Chr.16: 67406135-67406135 on Build GRCh38
rs924862710 Chr.16: 67406525-67406525 on Build GRCh38
rs985819117 Chr.16: 67406560-67406560 on Build GRCh38
rs762297548 Chr.16: 67406360-67406360 on Build GRCh38
rs376044283 Chr.16: 67406382-67406382 on Build GRCh38
rs769358583 Chr.16: 67406332-67406332 on Build GRCh38
rs566518264 Chr.16: 67406180-67406180 on Build GRCh38
rs8059722 Chr.16: 67406486-67406486 on Build GRCh38
rs893548086 Chr.16: 67406258-67406258 on Build GRCh38
rs201740223 Chr.16: 67406387-67406387 on Build GRCh38
rs8057903 Chr.16: 67406162-67406162 on Build GRCh38
rs770620951 Chr.16: 67406303-67406303 on Build GRCh38
rs199716510 Chr.16: 67406295-67406295 on Build GRCh38
rs766454880 Chr.16: 67406436-67406436 on Build GRCh38
rs923058341 Chr.16: 67406550-67406550 on Build GRCh38
rs143542767 Chr.16: 67406239-67406239 on Build GRCh38
rs200637668 Chr.16: 67406202-67406202 on Build GRCh38
rs747924012 Chr.16: 67406200-67406200 on Build GRCh38
rs748932806 Chr.16: 67406398-67406398 on Build GRCh38
rs759849248 Chr.16: 67406476-67406476 on Build GRCh38
rs745318366 Chr.16: 67406331-67406331 on Build GRCh38
rs905161219 Chr.16: 67406467-67406467 on Build GRCh38
rs953140846 Chr.16: 67406559-67406559 on Build GRCh38
rs149894745 Chr.16: 67406294-67406294 on Build GRCh38
rs756306844 Chr.16: 67406280-67406280 on Build GRCh38
rs199529796 Chr.16: 67406203-67406203 on Build GRCh38
rs529100810 Chr.16: 67406537-67406537 on Build GRCh38
rs568844374 Chr.16: 67406183-67406183 on Build GRCh38
rs778182426 Chr.16: 67406172-67406172 on Build GRCh38
rs1013603888 Chr.16: 67406421-67406421 on Build GRCh38
rs35530468 Chr.16: 67406185-67406186 on Build GRCh38
rs561859392 Chr.16: 67406516-67406516 on Build GRCh38
rs757275561 Chr.16: 67406178-67406178 on Build GRCh38
rs942451026 Chr.16: 67406571-67406571 on Build GRCh38
rs555524849 Chr.16: 67406257-67406257 on Build GRCh38
rs754070046 Chr.16: 67406368-67406368 on Build GRCh38
rs1013507490 Chr.16: 67406199-67406199 on Build GRCh38
rs750477709 Chr.16: 67406363-67406363 on Build GRCh38
rs751020663 Chr.16: 67406568-67406568 on Build GRCh38
rs758299772 Chr.16: 67406392-67406392 on Build GRCh38
rs8058306 Chr.16: 67406386-67406386 on Build GRCh38
rs369323711 Chr.16: 67406411-67406411 on Build GRCh38
rs758310277 Chr.16: 67406375-67406375 on Build GRCh38
rs991505036 Chr.16: 67406179-67406179 on Build GRCh38
rs1004500548 Chr.16: 67406509-67406509 on Build GRCh38
rs374370290 Chr.16: 67406184-67406184 on Build GRCh38
rs370038434 Chr.16: 67406245-67406245 on Build GRCh38
rs925160074 Chr.16: 67406565-67406565 on Build GRCh38
rs775156668 Chr.16: 67406333-67406333 on Build GRCh38
rs757365012 Chr.16: 67406269-67406269 on Build GRCh38
rs774907156 Chr.16: 67406340-67406340 on Build GRCh38
rs765063359 Chr.16: 67406351-67406351 on Build GRCh38
rs773720080 Chr.16: 67406419-67406419 on Build GRCh38
rs754181592 Chr.16: 67406475-67406475 on Build GRCh38
rs751666264 Chr.16: 67406174-67406174 on Build GRCh38
rs545506247 Chr.16: 67406418-67406418 on Build GRCh38
rs759312519 Chr.16: 67406339-67406339 on Build GRCh38
rs547627679 Chr.16: 67406545-67406545 on Build GRCh38
rs759996137 Chr.16: 67406159-67406159 on Build GRCh38
rs751340257 Chr.16: 67406379-67406379 on Build GRCh38
rs936154148 Chr.16: 67406255-67406255 on Build GRCh38
rs758842220 Chr.16: 67406164-67406164 on Build GRCh38
rs370858030 Chr.16: 67406256-67406256 on Build GRCh38
rs549945060 Chr.16: 67406489-67406489 on Build GRCh38
rs200282329 Chr.16: 67406186-67406186 on Build GRCh38
rs747927116 Chr.16: 67406284-67406284 on Build GRCh38
rs573935339 Chr.16: 67406278-67406278 on Build GRCh38
rs142114755 Chr.16: 67406330-67406330 on Build GRCh38
rs139149730 Chr.16: 67406230-67406230 on Build GRCh38
rs1015769356 Chr.16: 67406532-67406532 on Build GRCh38
rs112959868 Chr.16: 67406170-67406170 on Build GRCh38
rs373188989 Chr.16: 67406423-67406423 on Build GRCh38
rs965560967 Chr.16: 67406538-67406538 on Build GRCh38
rs757263885 Chr.16: 67406380-67406380 on Build GRCh38
rs772323214 Chr.16: 67406136-67406136 on Build GRCh38
rs766456247 Chr.16: 67406248-67406248 on Build GRCh38
rs776409408 Chr.16: 67406225-67406225 on Build GRCh38
rs564140126 Chr.16: 67406424-67406424 on Build GRCh38
rs767582429 Chr.16: 67406271-67406271 on Build GRCh38
rs776487512 Chr.16: 67406334-67406334 on Build GRCh38
rs114187593 Chr.16: 67406302-67406302 on Build GRCh38
rs781439226 Chr.16: 67406182-67406182 on Build GRCh38
rs372055076 Chr.16: 67406367-67406367 on Build GRCh38
rs746540603 Chr.16: 67406206-67406206 on Build GRCh38
rs768013359 Chr.16: 67406362-67406362 on Build GRCh38
rs781144632 Chr.16: 67406381-67406381 on Build GRCh38
rs952947905 Chr.16: 67406506-67406506 on Build GRCh38

View Full Product Details