Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69341410-69341908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIP7
Entrez Gene ID
51388
Gene Name
NIP7, nucleolar pre-rRNA processing protein
Gene Aliases
CGI-37, HSPC031, KD93
UniGene
Hs.585728
Gene Chromosome Location
Chr.16: 69339512-69343111 on Build GRCh38
Transcripts
NM_001199434.1 NM_016101.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1217347 c.446C>A p.S149Y Chr.16: 69341555-69341555 on Build GRCh38
6726323 c.533C>T p.T178M Chr.16: 69341642-69341642 on Build GRCh38
1324340 c.472C>T p.P158S Chr.16: 69341581-69341581 on Build GRCh38
32261 c.511G>C p.E171Q Chr.16: 69341620-69341620 on Build GRCh38
972881 c.480G>A p.A160A Chr.16: 69341589-69341589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189670796 Chr.16: 69341694-69341694 on Build GRCh38
rs1031617747 Chr.16: 69341482-69341482 on Build GRCh38
rs764930598 Chr.16: 69341643-69341645 on Build GRCh38
rs944338787 Chr.16: 69341599-69341599 on Build GRCh38
rs1021682885 Chr.16: 69341471-69341471 on Build GRCh38
rs989248821 Chr.16: 69341444-69341444 on Build GRCh38
rs774334140 Chr.16: 69341464-69341464 on Build GRCh38
rs532424115 Chr.16: 69341756-69341756 on Build GRCh38
rs564688522 Chr.16: 69341842-69341842 on Build GRCh38
rs148264436 Chr.16: 69341623-69341623 on Build GRCh38
rs751820427 Chr.16: 69341627-69341627 on Build GRCh38
rs968745579 Chr.16: 69341477-69341477 on Build GRCh38
rs964584036 Chr.16: 69341455-69341455 on Build GRCh38
rs748010016 Chr.16: 69341580-69341580 on Build GRCh38
rs767600362 Chr.16: 69341638-69341638 on Build GRCh38
rs143668884 Chr.16: 69341613-69341613 on Build GRCh38
rs916113059 Chr.16: 69341514-69341514 on Build GRCh38
rs200525173 Chr.16: 69341609-69341609 on Build GRCh38
rs771741273 Chr.16: 69341662-69341662 on Build GRCh38
rs141835490 Chr.16: 69341630-69341630 on Build GRCh38
rs899332163 Chr.16: 69341869-69341869 on Build GRCh38
rs369784611 Chr.16: 69341622-69341622 on Build GRCh38
rs377441956 Chr.16: 69341479-69341479 on Build GRCh38
rs973596335 Chr.16: 69341728-69341728 on Build GRCh38
rs528087620 Chr.16: 69341807-69341807 on Build GRCh38
rs200117312 Chr.16: 69341693-69341693 on Build GRCh38
rs764308412 Chr.16: 69341512-69341512 on Build GRCh38
rs756246125 Chr.16: 69341648-69341648 on Build GRCh38
rs975232787 Chr.16: 69341781-69341781 on Build GRCh38
rs973542211 Chr.16: 69341544-69341544 on Build GRCh38
rs371580331 Chr.16: 69341497-69341497 on Build GRCh38
rs756923321 Chr.16: 69341864-69341864 on Build GRCh38
rs922003421 Chr.16: 69341786-69341786 on Build GRCh38
rs944407908 Chr.16: 69341833-69341833 on Build GRCh38
rs561141926 Chr.16: 69341469-69341469 on Build GRCh38
rs755002243 Chr.16: 69341563-69341563 on Build GRCh38
rs539932241 Chr.16: 69341819-69341819 on Build GRCh38
rs768664197 Chr.16: 69341461-69341461 on Build GRCh38
rs956603889 Chr.16: 69341724-69341724 on Build GRCh38
rs764550979 Chr.16: 69341626-69341626 on Build GRCh38
rs772903739 Chr.16: 69341478-69341478 on Build GRCh38
rs758748820 Chr.16: 69341506-69341506 on Build GRCh38
rs367907296 Chr.16: 69341521-69341521 on Build GRCh38
rs900394284 Chr.16: 69341617-69341617 on Build GRCh38
rs557196940 Chr.16: 69341677-69341677 on Build GRCh38
rs745919863 Chr.16: 69341530-69341530 on Build GRCh38
rs373272972 Chr.16: 69341644-69341644 on Build GRCh38
rs201073050 Chr.16: 69341656-69341656 on Build GRCh38
rs759044976 Chr.16: 69341616-69341616 on Build GRCh38
rs8053850 Chr.16: 69341755-69341755 on Build GRCh38
rs777675472 Chr.16: 69341661-69341661 on Build GRCh38
rs375434121 Chr.16: 69341570-69341570 on Build GRCh38
rs960609305 Chr.16: 69341495-69341495 on Build GRCh38
rs573297829 Chr.16: 69341596-69341596 on Build GRCh38
rs960073730 Chr.16: 69341691-69341691 on Build GRCh38
rs989301299 Chr.16: 69341726-69341726 on Build GRCh38
rs746784045 Chr.16: 69341788-69341788 on Build GRCh38
rs148917509 Chr.16: 69341584-69341584 on Build GRCh38
rs746752621 Chr.16: 69341674-69341674 on Build GRCh38
rs747917774 Chr.16: 69341465-69341465 on Build GRCh38
rs114563518 Chr.16: 69341498-69341498 on Build GRCh38
rs113242511 Chr.16: 69341879-69341879 on Build GRCh38
rs181010195 Chr.16: 69341702-69341702 on Build GRCh38
rs550928461 Chr.16: 69341865-69341865 on Build GRCh38
rs765704225 Chr.16: 69341503-69341503 on Build GRCh38
rs369943294 Chr.16: 69341655-69341655 on Build GRCh38
rs77794544 Chr.16: 69341435-69341435 on Build GRCh38
rs200779986 Chr.16: 69341516-69341516 on Build GRCh38
rs778987296 Chr.16: 69341575-69341575 on Build GRCh38
rs752617802 Chr.16: 69341750-69341750 on Build GRCh38
rs780221560 Chr.16: 69341654-69341654 on Build GRCh38
rs145746516 Chr.16: 69341676-69341676 on Build GRCh38
rs769019429 Chr.16: 69341698-69341698 on Build GRCh38
rs1022073862 Chr.16: 69341643-69341643 on Build GRCh38
rs772846779 Chr.16: 69341588-69341588 on Build GRCh38
rs753128384 Chr.16: 69341504-69341504 on Build GRCh38
rs751809937 Chr.16: 69341515-69341515 on Build GRCh38
rs920742173 Chr.16: 69341740-69341740 on Build GRCh38
rs765949229 Chr.16: 69341490-69341490 on Build GRCh38
rs770470671 Chr.16: 69341603-69341603 on Build GRCh38
rs760182885 Chr.16: 69341489-69341489 on Build GRCh38
rs933328256 Chr.16: 69341792-69341792 on Build GRCh38
rs940864876 Chr.16: 69341581-69341581 on Build GRCh38
rs187120182 Chr.16: 69341859-69341859 on Build GRCh38
rs201518205 Chr.16: 69341602-69341602 on Build GRCh38
rs920354317 Chr.16: 69341550-69341550 on Build GRCh38

View Full Product Details