Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54075478-54076010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TARM1
Entrez Gene ID
441864
Gene Name
T cell-interacting, activating receptor on myeloid cells 1
Gene Aliases
OLT-2
UniGene
Hs.694674
Gene Chromosome Location
Chr.19: 54069342-54081365 on Build GRCh38
Transcripts
NM_001135686.2 XM_005278278.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4554754 c.62G>A p.R21K Chr.19: 54075891-54075891 on Build GRCh38
6354366 c.43G>A p.V15M Chr.19: 54075910-54075910 on Build GRCh38
5172037 c.42C>T p.C14C Chr.19: 54075911-54075911 on Build GRCh38
5390665 c.63G>A p.R21R Chr.19: 54075890-54075890 on Build GRCh38
5725469 c.53G>A p.G18E Chr.19: 54075900-54075900 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs958249992 Chr.19: 54075710-54075710 on Build GRCh38
rs772697930 Chr.19: 54075964-54075964 on Build GRCh38
rs587708485 Chr.19: 54075716-54075716 on Build GRCh38
rs989778642 Chr.19: 54075718-54075718 on Build GRCh38
rs369599381 Chr.19: 54075740-54075740 on Build GRCh38
rs183293125 Chr.19: 54075579-54075579 on Build GRCh38
rs957054862 Chr.19: 54075514-54075514 on Build GRCh38
rs919847937 Chr.19: 54075841-54075841 on Build GRCh38
rs1019685334 Chr.19: 54075670-54075670 on Build GRCh38
rs762656720 Chr.19: 54075523-54075523 on Build GRCh38
rs755231427 Chr.19: 54075910-54075910 on Build GRCh38
rs587596902 Chr.19: 54075689-54075689 on Build GRCh38
rs779088450 Chr.19: 54075935-54075935 on Build GRCh38
rs587691776 Chr.19: 54075533-54075533 on Build GRCh38
rs887138622 Chr.19: 54075870-54075870 on Build GRCh38
rs778847868 Chr.19: 54075831-54075831 on Build GRCh38
rs145741251 Chr.19: 54075787-54075787 on Build GRCh38
rs1033022509 Chr.19: 54075703-54075703 on Build GRCh38
rs60063583 Chr.19: 54075550-54075550 on Build GRCh38
rs945761129 Chr.19: 54075522-54075522 on Build GRCh38
rs749832998 Chr.19: 54075595-54075595 on Build GRCh38
rs925541326 Chr.19: 54075512-54075512 on Build GRCh38
rs752645367 Chr.19: 54075904-54075904 on Build GRCh38
rs779034049 Chr.19: 54075983-54075983 on Build GRCh38
rs958197632 Chr.19: 54075971-54075971 on Build GRCh38
rs766346373 Chr.19: 54075532-54075532 on Build GRCh38
rs587673311 Chr.19: 54075799-54075799 on Build GRCh38
rs587715773 Chr.19: 54075613-54075613 on Build GRCh38
rs372926695 Chr.19: 54075669-54075669 on Build GRCh38
rs372329421 Chr.19: 54075807-54075807 on Build GRCh38
rs587694779 Chr.19: 54075704-54075704 on Build GRCh38
rs587675009 Chr.19: 54075658-54075658 on Build GRCh38
rs781626749 Chr.19: 54075779-54075779 on Build GRCh38
rs201942479 Chr.19: 54075622-54075622 on Build GRCh38
rs912993116 Chr.19: 54075519-54075519 on Build GRCh38
rs1055602762 Chr.19: 54075810-54075810 on Build GRCh38
rs928385421 Chr.19: 54075781-54075781 on Build GRCh38
rs113936834 Chr.19: 54075685-54075685 on Build GRCh38
rs899888207 Chr.19: 54075911-54075911 on Build GRCh38
rs967010130 Chr.19: 54075766-54075766 on Build GRCh38
rs1008215501 Chr.19: 54075875-54075875 on Build GRCh38
rs995519460 Chr.19: 54075580-54075580 on Build GRCh38
rs587658712 Chr.19: 54075770-54075770 on Build GRCh38
rs964586833 Chr.19: 54075671-54075671 on Build GRCh38
rs199987542 Chr.19: 54075656-54075656 on Build GRCh38
rs931177543 Chr.19: 54075850-54075850 on Build GRCh38
rs912939286 Chr.19: 54075756-54075756 on Build GRCh38
rs77043638 Chr.19: 54075936-54075936 on Build GRCh38
rs749429056 Chr.19: 54075863-54075863 on Build GRCh38
rs1047061600 Chr.19: 54075605-54075605 on Build GRCh38
rs1047008738 Chr.19: 54075867-54075867 on Build GRCh38
rs587734155 Chr.19: 54075771-54075771 on Build GRCh38
rs111509031 Chr.19: 54075747-54075747 on Build GRCh38
rs997003497 Chr.19: 54075678-54075678 on Build GRCh38
rs587634469 Chr.19: 54075511-54075511 on Build GRCh38
rs587646372 Chr.19: 54075591-54075591 on Build GRCh38
rs112452835 Chr.19: 54075803-54075803 on Build GRCh38
rs989747637 Chr.19: 54075515-54075515 on Build GRCh38
rs1056140685 Chr.19: 54075576-54075576 on Build GRCh38

View Full Product Details