Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54161912-54162427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMC4
Entrez Gene ID
147798
Gene Name
transmembrane channel like 4
Gene Aliases
-
UniGene
Hs.355126
Gene Chromosome Location
Chr.19: 54160095-54173250 on Build GRCh38
Transcripts
NM_001145303.2 NM_144686.3 XM_011526486.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5105094 c.1686+5G>A p.? Chr.19: 54162097-54162097 on Build GRCh38
87822 c.1601C>T p.T534M Chr.19: 54162187-54162187 on Build GRCh38
4982745 c.438C>T p.F146F Chr.19: 54162114-54162114 on Build GRCh38
440254 c.1573G>A p.E525K Chr.19: 54162215-54162215 on Build GRCh38
5105095 c.450+5G>A p.? Chr.19: 54162097-54162097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764621289 Chr.19: 54162198-54162198 on Build GRCh38
rs757833720 Chr.19: 54162320-54162320 on Build GRCh38
rs775674628 Chr.19: 54162111-54162111 on Build GRCh38
rs111613244 Chr.19: 54162392-54162392 on Build GRCh38
rs751160737 Chr.19: 54162322-54162322 on Build GRCh38
rs963925461 Chr.19: 54162036-54162036 on Build GRCh38
rs763603298 Chr.19: 54162321-54162321 on Build GRCh38
rs36655 Chr.19: 54162234-54162234 on Build GRCh38
rs770848254 Chr.19: 54162061-54162061 on Build GRCh38
rs762337344 Chr.19: 54162144-54162144 on Build GRCh38
rs550289323 Chr.19: 54161947-54161947 on Build GRCh38
rs553889804 Chr.19: 54162361-54162361 on Build GRCh38
rs756983345 Chr.19: 54162216-54162216 on Build GRCh38
rs780356098 Chr.19: 54162259-54162272 on Build GRCh38
rs370476133 Chr.19: 54162273-54162273 on Build GRCh38
rs780823221 Chr.19: 54162326-54162326 on Build GRCh38
rs373591201 Chr.19: 54162275-54162275 on Build GRCh38
rs779701144 Chr.19: 54162391-54162392 on Build GRCh38
rs572205842 Chr.19: 54162393-54162393 on Build GRCh38
rs199687894 Chr.19: 54162163-54162163 on Build GRCh38
rs369129546 Chr.19: 54162386-54162387 on Build GRCh38
rs370886990 Chr.19: 54162267-54162267 on Build GRCh38
rs746328269 Chr.19: 54162250-54162259 on Build GRCh38
rs12978996 Chr.19: 54162324-54162324 on Build GRCh38
rs547056678 Chr.19: 54162004-54162004 on Build GRCh38
rs67618081 Chr.19: 54162387-54162387 on Build GRCh38
rs3816129 Chr.19: 54162342-54162342 on Build GRCh38
rs751573380 Chr.19: 54162111-54162112 on Build GRCh38
rs756491176 Chr.19: 54162252-54162258 on Build GRCh38
rs752299652 Chr.19: 54162199-54162199 on Build GRCh38
rs761063744 Chr.19: 54162293-54162293 on Build GRCh38
rs760989445 Chr.19: 54162079-54162079 on Build GRCh38
rs746666219 Chr.19: 54162215-54162215 on Build GRCh38
rs781554171 Chr.19: 54162248-54162248 on Build GRCh38
rs377550147 Chr.19: 54162181-54162181 on Build GRCh38
rs992792027 Chr.19: 54162348-54162348 on Build GRCh38
rs373393088 Chr.19: 54162182-54162182 on Build GRCh38
rs766966046 Chr.19: 54162294-54162294 on Build GRCh38
rs538104744 Chr.19: 54161945-54161945 on Build GRCh38
rs756674689 Chr.19: 54162186-54162186 on Build GRCh38
rs777466101 Chr.19: 54162212-54162212 on Build GRCh38
rs144865322 Chr.19: 54162243-54162243 on Build GRCh38
rs918192278 Chr.19: 54162354-54162354 on Build GRCh38
rs775413179 Chr.19: 54162257-54162257 on Build GRCh38
rs757845372 Chr.19: 54162238-54162246 on Build GRCh38
rs538545933 Chr.19: 54162268-54162268 on Build GRCh38
rs375869770 Chr.19: 54162264-54162264 on Build GRCh38
rs373394460 Chr.19: 54162090-54162090 on Build GRCh38
rs955789676 Chr.19: 54162404-54162404 on Build GRCh38
rs79161038 Chr.19: 54162403-54162403 on Build GRCh38
rs370724130 Chr.19: 54162256-54162256 on Build GRCh38
rs779192332 Chr.19: 54162011-54162011 on Build GRCh38
rs936772498 Chr.19: 54162397-54162398 on Build GRCh38
rs764462813 Chr.19: 54162136-54162136 on Build GRCh38
rs973748714 Chr.19: 54162125-54162125 on Build GRCh38
rs545931335 Chr.19: 54162394-54162394 on Build GRCh38
rs948000443 Chr.19: 54162392-54162393 on Build GRCh38
rs891593334 Chr.19: 54162399-54162400 on Build GRCh38
rs1016522478 Chr.19: 54161995-54161995 on Build GRCh38
rs60122470 Chr.19: 54162387-54162387 on Build GRCh38
rs905544784 Chr.19: 54162350-54162350 on Build GRCh38
rs773664919 Chr.19: 54162278-54162278 on Build GRCh38
rs774117731 Chr.19: 54162134-54162134 on Build GRCh38
rs567044941 Chr.19: 54162313-54162313 on Build GRCh38
rs780983859 Chr.19: 54162233-54162233 on Build GRCh38
rs111291755 Chr.19: 54162391-54162391 on Build GRCh38
rs901491649 Chr.19: 54162258-54162258 on Build GRCh38
rs375944512 Chr.19: 54161965-54161965 on Build GRCh38
rs777521161 Chr.19: 54162055-54162055 on Build GRCh38
rs750838464 Chr.19: 54162185-54162185 on Build GRCh38
rs776480502 Chr.19: 54162075-54162075 on Build GRCh38
rs138980997 Chr.19: 54162194-54162194 on Build GRCh38
rs764315607 Chr.19: 54162092-54162092 on Build GRCh38
rs767998477 Chr.19: 54162169-54162169 on Build GRCh38
rs774845118 Chr.19: 54162143-54162143 on Build GRCh38
rs375250638 Chr.19: 54162261-54162261 on Build GRCh38
rs999779500 Chr.19: 54162358-54162358 on Build GRCh38
rs769717168 Chr.19: 54162108-54162108 on Build GRCh38
rs373710661 Chr.19: 54161958-54161958 on Build GRCh38
rs746862124 Chr.19: 54162060-54162060 on Build GRCh38
rs981318056 Chr.19: 54161985-54161985 on Build GRCh38
rs771795779 Chr.19: 54162052-54162052 on Build GRCh38
rs757971688 Chr.19: 54162206-54162206 on Build GRCh38
rs564515730 Chr.19: 54162399-54162399 on Build GRCh38
rs200760133 Chr.19: 54162080-54162080 on Build GRCh38

View Full Product Details