Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 106805260-106805762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST6GAL2
Entrez Gene ID
84620
Gene Name
ST6 beta-galactoside alpha-2,6-sialyltransferase 2
Gene Aliases
SIAT2, ST6GalII
UniGene
Hs.98265
Gene Chromosome Location
Chr.2: 106801600-106887410 on Build GRCh38
Transcripts
NM_001142351.1 NM_001322362.1 NM_032528.2 NR_136313.1 XM_006712802.1 XM_011511999.2

SNP Detail

SNP ID Chromosome Location
rs947459078 Chr.2: 106805579-106805580 on Build GRCh38
rs781702433 Chr.2: 106805703-106805703 on Build GRCh38
rs912734250 Chr.2: 106805453-106805453 on Build GRCh38
rs933108364 Chr.2: 106805412-106805412 on Build GRCh38
rs757491993 Chr.2: 106805612-106805612 on Build GRCh38
rs568144904 Chr.2: 106805408-106805408 on Build GRCh38
rs950901191 Chr.2: 106805329-106805329 on Build GRCh38
rs1041731740 Chr.2: 106805601-106805601 on Build GRCh38
rs764031957 Chr.2: 106805304-106805304 on Build GRCh38
rs1051440821 Chr.2: 106805431-106805431 on Build GRCh38
rs962417848 Chr.2: 106805345-106805345 on Build GRCh38
rs533531552 Chr.2: 106805698-106805698 on Build GRCh38
rs763067972 Chr.2: 106805300-106805300 on Build GRCh38
rs909461959 Chr.2: 106805713-106805713 on Build GRCh38
rs891656559 Chr.2: 106805459-106805459 on Build GRCh38
rs950966801 Chr.2: 106805661-106805661 on Build GRCh38
rs914619342 Chr.2: 106805578-106805578 on Build GRCh38
rs909443029 Chr.2: 106805357-106805357 on Build GRCh38
rs1025034381 Chr.2: 106805621-106805621 on Build GRCh38
rs898262902 Chr.2: 106805738-106805738 on Build GRCh38
rs935287567 Chr.2: 106805632-106805632 on Build GRCh38
rs1033720878 Chr.2: 106805554-106805554 on Build GRCh38
rs189158200 Chr.2: 106805692-106805692 on Build GRCh38
rs975538658 Chr.2: 106805552-106805552 on Build GRCh38
rs1011999340 Chr.2: 106805321-106805321 on Build GRCh38
rs904407737 Chr.2: 106805506-106805506 on Build GRCh38
rs535109194 Chr.2: 106805660-106805660 on Build GRCh38
rs987048116 Chr.2: 106805437-106805437 on Build GRCh38
rs941693236 Chr.2: 106805383-106805383 on Build GRCh38
rs549410674 Chr.2: 106805330-106805330 on Build GRCh38
rs552434793 Chr.2: 106805395-106805395 on Build GRCh38
rs1043370973 Chr.2: 106805476-106805476 on Build GRCh38
rs148608067 Chr.2: 106805433-106805433 on Build GRCh38
rs531009326 Chr.2: 106805363-106805363 on Build GRCh38
rs921592934 Chr.2: 106805405-106805405 on Build GRCh38
rs528424593 Chr.2: 106805712-106805712 on Build GRCh38
rs568090156 Chr.2: 106805405-106805406 on Build GRCh38
rs1025295601 Chr.2: 106805393-106805393 on Build GRCh38
rs1056743294 Chr.2: 106805662-106805662 on Build GRCh38
rs1001851020 Chr.2: 106805528-106805528 on Build GRCh38
rs1044838663 Chr.2: 106805722-106805722 on Build GRCh38
rs959343365 Chr.2: 106805600-106805600 on Build GRCh38
rs1010050777 Chr.2: 106805464-106805464 on Build GRCh38
rs1038639939 Chr.2: 106805394-106805394 on Build GRCh38

View Full Product Details