Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2708949-2709448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMZ1
Entrez Gene ID
155185
Gene Name
archaelysin family metallopeptidase 1
Gene Aliases
-
UniGene
Hs.42221
Gene Chromosome Location
Chr.7: 2679522-2767486 on Build GRCh38
Transcripts
NM_001284355.2 NM_001321766.1 NM_133463.3 XM_011515150.2 XM_011515151.2 XM_017011774.1 XM_017011775.1 XM_017011776.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1450192 c.610G>A p.V204I Chr.7: 2709083-2709083 on Build GRCh38
3637754 c.602-2A>G p.? Chr.7: 2709073-2709073 on Build GRCh38
5601015 c.637G>A p.E213K Chr.7: 2709110-2709110 on Build GRCh38
1450191 c.609C>T p.G203G Chr.7: 2709082-2709082 on Build GRCh38
3674910 c.725G>A p.G242D Chr.7: 2709198-2709198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772788685 Chr.7: 2709237-2709237 on Build GRCh38
rs375716276 Chr.7: 2709266-2709266 on Build GRCh38
rs999550677 Chr.7: 2709099-2709099 on Build GRCh38
rs918027721 Chr.7: 2709388-2709388 on Build GRCh38
rs773929996 Chr.7: 2709167-2709167 on Build GRCh38
rs1013476923 Chr.7: 2709204-2709204 on Build GRCh38
rs375770353 Chr.7: 2709325-2709325 on Build GRCh38
rs759522759 Chr.7: 2709043-2709044 on Build GRCh38
rs1048744623 Chr.7: 2709299-2709299 on Build GRCh38
rs527501031 Chr.7: 2709127-2709127 on Build GRCh38
rs768713874 Chr.7: 2709032-2709032 on Build GRCh38
rs923669608 Chr.7: 2709353-2709353 on Build GRCh38
rs371443958 Chr.7: 2709186-2709186 on Build GRCh38
rs116695170 Chr.7: 2709014-2709014 on Build GRCh38
rs555387120 Chr.7: 2709139-2709139 on Build GRCh38
rs1022332852 Chr.7: 2709013-2709013 on Build GRCh38
rs574231697 Chr.7: 2709381-2709381 on Build GRCh38
rs976338706 Chr.7: 2709261-2709261 on Build GRCh38
rs200567828 Chr.7: 2709072-2709072 on Build GRCh38
rs773255375 Chr.7: 2709035-2709035 on Build GRCh38
rs575374021 Chr.7: 2709024-2709024 on Build GRCh38
rs540537228 Chr.7: 2709061-2709061 on Build GRCh38
rs750923034 Chr.7: 2709102-2709102 on Build GRCh38
rs754341337 Chr.7: 2709180-2709180 on Build GRCh38
rs749616241 Chr.7: 2709106-2709106 on Build GRCh38
rs778661364 Chr.7: 2709264-2709264 on Build GRCh38
rs754438467 Chr.7: 2709187-2709189 on Build GRCh38
rs146152915 Chr.7: 2709310-2709310 on Build GRCh38
rs188678758 Chr.7: 2709059-2709059 on Build GRCh38
rs776236822 Chr.7: 2709274-2709274 on Build GRCh38
rs779141297 Chr.7: 2709116-2709116 on Build GRCh38
rs752902440 Chr.7: 2709191-2709191 on Build GRCh38
rs144574079 Chr.7: 2709064-2709065 on Build GRCh38
rs949430948 Chr.7: 2709293-2709293 on Build GRCh38
rs979712957 Chr.7: 2709331-2709331 on Build GRCh38
rs779776373 Chr.7: 2709270-2709271 on Build GRCh38
rs59438885 Chr.7: 2709124-2709124 on Build GRCh38
rs931153834 Chr.7: 2709298-2709298 on Build GRCh38
rs750207124 Chr.7: 2709178-2709178 on Build GRCh38
rs568878640 Chr.7: 2709313-2709313 on Build GRCh38
rs1029239794 Chr.7: 2709319-2709319 on Build GRCh38
rs763665494 Chr.7: 2709125-2709125 on Build GRCh38
rs969206171 Chr.7: 2709045-2709045 on Build GRCh38
rs763279392 Chr.7: 2709255-2709255 on Build GRCh38
rs377323696 Chr.7: 2709252-2709252 on Build GRCh38
rs925358685 Chr.7: 2709269-2709269 on Build GRCh38
rs368696591 Chr.7: 2709128-2709128 on Build GRCh38
rs200587200 Chr.7: 2709263-2709263 on Build GRCh38
rs958953633 Chr.7: 2709321-2709321 on Build GRCh38
rs757734454 Chr.7: 2709056-2709056 on Build GRCh38
rs764987917 Chr.7: 2709049-2709049 on Build GRCh38
rs772735714 Chr.7: 2709174-2709174 on Build GRCh38
rs749039717 Chr.7: 2709158-2709158 on Build GRCh38
rs777125703 Chr.7: 2709054-2709054 on Build GRCh38
rs745482707 Chr.7: 2709026-2709026 on Build GRCh38
rs58726146 Chr.7: 2709169-2709169 on Build GRCh38
rs931889265 Chr.7: 2709415-2709415 on Build GRCh38
rs767724087 Chr.7: 2709364-2709364 on Build GRCh38
rs781378543 Chr.7: 2709025-2709025 on Build GRCh38
rs139942397 Chr.7: 2709327-2709327 on Build GRCh38
rs150799683 Chr.7: 2709153-2709153 on Build GRCh38
rs6960470 Chr.7: 2709424-2709424 on Build GRCh38
rs143141138 Chr.7: 2709094-2709094 on Build GRCh38
rs200752635 Chr.7: 2709131-2709131 on Build GRCh38
rs949397975 Chr.7: 2709396-2709396 on Build GRCh38
rs554394856 Chr.7: 2709380-2709380 on Build GRCh38
rs777970260 Chr.7: 2709141-2709141 on Build GRCh38
rs752936652 Chr.7: 2709156-2709157 on Build GRCh38
rs749302796 Chr.7: 2709179-2709180 on Build GRCh38
rs567444118 Chr.7: 2709303-2709303 on Build GRCh38
rs201824612 Chr.7: 2709117-2709117 on Build GRCh38
rs114022522 Chr.7: 2709030-2709030 on Build GRCh38
rs757337038 Chr.7: 2709208-2709208 on Build GRCh38
rs57805268 Chr.7: 2708992-2708992 on Build GRCh38
rs752394822 Chr.7: 2709050-2709050 on Build GRCh38
rs752146808 Chr.7: 2709055-2709055 on Build GRCh38
rs75195869 Chr.7: 2709210-2709210 on Build GRCh38
rs976742061 Chr.7: 2709163-2709163 on Build GRCh38
rs1032480303 Chr.7: 2709337-2709337 on Build GRCh38
rs373885832 Chr.7: 2709254-2709254 on Build GRCh38
rs887811963 Chr.7: 2709423-2709423 on Build GRCh38
rs765112468 Chr.7: 2709086-2709086 on Build GRCh38
rs201597646 Chr.7: 2709196-2709196 on Build GRCh38
rs780317642 Chr.7: 2709062-2709062 on Build GRCh38
rs944523090 Chr.7: 2709265-2709265 on Build GRCh38
rs775791774 Chr.7: 2709286-2709286 on Build GRCh38
rs376387894 Chr.7: 2709040-2709040 on Build GRCh38
rs376896309 Chr.7: 2709170-2709170 on Build GRCh38
rs58748470 Chr.7: 2709267-2709267 on Build GRCh38
rs138200553 Chr.7: 2709176-2709176 on Build GRCh38
rs1052341733 Chr.7: 2709100-2709100 on Build GRCh38
rs370587760 Chr.7: 2709253-2709253 on Build GRCh38
rs749076310 Chr.7: 2709028-2709028 on Build GRCh38
rs935365472 Chr.7: 2709280-2709280 on Build GRCh38
rs1040415763 Chr.7: 2708972-2708972 on Build GRCh38
rs35034694 Chr.7: 2709402-2709403 on Build GRCh38
rs770789436 Chr.7: 2709082-2709082 on Build GRCh38
rs374537415 Chr.7: 2709155-2709155 on Build GRCh38
rs753754356 Chr.7: 2709260-2709260 on Build GRCh38
rs755943997 Chr.7: 2709177-2709178 on Build GRCh38
rs149639626 Chr.7: 2709079-2709079 on Build GRCh38
rs935314552 Chr.7: 2709178-2709178 on Build GRCh38
rs990440865 Chr.7: 2709214-2709214 on Build GRCh38
rs758747166 Chr.7: 2709166-2709176 on Build GRCh38
rs756525635 Chr.7: 2709103-2709103 on Build GRCh38
rs767911544 Chr.7: 2709098-2709098 on Build GRCh38
rs770416776 Chr.7: 2709240-2709240 on Build GRCh38
rs900829564 Chr.7: 2709311-2709311 on Build GRCh38
rs113338257 Chr.7: 2709051-2709051 on Build GRCh38
rs780370603 Chr.7: 2709104-2709104 on Build GRCh38
rs755181336 Chr.7: 2709115-2709115 on Build GRCh38
rs770470084 Chr.7: 2709273-2709273 on Build GRCh38
rs777054420 Chr.7: 2709245-2709245 on Build GRCh38
rs977898702 Chr.7: 2709333-2709333 on Build GRCh38
rs764437978 Chr.7: 2709170-2709176 on Build GRCh38
rs376694807 Chr.7: 2709095-2709095 on Build GRCh38
rs770348171 Chr.7: 2709285-2709285 on Build GRCh38
rs775629125 Chr.7: 2709029-2709029 on Build GRCh38
rs745313964 Chr.7: 2709283-2709283 on Build GRCh38
rs798568 Chr.7: 2709365-2709365 on Build GRCh38
rs142796061 Chr.7: 2709309-2709309 on Build GRCh38
rs369383324 Chr.7: 2709048-2709048 on Build GRCh38
rs779795364 Chr.7: 2709180-2709180 on Build GRCh38
rs760057391 Chr.7: 2709247-2709247 on Build GRCh38
rs746201767 Chr.7: 2709027-2709027 on Build GRCh38
rs775332053 Chr.7: 2709088-2709088 on Build GRCh38
rs753600043 Chr.7: 2709046-2709046 on Build GRCh38
rs140523812 Chr.7: 2709133-2709133 on Build GRCh38
rs144255482 Chr.7: 2709083-2709083 on Build GRCh38
rs571724342 Chr.7: 2709363-2709363 on Build GRCh38
rs757467508 Chr.7: 2709258-2709258 on Build GRCh38
rs1013424703 Chr.7: 2709111-2709111 on Build GRCh38
rs768491440 Chr.7: 2709162-2709162 on Build GRCh38
rs766303750 Chr.7: 2709041-2709041 on Build GRCh38
rs905135163 Chr.7: 2709297-2709297 on Build GRCh38
rs999416525 Chr.7: 2709021-2709021 on Build GRCh38
rs778091933 Chr.7: 2709202-2709202 on Build GRCh38
rs896493188 Chr.7: 2709192-2709192 on Build GRCh38
rs776768133 Chr.7: 2709121-2709121 on Build GRCh38
rs981213991 Chr.7: 2709418-2709418 on Build GRCh38
rs747045351 Chr.7: 2709143-2709143 on Build GRCh38
rs557784148 Chr.7: 2709330-2709330 on Build GRCh38
rs760124282 Chr.7: 2709175-2709175 on Build GRCh38
rs765613498 Chr.7: 2709184-2709184 on Build GRCh38
rs200226968 Chr.7: 2709042-2709042 on Build GRCh38
rs772189961 Chr.7: 2709118-2709118 on Build GRCh38
rs766826076 Chr.7: 2709129-2709129 on Build GRCh38
rs763477359 Chr.7: 2709294-2709294 on Build GRCh38
rs760247065 Chr.7: 2709238-2709238 on Build GRCh38
rs560863383 Chr.7: 2709067-2709067 on Build GRCh38
rs996571033 Chr.7: 2709005-2709005 on Build GRCh38
rs769751094 Chr.7: 2709123-2709123 on Build GRCh38
rs536989147 Chr.7: 2709416-2709416 on Build GRCh38
rs142146201 Chr.7: 2709134-2709134 on Build GRCh38
rs547609451 Chr.7: 2709270-2709270 on Build GRCh38
rs749495362 Chr.7: 2709063-2709063 on Build GRCh38
rs143291176 Chr.7: 2709222-2709222 on Build GRCh38
rs768381532 Chr.7: 2709221-2709221 on Build GRCh38
rs562498150 Chr.7: 2709226-2709226 on Build GRCh38
rs751825267 Chr.7: 2709257-2709257 on Build GRCh38

View Full Product Details