Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 43103677-43104130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POMK
Entrez Gene ID
84197
Gene Name
protein-O-mannose kinase
Gene Aliases
MDDGA12, MDDGC12, SGK196
UniGene
Hs.491646
Gene Chromosome Location
Chr.8: 43093506-43123180 on Build GRCh38
Transcripts
NM_001277971.1 NM_032237.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7247014 c.134C>G p.P45R Chr.8: 43103682-43103682 on Build GRCh38
5431496 c.195G>A p.M65I Chr.8: 43103743-43103743 on Build GRCh38
144081 c.208C>T p.P70S Chr.8: 43103756-43103756 on Build GRCh38
7102996 c.231G>A p.L77L Chr.8: 43103779-43103779 on Build GRCh38
2962137 c.162C>T p.H54H Chr.8: 43103710-43103710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147680806 Chr.8: 43103770-43103770 on Build GRCh38
rs918014375 Chr.8: 43103993-43103993 on Build GRCh38
rs145831713 Chr.8: 43103712-43103712 on Build GRCh38
rs376305406 Chr.8: 43103838-43103838 on Build GRCh38
rs758336306 Chr.8: 43103807-43103807 on Build GRCh38
rs1035713559 Chr.8: 43103711-43103711 on Build GRCh38
rs781412137 Chr.8: 43103700-43103700 on Build GRCh38
rs181979471 Chr.8: 43103850-43103850 on Build GRCh38
rs779527053 Chr.8: 43103995-43103995 on Build GRCh38
rs201095843 Chr.8: 43103805-43103805 on Build GRCh38
rs974635918 Chr.8: 43103963-43103963 on Build GRCh38
rs148978342 Chr.8: 43103726-43103726 on Build GRCh38
rs774951327 Chr.8: 43103857-43103857 on Build GRCh38
rs746739561 Chr.8: 43103748-43103748 on Build GRCh38
rs748059884 Chr.8: 43103867-43103867 on Build GRCh38
rs778749095 Chr.8: 43103731-43103731 on Build GRCh38
rs188920070 Chr.8: 43103767-43103767 on Build GRCh38
rs150669524 Chr.8: 43103919-43103919 on Build GRCh38
rs908777905 Chr.8: 43103930-43103930 on Build GRCh38
rs573433327 Chr.8: 43104039-43104039 on Build GRCh38
rs138772640 Chr.8: 43103921-43103921 on Build GRCh38
rs755313239 Chr.8: 43103798-43103798 on Build GRCh38
rs767010506 Chr.8: 43103853-43103853 on Build GRCh38
rs1000413453 Chr.8: 43103874-43103874 on Build GRCh38
rs34936819 Chr.8: 43104047-43104047 on Build GRCh38
rs771734462 Chr.8: 43103929-43103929 on Build GRCh38
rs571809721 Chr.8: 43103959-43103959 on Build GRCh38
rs763543073 Chr.8: 43103789-43103789 on Build GRCh38
rs773352984 Chr.8: 43104086-43104086 on Build GRCh38
rs55636430 Chr.8: 43103913-43103913 on Build GRCh38
rs757819424 Chr.8: 43103756-43103756 on Build GRCh38
rs370978230 Chr.8: 43103804-43103804 on Build GRCh38
rs553438011 Chr.8: 43104007-43104007 on Build GRCh38
rs555746391 Chr.8: 43104089-43104089 on Build GRCh38
rs759851830 Chr.8: 43103861-43103861 on Build GRCh38
rs142346196 Chr.8: 43103793-43103793 on Build GRCh38
rs142894606 Chr.8: 43103732-43103732 on Build GRCh38
rs778835046 Chr.8: 43103829-43103829 on Build GRCh38
rs964120858 Chr.8: 43103886-43103886 on Build GRCh38
rs775260814 Chr.8: 43103745-43103745 on Build GRCh38
rs752925552 Chr.8: 43103802-43103802 on Build GRCh38
rs56224218 Chr.8: 43103952-43103952 on Build GRCh38
rs926673143 Chr.8: 43103706-43103706 on Build GRCh38
rs938092349 Chr.8: 43103966-43103966 on Build GRCh38
rs544817446 Chr.8: 43104070-43104072 on Build GRCh38
rs758397612 Chr.8: 43103879-43103880 on Build GRCh38
rs779908103 Chr.8: 43103822-43103822 on Build GRCh38
rs775727817 Chr.8: 43103875-43103875 on Build GRCh38
rs925752217 Chr.8: 43103813-43103813 on Build GRCh38
rs746797662 Chr.8: 43103823-43103823 on Build GRCh38
rs1020334634 Chr.8: 43104028-43104028 on Build GRCh38
rs772847115 Chr.8: 43103845-43103845 on Build GRCh38
rs535809302 Chr.8: 43104068-43104068 on Build GRCh38
rs375189312 Chr.8: 43103851-43103851 on Build GRCh38
rs773908820 Chr.8: 43103784-43103784 on Build GRCh38
rs779331401 Chr.8: 43103839-43103839 on Build GRCh38
rs371623050 Chr.8: 43103873-43103873 on Build GRCh38
rs201953159 Chr.8: 43103729-43103729 on Build GRCh38
rs535321605 Chr.8: 43103992-43103992 on Build GRCh38
rs897842541 Chr.8: 43103912-43103912 on Build GRCh38
rs1009832273 Chr.8: 43104004-43104004 on Build GRCh38
rs202005139 Chr.8: 43103862-43103862 on Build GRCh38
rs751981817 Chr.8: 43103795-43103795 on Build GRCh38
rs967023100 Chr.8: 43104036-43104036 on Build GRCh38
rs747811554 Chr.8: 43103837-43103837 on Build GRCh38
rs962946763 Chr.8: 43103939-43103939 on Build GRCh38
rs1036748111 Chr.8: 43103883-43103883 on Build GRCh38
rs767655500 Chr.8: 43103799-43103799 on Build GRCh38
rs769643751 Chr.8: 43103965-43103965 on Build GRCh38
rs776184446 Chr.8: 43103834-43103834 on Build GRCh38
rs372826172 Chr.8: 43103718-43103718 on Build GRCh38
rs764770753 Chr.8: 43103783-43103783 on Build GRCh38
rs752918721 Chr.8: 43103863-43103863 on Build GRCh38
rs576208272 Chr.8: 43104099-43104099 on Build GRCh38
rs534169939 Chr.8: 43103968-43103968 on Build GRCh38
rs5024011 Chr.8: 43103864-43103864 on Build GRCh38

View Full Product Details