Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 43122456-43122950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POMK
Entrez Gene ID
84197
Gene Name
protein-O-mannose kinase
Gene Aliases
MDDGA12, MDDGC12, SGK196
UniGene
Hs.491646
Gene Chromosome Location
Chr.8: 43093506-43123180 on Build GRCh38
Transcripts
NM_001277971.1 NM_032237.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100109 c.742G>A p.E248K Chr.8: 43122566-43122566 on Build GRCh38
1100105 c.687C>T p.D229D Chr.8: 43122511-43122511 on Build GRCh38
6275395 c.699G>T p.L233L Chr.8: 43122523-43122523 on Build GRCh38
1100107 c.714C>A p.S238S Chr.8: 43122538-43122538 on Build GRCh38
6180883 c.859G>C p.D287H Chr.8: 43122683-43122683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200555259 Chr.8: 43122837-43122837 on Build GRCh38
rs34715198 Chr.8: 43122584-43122584 on Build GRCh38
rs757600130 Chr.8: 43122657-43122657 on Build GRCh38
rs56282240 Chr.8: 43122706-43122706 on Build GRCh38
rs1029880562 Chr.8: 43122553-43122553 on Build GRCh38
rs781318268 Chr.8: 43122885-43122885 on Build GRCh38
rs762404960 Chr.8: 43122848-43122848 on Build GRCh38
rs755763171 Chr.8: 43122887-43122887 on Build GRCh38
rs757744253 Chr.8: 43122734-43122734 on Build GRCh38
rs758695817 Chr.8: 43122760-43122760 on Build GRCh38
rs750736847 Chr.8: 43122750-43122750 on Build GRCh38
rs1005243869 Chr.8: 43122853-43122853 on Build GRCh38
rs772331741 Chr.8: 43122680-43122680 on Build GRCh38
rs756551345 Chr.8: 43122529-43122529 on Build GRCh38
rs771820969 Chr.8: 43122928-43122928 on Build GRCh38
rs899980411 Chr.8: 43122778-43122778 on Build GRCh38
rs747083630 Chr.8: 43122789-43122789 on Build GRCh38
rs754174652 Chr.8: 43122631-43122631 on Build GRCh38
rs778767217 Chr.8: 43122915-43122915 on Build GRCh38
rs758784196 Chr.8: 43122873-43122873 on Build GRCh38
rs772247645 Chr.8: 43122558-43122558 on Build GRCh38
rs939368243 Chr.8: 43122521-43122521 on Build GRCh38
rs777615691 Chr.8: 43122791-43122791 on Build GRCh38
rs749227228 Chr.8: 43122795-43122795 on Build GRCh38
rs749588884 Chr.8: 43122836-43122837 on Build GRCh38
rs142479040 Chr.8: 43122556-43122556 on Build GRCh38
rs750830493 Chr.8: 43122871-43122871 on Build GRCh38
rs1029874324 Chr.8: 43122792-43122792 on Build GRCh38
rs746065285 Chr.8: 43122674-43122674 on Build GRCh38
rs745687598 Chr.8: 43122916-43122916 on Build GRCh38
rs546931054 Chr.8: 43122803-43122803 on Build GRCh38
rs762691852 Chr.8: 43122701-43122701 on Build GRCh38
rs769693529 Chr.8: 43122573-43122573 on Build GRCh38
rs190646380 Chr.8: 43122685-43122685 on Build GRCh38
rs1022266326 Chr.8: 43122486-43122486 on Build GRCh38
rs947882915 Chr.8: 43122599-43122599 on Build GRCh38
rs755062802 Chr.8: 43122880-43122880 on Build GRCh38
rs765678572 Chr.8: 43122738-43122738 on Build GRCh38
rs1043858087 Chr.8: 43122759-43122759 on Build GRCh38
rs1014642009 Chr.8: 43122571-43122571 on Build GRCh38
rs397509386 Chr.8: 43122597-43122597 on Build GRCh38
rs762504780 Chr.8: 43122498-43122498 on Build GRCh38
rs771065242 Chr.8: 43122545-43122545 on Build GRCh38
rs370946940 Chr.8: 43122732-43122732 on Build GRCh38
rs199892708 Chr.8: 43122900-43122900 on Build GRCh38
rs148949414 Chr.8: 43122538-43122538 on Build GRCh38
rs751085294 Chr.8: 43122511-43122511 on Build GRCh38
rs375850409 Chr.8: 43122766-43122766 on Build GRCh38
rs145931566 Chr.8: 43122487-43122487 on Build GRCh38
rs780273144 Chr.8: 43122684-43122684 on Build GRCh38
rs772885359 Chr.8: 43122574-43122574 on Build GRCh38
rs773155264 Chr.8: 43122688-43122688 on Build GRCh38
rs761639735 Chr.8: 43122566-43122566 on Build GRCh38
rs749293857 Chr.8: 43122894-43122894 on Build GRCh38
rs200742772 Chr.8: 43122528-43122528 on Build GRCh38
rs751775266 Chr.8: 43122876-43122876 on Build GRCh38
rs754439246 Chr.8: 43122520-43122520 on Build GRCh38
rs767081710 Chr.8: 43122725-43122725 on Build GRCh38
rs528307346 Chr.8: 43122731-43122731 on Build GRCh38
rs753305627 Chr.8: 43122616-43122616 on Build GRCh38
rs886041651 Chr.8: 43122741-43122742 on Build GRCh38
rs543448796 Chr.8: 43122620-43122620 on Build GRCh38
rs1016660986 Chr.8: 43122895-43122895 on Build GRCh38
rs6990761 Chr.8: 43122710-43122710 on Build GRCh38
rs900073683 Chr.8: 43122547-43122547 on Build GRCh38
rs765987803 Chr.8: 43122505-43122505 on Build GRCh38
rs751177038 Chr.8: 43122590-43122590 on Build GRCh38
rs755047768 Chr.8: 43122790-43122790 on Build GRCh38
rs113361507 Chr.8: 43122830-43122830 on Build GRCh38
rs33920561 Chr.8: 43122726-43122726 on Build GRCh38
rs779226611 Chr.8: 43122550-43122550 on Build GRCh38
rs201232766 Chr.8: 43122583-43122583 on Build GRCh38
rs766542633 Chr.8: 43122874-43122874 on Build GRCh38
rs759091832 Chr.8: 43122598-43122598 on Build GRCh38
rs377648307 Chr.8: 43122802-43122802 on Build GRCh38
rs35104071 Chr.8: 43122539-43122539 on Build GRCh38
rs564512395 Chr.8: 43122711-43122711 on Build GRCh38
rs775857497 Chr.8: 43122803-43122803 on Build GRCh38
rs199756983 Chr.8: 43122729-43122729 on Build GRCh38
rs1018638122 Chr.8: 43122512-43122512 on Build GRCh38
rs36071326 Chr.8: 43122619-43122619 on Build GRCh38
rs769629128 Chr.8: 43122686-43122686 on Build GRCh38
rs760140163 Chr.8: 43122834-43122834 on Build GRCh38
rs776753930 Chr.8: 43122561-43122561 on Build GRCh38
rs996987445 Chr.8: 43122781-43122781 on Build GRCh38
rs765772229 Chr.8: 43122849-43122849 on Build GRCh38
rs868057078 Chr.8: 43122644-43122644 on Build GRCh38
rs779318258 Chr.8: 43122668-43122668 on Build GRCh38
rs777058315 Chr.8: 43122847-43122847 on Build GRCh38
rs771728909 Chr.8: 43122827-43122827 on Build GRCh38
rs370495356 Chr.8: 43122813-43122813 on Build GRCh38
rs185390123 Chr.8: 43122525-43122525 on Build GRCh38
rs946823916 Chr.8: 43122743-43122743 on Build GRCh38

View Full Product Details