Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4834049-4834548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCL1
Entrez Gene ID
10171
Gene Name
RNA terminal phosphate cyclase like 1
Gene Aliases
RNAC, RPCL1
UniGene
Hs.194121
Gene Chromosome Location
Chr.9: 4792834-4861077 on Build GRCh38
Transcripts
NM_001286699.1 NM_001286700.1 NM_005772.4 XM_006716715.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4879342 c.517G>A p.V173M Chr.9: 4834198-4834198 on Build GRCh38
1109340 c.528C>A p.V176V Chr.9: 4834209-4834209 on Build GRCh38
5998419 c.584+2T>C p.? Chr.9: 4834267-4834267 on Build GRCh38
2777176 c.484G>A p.G162R Chr.9: 4834165-4834165 on Build GRCh38
1720237 c.506T>C p.F169S Chr.9: 4834187-4834187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113668857 Chr.9: 4834188-4834188 on Build GRCh38
rs920038809 Chr.9: 4834075-4834075 on Build GRCh38
rs999738852 Chr.9: 4834073-4834074 on Build GRCh38
rs982951603 Chr.9: 4834080-4834080 on Build GRCh38
rs769417269 Chr.9: 4834292-4834294 on Build GRCh38
rs999792100 Chr.9: 4834183-4834183 on Build GRCh38
rs369255649 Chr.9: 4834135-4834135 on Build GRCh38
rs5896097 Chr.9: 4834490-4834490 on Build GRCh38
rs141431065 Chr.9: 4834292-4834292 on Build GRCh38
rs761023720 Chr.9: 4834472-4834474 on Build GRCh38
rs750642259 Chr.9: 4834266-4834266 on Build GRCh38
rs952683335 Chr.9: 4834364-4834364 on Build GRCh38
rs1035328505 Chr.9: 4834324-4834324 on Build GRCh38
rs1031182476 Chr.9: 4834073-4834073 on Build GRCh38
rs372465798 Chr.9: 4834087-4834087 on Build GRCh38
rs918493032 Chr.9: 4834358-4834358 on Build GRCh38
rs1035380096 Chr.9: 4834499-4834499 on Build GRCh38
rs116769238 Chr.9: 4834164-4834164 on Build GRCh38
rs761996748 Chr.9: 4834252-4834252 on Build GRCh38
rs775302168 Chr.9: 4834241-4834241 on Build GRCh38
rs969401966 Chr.9: 4834245-4834245 on Build GRCh38
rs10217603 Chr.9: 4834295-4834295 on Build GRCh38
rs760192282 Chr.9: 4834213-4834213 on Build GRCh38
rs770352145 Chr.9: 4834219-4834219 on Build GRCh38
rs1041122471 Chr.9: 4834403-4834403 on Build GRCh38
rs995386057 Chr.9: 4834471-4834471 on Build GRCh38
rs762468579 Chr.9: 4834142-4834142 on Build GRCh38
rs771902545 Chr.9: 4834208-4834208 on Build GRCh38
rs780566380 Chr.9: 4834169-4834169 on Build GRCh38
rs457287 Chr.9: 4834394-4834394 on Build GRCh38
rs905327504 Chr.9: 4834460-4834460 on Build GRCh38
rs757002349 Chr.9: 4834277-4834277 on Build GRCh38
rs570779849 Chr.9: 4834471-4834471 on Build GRCh38
rs201288279 Chr.9: 4834312-4834312 on Build GRCh38
rs532877046 Chr.9: 4834209-4834209 on Build GRCh38
rs781217225 Chr.9: 4834163-4834163 on Build GRCh38
rs116589736 Chr.9: 4834150-4834150 on Build GRCh38
rs879031460 Chr.9: 4834243-4834243 on Build GRCh38
rs183144815 Chr.9: 4834411-4834411 on Build GRCh38
rs772821344 Chr.9: 4834131-4834131 on Build GRCh38
rs375761110 Chr.9: 4834095-4834095 on Build GRCh38
rs1023997865 Chr.9: 4834204-4834204 on Build GRCh38
rs191272144 Chr.9: 4834390-4834390 on Build GRCh38
rs765010808 Chr.9: 4834234-4834234 on Build GRCh38
rs138076058 Chr.9: 4834148-4834148 on Build GRCh38
rs115862235 Chr.9: 4834151-4834151 on Build GRCh38
rs750168003 Chr.9: 4834264-4834264 on Build GRCh38
rs934564958 Chr.9: 4834492-4834492 on Build GRCh38
rs148106832 Chr.9: 4834299-4834300 on Build GRCh38
rs767726268 Chr.9: 4834254-4834254 on Build GRCh38
rs753750304 Chr.9: 4834137-4834137 on Build GRCh38
rs983966881 Chr.9: 4834379-4834379 on Build GRCh38
rs951394814 Chr.9: 4834490-4834490 on Build GRCh38
rs762704789 Chr.9: 4834292-4834293 on Build GRCh38
rs148074533 Chr.9: 4834165-4834165 on Build GRCh38
rs755583736 Chr.9: 4834296-4834296 on Build GRCh38
rs1026906636 Chr.9: 4834479-4834479 on Build GRCh38
rs187693726 Chr.9: 4834332-4834332 on Build GRCh38
rs924951085 Chr.9: 4834416-4834416 on Build GRCh38
rs942806793 Chr.9: 4834396-4834396 on Build GRCh38
rs982835603 Chr.9: 4834303-4834303 on Build GRCh38
rs749760183 Chr.9: 4834176-4834176 on Build GRCh38
rs150833689 Chr.9: 4834079-4834079 on Build GRCh38
rs774315982 Chr.9: 4834296-4834297 on Build GRCh38
rs546480390 Chr.9: 4834447-4834447 on Build GRCh38
rs749612149 Chr.9: 4834099-4834099 on Build GRCh38
rs10974805 Chr.9: 4834420-4834420 on Build GRCh38
rs761568755 Chr.9: 4834114-4834114 on Build GRCh38
rs374022520 Chr.9: 4834227-4834227 on Build GRCh38
rs78943484 Chr.9: 4834438-4834438 on Build GRCh38
rs931671056 Chr.9: 4834143-4834143 on Build GRCh38
rs554408302 Chr.9: 4834291-4834292 on Build GRCh38
rs975671236 Chr.9: 4834365-4834365 on Build GRCh38
rs780823579 Chr.9: 4834474-4834474 on Build GRCh38
rs535294636 Chr.9: 4834498-4834498 on Build GRCh38
rs1001255248 Chr.9: 4834462-4834462 on Build GRCh38
rs756150959 Chr.9: 4834275-4834275 on Build GRCh38
rs139261340 Chr.9: 4834133-4834133 on Build GRCh38
rs1040371802 Chr.9: 4834110-4834110 on Build GRCh38
rs769170839 Chr.9: 4834107-4834107 on Build GRCh38
rs751208085 Chr.9: 4834159-4834159 on Build GRCh38
rs1044980529 Chr.9: 4834448-4834448 on Build GRCh38
rs751487831 Chr.9: 4834473-4834474 on Build GRCh38
rs538540542 Chr.9: 4834389-4834389 on Build GRCh38
rs75298111 Chr.9: 4834459-4834459 on Build GRCh38
rs377100122 Chr.9: 4834113-4834113 on Build GRCh38
rs2296769 Chr.9: 4834299-4834299 on Build GRCh38
rs1004228796 Chr.9: 4834520-4834520 on Build GRCh38
rs552777359 Chr.9: 4834086-4834086 on Build GRCh38
rs747774431 Chr.9: 4834191-4834191 on Build GRCh38
rs1041068552 Chr.9: 4834180-4834180 on Build GRCh38
rs965946353 Chr.9: 4834353-4834353 on Build GRCh38
rs769081196 Chr.9: 4834182-4834182 on Build GRCh38
rs757541031 Chr.9: 4834162-4834162 on Build GRCh38
rs776575779 Chr.9: 4834224-4834224 on Build GRCh38
rs935039662 Chr.9: 4834417-4834417 on Build GRCh38
rs935298481 Chr.9: 4834412-4834412 on Build GRCh38

View Full Product Details