Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113589533-113590055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HABP2
Entrez Gene ID
3026
Gene Name
hyaluronan binding protein 2
Gene Aliases
FSAP, HABP, HGFAL, NMTC5, PHBP
UniGene
Hs.422542
Gene Chromosome Location
Chr.10: 113550831-113589602 on Build GRCh38
Transcripts
NM_001177660.2 NM_004132.4 NM_001261463.1 NM_001322945.1 NM_006175.4 NM_198060.3 XM_005269864.2 XM_005269865.2 XM_005269867.2 XM_006717870.2 XM_011539832.2

Gene Symbol
NRAP
Entrez Gene ID
4892
Gene Name
nebulin related anchoring protein
Gene Aliases
N-RAP
UniGene
Hs.268788
Gene Chromosome Location
Chr.10: 113588824-113664070 on Build GRCh38
Transcripts
NM_001177660.2 NM_004132.4 NM_001261463.1 NM_001322945.1 NM_006175.4 NM_198060.3 XM_005269864.2 XM_005269865.2 XM_005269867.2 XM_006717870.2 XM_011539832.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6729389 c.4978A>G p.N1660D Chr.10: 113589776-113589776 on Build GRCh38
1217808 c.5027A>G p.E1676G Chr.10: 113589727-113589727 on Build GRCh38
4518039 c.5031_5032GG>AA p.M1677_A1678>IT Chr.10: 113589722-113589723 on Build GRCh38
6729392 c.5071G>T p.G1691C Chr.10: 113589683-113589683 on Build GRCh38
4454498 c.5082A>T p.G1694G Chr.10: 113589672-113589672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937169150 Chr.10: 113589571-113589571 on Build GRCh38
rs367726303 Chr.10: 113589846-113589846 on Build GRCh38
rs967796564 Chr.10: 113589892-113589892 on Build GRCh38
rs11196388 Chr.10: 113589639-113589639 on Build GRCh38
rs368972487 Chr.10: 113589651-113589651 on Build GRCh38
rs768678100 Chr.10: 113589750-113589750 on Build GRCh38
rs968758563 Chr.10: 113589825-113589825 on Build GRCh38
rs767490943 Chr.10: 113590003-113590003 on Build GRCh38
rs113411292 Chr.10: 113589715-113589715 on Build GRCh38
rs747330406 Chr.10: 113589658-113589658 on Build GRCh38
rs11575792 Chr.10: 113589976-113589976 on Build GRCh38
rs750606534 Chr.10: 113589562-113589562 on Build GRCh38
rs145862371 Chr.10: 113589740-113589740 on Build GRCh38
rs781496592 Chr.10: 113589726-113589726 on Build GRCh38
rs370372596 Chr.10: 113589643-113589643 on Build GRCh38
rs11575791 Chr.10: 113589932-113589932 on Build GRCh38
rs879182838 Chr.10: 113589729-113589729 on Build GRCh38
rs201125144 Chr.10: 113589719-113589719 on Build GRCh38
rs1052205771 Chr.10: 113589897-113589897 on Build GRCh38
rs878901087 Chr.10: 113589683-113589683 on Build GRCh38
rs11575789 Chr.10: 113589665-113589665 on Build GRCh38
rs548663497 Chr.10: 113589963-113589963 on Build GRCh38
rs1005679742 Chr.10: 113589590-113589590 on Build GRCh38
rs778522668 Chr.10: 113589641-113589641 on Build GRCh38
rs11575790 Chr.10: 113589708-113589708 on Build GRCh38
rs1002510900 Chr.10: 113590004-113590004 on Build GRCh38
rs751181314 Chr.10: 113589817-113589817 on Build GRCh38
rs201572004 Chr.10: 113589613-113589613 on Build GRCh38
rs544482064 Chr.10: 113590010-113590010 on Build GRCh38
rs4918857 Chr.10: 113589703-113589703 on Build GRCh38
rs878998890 Chr.10: 113589785-113589785 on Build GRCh38
rs113659972 Chr.10: 113589911-113589911 on Build GRCh38
rs774507330 Chr.10: 113589697-113589697 on Build GRCh38
rs747639408 Chr.10: 113589650-113589650 on Build GRCh38
rs200987782 Chr.10: 113589725-113589725 on Build GRCh38
rs1040721338 Chr.10: 113589713-113589713 on Build GRCh38
rs577870560 Chr.10: 113589797-113589797 on Build GRCh38
rs766438314 Chr.10: 113589879-113589879 on Build GRCh38
rs769091074 Chr.10: 113589757-113589758 on Build GRCh38
rs774615104 Chr.10: 113589751-113589751 on Build GRCh38
rs376972937 Chr.10: 113589620-113589620 on Build GRCh38
rs112213268 Chr.10: 113589798-113589798 on Build GRCh38
rs987842522 Chr.10: 113589687-113589687 on Build GRCh38
rs759370424 Chr.10: 113589954-113589955 on Build GRCh38
rs923144114 Chr.10: 113589995-113589995 on Build GRCh38
rs969637898 Chr.10: 113589619-113589619 on Build GRCh38
rs149864957 Chr.10: 113589716-113589716 on Build GRCh38
rs576102215 Chr.10: 113590025-113590025 on Build GRCh38
rs397737729 Chr.10: 113589642-113589642 on Build GRCh38
rs371816136 Chr.10: 113589847-113589847 on Build GRCh38
rs202079471 Chr.10: 113589773-113589773 on Build GRCh38
rs913580702 Chr.10: 113589886-113589886 on Build GRCh38
rs550969861 Chr.10: 113589696-113589696 on Build GRCh38
rs755142960 Chr.10: 113589801-113589801 on Build GRCh38
rs774950232 Chr.10: 113589862-113589863 on Build GRCh38
rs770230689 Chr.10: 113589732-113589732 on Build GRCh38
rs113506975 Chr.10: 113589800-113589800 on Build GRCh38
rs1037867366 Chr.10: 113589942-113589944 on Build GRCh38
rs780931577 Chr.10: 113589805-113589805 on Build GRCh38
rs780836625 Chr.10: 113589822-113589822 on Build GRCh38
rs751592372 Chr.10: 113589880-113589880 on Build GRCh38
rs765105423 Chr.10: 113589712-113589712 on Build GRCh38
rs370742966 Chr.10: 113589695-113589695 on Build GRCh38
rs756504691 Chr.10: 113589835-113589835 on Build GRCh38
rs144506958 Chr.10: 113589721-113589721 on Build GRCh38
rs533626220 Chr.10: 113589682-113589682 on Build GRCh38
rs777477107 Chr.10: 113589718-113589718 on Build GRCh38
rs933307258 Chr.10: 113589894-113589894 on Build GRCh38
rs929215649 Chr.10: 113589996-113589996 on Build GRCh38
rs753888267 Chr.10: 113589623-113589623 on Build GRCh38
rs758411362 Chr.10: 113589828-113589828 on Build GRCh38
rs756854603 Chr.10: 113589820-113589820 on Build GRCh38
rs376381578 Chr.10: 113589833-113589833 on Build GRCh38
rs113590679 Chr.10: 113589666-113589666 on Build GRCh38
rs200399883 Chr.10: 113589730-113589730 on Build GRCh38
rs755981418 Chr.10: 113589615-113589615 on Build GRCh38
rs766330010 Chr.10: 113589704-113589704 on Build GRCh38
rs1017080064 Chr.10: 113589780-113589780 on Build GRCh38
rs574003797 Chr.10: 113589738-113589738 on Build GRCh38
rs750721103 Chr.10: 113589830-113589830 on Build GRCh38
rs551118310 Chr.10: 113589636-113589636 on Build GRCh38
rs776514915 Chr.10: 113589782-113589782 on Build GRCh38
rs777301556 Chr.10: 113589652-113589652 on Build GRCh38
rs367960069 Chr.10: 113589734-113589734 on Build GRCh38
rs199520396 Chr.10: 113589838-113589838 on Build GRCh38
rs140014381 Chr.10: 113589709-113589709 on Build GRCh38
rs573303899 Chr.10: 113589922-113589922 on Build GRCh38
rs762169922 Chr.10: 113589755-113589755 on Build GRCh38
rs991746787 Chr.10: 113589592-113589592 on Build GRCh38
rs868286955 Chr.10: 113589916-113589916 on Build GRCh38
rs763562082 Chr.10: 113589811-113589811 on Build GRCh38
rs780602056 Chr.10: 113589627-113589627 on Build GRCh38

View Full Product Details