Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25863192-25863680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAQR7
Entrez Gene ID
164091
Gene Name
progestin and adipoQ receptor family member 7
Gene Aliases
MPRA, PGLP, mSR
UniGene
Hs.523652
Gene Chromosome Location
Chr.1: 25861484-25876707 on Build GRCh38
Transcripts
NM_178422.5 XM_005245745.3 XM_005245746.3 XM_011540861.1 XM_011540862.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6737614 c.630C>T p.S210S Chr.1: 25863210-25863210 on Build GRCh38
4711086 c.264G>A p.R88R Chr.1: 25863576-25863576 on Build GRCh38
6737612 c.440C>T p.A147V Chr.1: 25863400-25863400 on Build GRCh38
4617408 c.442G>A p.V148M Chr.1: 25863398-25863398 on Build GRCh38
6737610 c.242C>T p.A81V Chr.1: 25863598-25863598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975396509 Chr.1: 25863483-25863483 on Build GRCh38
rs150642797 Chr.1: 25863397-25863398 on Build GRCh38
rs534306534 Chr.1: 25863583-25863583 on Build GRCh38
rs759761837 Chr.1: 25863291-25863291 on Build GRCh38
rs770666410 Chr.1: 25863658-25863658 on Build GRCh38
rs769014018 Chr.1: 25863229-25863229 on Build GRCh38
rs779609918 Chr.1: 25863631-25863631 on Build GRCh38
rs552087905 Chr.1: 25863399-25863399 on Build GRCh38
rs148705012 Chr.1: 25863258-25863258 on Build GRCh38
rs937878699 Chr.1: 25863633-25863633 on Build GRCh38
rs755659850 Chr.1: 25863618-25863618 on Build GRCh38
rs779474089 Chr.1: 25863342-25863342 on Build GRCh38
rs150686205 Chr.1: 25863597-25863597 on Build GRCh38
rs757664149 Chr.1: 25863630-25863630 on Build GRCh38
rs1047920841 Chr.1: 25863660-25863660 on Build GRCh38
rs61743806 Chr.1: 25863555-25863555 on Build GRCh38
rs370095439 Chr.1: 25863577-25863577 on Build GRCh38
rs772938279 Chr.1: 25863572-25863572 on Build GRCh38
rs746625276 Chr.1: 25863222-25863222 on Build GRCh38
rs754510817 Chr.1: 25863596-25863596 on Build GRCh38
rs746064957 Chr.1: 25863536-25863554 on Build GRCh38
rs138657959 Chr.1: 25863448-25863448 on Build GRCh38
rs151265646 Chr.1: 25863347-25863347 on Build GRCh38
rs747710601 Chr.1: 25863515-25863515 on Build GRCh38
rs975491122 Chr.1: 25863214-25863214 on Build GRCh38
rs757388951 Chr.1: 25863451-25863452 on Build GRCh38
rs770036785 Chr.1: 25863366-25863366 on Build GRCh38
rs1020868374 Chr.1: 25863621-25863621 on Build GRCh38
rs749811681 Chr.1: 25863527-25863527 on Build GRCh38
rs146982273 Chr.1: 25863354-25863354 on Build GRCh38
rs569125464 Chr.1: 25863436-25863436 on Build GRCh38
rs369406955 Chr.1: 25863486-25863486 on Build GRCh38
rs913685860 Chr.1: 25863263-25863263 on Build GRCh38
rs768357485 Chr.1: 25863348-25863348 on Build GRCh38
rs374295612 Chr.1: 25863648-25863648 on Build GRCh38
rs182757920 Chr.1: 25863232-25863232 on Build GRCh38
rs763325248 Chr.1: 25863382-25863382 on Build GRCh38
rs757947446 Chr.1: 25863339-25863339 on Build GRCh38
rs934047946 Chr.1: 25863254-25863254 on Build GRCh38
rs760768524 Chr.1: 25863557-25863557 on Build GRCh38
rs909491269 Chr.1: 25863494-25863494 on Build GRCh38
rs529741953 Chr.1: 25863296-25863296 on Build GRCh38
rs751479758 Chr.1: 25863595-25863595 on Build GRCh38
rs762787343 Chr.1: 25863653-25863653 on Build GRCh38
rs765095615 Chr.1: 25863563-25863563 on Build GRCh38
rs955459504 Chr.1: 25863553-25863553 on Build GRCh38
rs778951565 Chr.1: 25863515-25863516 on Build GRCh38
rs761716951 Chr.1: 25863427-25863427 on Build GRCh38
rs766292150 Chr.1: 25863469-25863469 on Build GRCh38
rs748588917 Chr.1: 25863363-25863363 on Build GRCh38
rs149765684 Chr.1: 25863629-25863629 on Build GRCh38
rs763145536 Chr.1: 25863264-25863264 on Build GRCh38
rs148531111 Chr.1: 25863643-25863643 on Build GRCh38
rs756537005 Chr.1: 25863518-25863518 on Build GRCh38
rs142174295 Chr.1: 25863420-25863420 on Build GRCh38
rs1030256980 Chr.1: 25863210-25863210 on Build GRCh38
rs751857522 Chr.1: 25863272-25863272 on Build GRCh38
rs376335279 Chr.1: 25863444-25863444 on Build GRCh38
rs142505796 Chr.1: 25863530-25863530 on Build GRCh38
rs773795405 Chr.1: 25863373-25863373 on Build GRCh38
rs1054767814 Chr.1: 25863344-25863344 on Build GRCh38
rs779730331 Chr.1: 25863216-25863216 on Build GRCh38
rs780873688 Chr.1: 25863492-25863492 on Build GRCh38
rs376473466 Chr.1: 25863624-25863624 on Build GRCh38
rs750304661 Chr.1: 25863332-25863332 on Build GRCh38
rs770401597 Chr.1: 25863233-25863233 on Build GRCh38
rs751138332 Chr.1: 25863474-25863474 on Build GRCh38
rs1047960753 Chr.1: 25863485-25863485 on Build GRCh38
rs1044023486 Chr.1: 25863277-25863277 on Build GRCh38
rs373339928 Chr.1: 25863531-25863531 on Build GRCh38
rs774512066 Chr.1: 25863398-25863398 on Build GRCh38
rs201392809 Chr.1: 25863304-25863304 on Build GRCh38
rs1054801245 Chr.1: 25863636-25863636 on Build GRCh38
rs777929299 Chr.1: 25863525-25863525 on Build GRCh38
rs749258548 Chr.1: 25863420-25863422 on Build GRCh38
rs369827250 Chr.1: 25863390-25863390 on Build GRCh38
rs760375332 Chr.1: 25863455-25863455 on Build GRCh38
rs752395755 Chr.1: 25863598-25863598 on Build GRCh38
rs147694393 Chr.1: 25863649-25863649 on Build GRCh38
rs747099473 Chr.1: 25863645-25863645 on Build GRCh38
rs746327855 Chr.1: 25863637-25863637 on Build GRCh38
rs200277795 Chr.1: 25863640-25863640 on Build GRCh38
rs1021314393 Chr.1: 25863356-25863356 on Build GRCh38
rs375757917 Chr.1: 25863554-25863554 on Build GRCh38
rs201686343 Chr.1: 25863652-25863652 on Build GRCh38
rs766323452 Chr.1: 25863271-25863271 on Build GRCh38
rs772459555 Chr.1: 25863571-25863572 on Build GRCh38
rs143463212 Chr.1: 25863528-25863528 on Build GRCh38
rs77010008 Chr.1: 25863297-25863297 on Build GRCh38

View Full Product Details