Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154449951-154450439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM50A
Entrez Gene ID
9130
Gene Name
family with sequence similarity 50 member A
Gene Aliases
9F, DXS9928E, HXC-26, HXC26, XAP5
UniGene
Hs.54277
Gene Chromosome Location
Chr.X: 154444126-154450654 on Build GRCh38
Transcripts
NM_004699.3 NR_106917.1

Gene Symbol
MIR6858
Entrez Gene ID
102465517
Gene Name
microRNA 6858
Gene Aliases
hsa-mir-6858
UniGene
Gene Chromosome Location
Chr.X: 154450320-154450386 on Build GRCh38
Transcripts
NM_004699.3 NR_106917.1

SNP Detail

SNP ID Chromosome Location
rs782149905 Chr.X: 154450121-154450121 on Build GRCh38
rs955555611 Chr.X: 154450369-154450369 on Build GRCh38
rs782284284 Chr.X: 154450244-154450244 on Build GRCh38
rs782207326 Chr.X: 154450169-154450169 on Build GRCh38
rs373505345 Chr.X: 154450136-154450136 on Build GRCh38
rs782819337 Chr.X: 154450333-154450333 on Build GRCh38
rs782022917 Chr.X: 154450119-154450119 on Build GRCh38
rs782537729 Chr.X: 154450365-154450365 on Build GRCh38
rs202185693 Chr.X: 154449971-154449971 on Build GRCh38
rs2070645 Chr.X: 154450384-154450384 on Build GRCh38
rs782716619 Chr.X: 154449972-154449972 on Build GRCh38
rs782628005 Chr.X: 154450057-154450057 on Build GRCh38
rs200500112 Chr.X: 154450013-154450013 on Build GRCh38
rs781946811 Chr.X: 154450295-154450295 on Build GRCh38
rs782651637 Chr.X: 154450162-154450162 on Build GRCh38
rs782769485 Chr.X: 154450117-154450117 on Build GRCh38
rs146149675 Chr.X: 154450078-154450078 on Build GRCh38
rs782497760 Chr.X: 154449975-154449975 on Build GRCh38
rs781950938 Chr.X: 154450109-154450109 on Build GRCh38
rs782483106 Chr.X: 154450132-154450132 on Build GRCh38
rs782477796 Chr.X: 154450355-154450355 on Build GRCh38
rs781944850 Chr.X: 154450084-154450084 on Build GRCh38
rs781890876 Chr.X: 154449997-154449997 on Build GRCh38
rs781954626 Chr.X: 154450186-154450186 on Build GRCh38
rs781843516 Chr.X: 154450354-154450354 on Build GRCh38
rs782674967 Chr.X: 154450163-154450163 on Build GRCh38
rs781940414 Chr.X: 154450256-154450256 on Build GRCh38
rs782281800 Chr.X: 154449974-154449974 on Build GRCh38
rs893726478 Chr.X: 154450020-154450020 on Build GRCh38
rs2070644 Chr.X: 154450358-154450358 on Build GRCh38
rs201052035 Chr.X: 154450268-154450268 on Build GRCh38
rs372392901 Chr.X: 154450405-154450405 on Build GRCh38
rs781904364 Chr.X: 154450010-154450011 on Build GRCh38
rs375504099 Chr.X: 154450130-154450130 on Build GRCh38
rs782546042 Chr.X: 154450158-154450158 on Build GRCh38
rs201845443 Chr.X: 154450195-154450195 on Build GRCh38
rs183827901 Chr.X: 154450183-154450183 on Build GRCh38
rs372569088 Chr.X: 154450116-154450116 on Build GRCh38
rs782324011 Chr.X: 154450418-154450418 on Build GRCh38
rs781979644 Chr.X: 154450182-154450182 on Build GRCh38
rs782471580 Chr.X: 154450346-154450346 on Build GRCh38
rs782774175 Chr.X: 154450138-154450138 on Build GRCh38
rs782547697 Chr.X: 154450289-154450289 on Build GRCh38
rs782394821 Chr.X: 154450250-154450250 on Build GRCh38
rs782485233 Chr.X: 154450397-154450397 on Build GRCh38
rs782395886 Chr.X: 154450036-154450036 on Build GRCh38
rs781858537 Chr.X: 154450151-154450151 on Build GRCh38
rs368796448 Chr.X: 154450131-154450131 on Build GRCh38
rs782190969 Chr.X: 154450383-154450383 on Build GRCh38
rs782121006 Chr.X: 154450318-154450318 on Build GRCh38
rs782365496 Chr.X: 154450107-154450107 on Build GRCh38
rs782242119 Chr.X: 154450359-154450359 on Build GRCh38
rs781885732 Chr.X: 154450339-154450339 on Build GRCh38
rs782696334 Chr.X: 154450350-154450350 on Build GRCh38
rs782387130 Chr.X: 154450191-154450191 on Build GRCh38
rs782222242 Chr.X: 154450034-154450034 on Build GRCh38
rs782667134 Chr.X: 154450377-154450377 on Build GRCh38
rs1051536455 Chr.X: 154449970-154449970 on Build GRCh38
rs782189294 Chr.X: 154450106-154450106 on Build GRCh38
rs782290010 Chr.X: 154450075-154450075 on Build GRCh38
rs201070149 Chr.X: 154449983-154449983 on Build GRCh38

View Full Product Details