Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 19076940-19077444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTTY14
Entrez Gene ID
83869
Gene Name
testis-specific transcript, Y-linked 14 (non-protein coding)
Gene Aliases
CYorf14, NCRNA00137, NCRNA00185, PRO2834, TTY14
UniGene
Hs.138453
Gene Chromosome Location
Chr.Y: 18872501-19077547 on Build GRCh38
Transcripts
NR_001543.3 NR_125733.1 NR_125734.1 NR_125735.1

SNP Detail

SNP ID Chromosome Location
rs770517809 Chr.Y: 19077230-19077230 on Build GRCh38
rs887054835 Chr.Y: 19077092-19077092 on Build GRCh38
rs1038670687 Chr.Y: 19077112-19077112 on Build GRCh38
rs939375648 Chr.Y: 19076999-19076999 on Build GRCh38
rs955122623 Chr.Y: 19077375-19077375 on Build GRCh38
rs778266990 Chr.Y: 19077242-19077242 on Build GRCh38
rs757944975 Chr.Y: 19077224-19077224 on Build GRCh38
rs1056473196 Chr.Y: 19077066-19077066 on Build GRCh38
rs767438348 Chr.Y: 19077374-19077374 on Build GRCh38
rs1028032081 Chr.Y: 19077252-19077252 on Build GRCh38
rs755004542 Chr.Y: 19077284-19077284 on Build GRCh38
rs777588103 Chr.Y: 19076987-19076987 on Build GRCh38
rs898341865 Chr.Y: 19077133-19077133 on Build GRCh38
rs1004115264 Chr.Y: 19077097-19077097 on Build GRCh38

View Full Product Details