Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71073193-71073691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX12
Entrez Gene ID
29934
Gene Name
sorting nexin 12
Gene Aliases
-
UniGene
Hs.260750
Gene Chromosome Location
Chr.X: 71059247-71073426 on Build GRCh38
Transcripts
NM_001256185.1

SNP Detail

SNP ID Chromosome Location
rs375130664 Chr.X: 71073546-71073546 on Build GRCh38
rs958335429 Chr.X: 71073266-71073266 on Build GRCh38
rs184007544 Chr.X: 71073386-71073386 on Build GRCh38
rs772913215 Chr.X: 71073274-71073274 on Build GRCh38
rs145321617 Chr.X: 71073273-71073273 on Build GRCh38
rs936455939 Chr.X: 71073311-71073311 on Build GRCh38
rs754197885 Chr.X: 71073663-71073663 on Build GRCh38
rs766134908 Chr.X: 71073367-71073367 on Build GRCh38
rs1046649065 Chr.X: 71073377-71073377 on Build GRCh38
rs760401365 Chr.X: 71073345-71073345 on Build GRCh38
rs144996267 Chr.X: 71073330-71073330 on Build GRCh38
rs908676961 Chr.X: 71073632-71073632 on Build GRCh38
rs983347479 Chr.X: 71073574-71073574 on Build GRCh38
rs906767803 Chr.X: 71073411-71073411 on Build GRCh38
rs1034058920 Chr.X: 71073477-71073477 on Build GRCh38
rs1024957922 Chr.X: 71073346-71073346 on Build GRCh38
rs776492274 Chr.X: 71073303-71073303 on Build GRCh38
rs1009675143 Chr.X: 71073349-71073349 on Build GRCh38
rs772991160 Chr.X: 71073647-71073647 on Build GRCh38
rs1002219814 Chr.X: 71073417-71073417 on Build GRCh38
rs1006076414 Chr.X: 71073598-71073598 on Build GRCh38

View Full Product Details