Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28505967-28506536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCC1
Entrez Gene ID
1104
Gene Name
regulator of chromosome condensation 1
Gene Aliases
CHC1, RCC1-I, SNHG3-RCC1
UniGene
Hs.469723
Gene Chromosome Location
Chr.1: 28505943-28539196 on Build GRCh38
Transcripts
NM_001048199.2 NR_030725.1 NR_030726.1 NR_002909.2 NR_036473.1

Gene Symbol
SNHG3
Entrez Gene ID
8420
Gene Name
small nucleolar RNA host gene 3
Gene Aliases
NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
UniGene
Hs.469723
Gene Chromosome Location
Chr.1: 28505943-28510892 on Build GRCh38
Transcripts
NM_001048199.2 NR_030725.1 NR_030726.1 NR_002909.2 NR_036473.1

SNP Detail

SNP ID Chromosome Location
rs946041557 Chr.1: 28506076-28506076 on Build GRCh38
rs892296880 Chr.1: 28506036-28506037 on Build GRCh38
rs142712867 Chr.1: 28506294-28506294 on Build GRCh38
rs546184610 Chr.1: 28506487-28506487 on Build GRCh38
rs961578387 Chr.1: 28506153-28506153 on Build GRCh38
rs987516413 Chr.1: 28506072-28506072 on Build GRCh38
rs1046839957 Chr.1: 28506145-28506145 on Build GRCh38
rs945180421 Chr.1: 28506096-28506096 on Build GRCh38
rs774203093 Chr.1: 28506202-28506203 on Build GRCh38
rs997302955 Chr.1: 28506125-28506125 on Build GRCh38
rs970630114 Chr.1: 28506498-28506498 on Build GRCh38
rs751024327 Chr.1: 28506217-28506217 on Build GRCh38
rs765371028 Chr.1: 28506509-28506509 on Build GRCh38
rs895543800 Chr.1: 28506023-28506023 on Build GRCh38
rs373823348 Chr.1: 28506278-28506278 on Build GRCh38
rs565714290 Chr.1: 28506169-28506169 on Build GRCh38
rs774289710 Chr.1: 28506089-28506089 on Build GRCh38
rs997177221 Chr.1: 28506179-28506179 on Build GRCh38
rs762410440 Chr.1: 28506200-28506201 on Build GRCh38
rs557662811 Chr.1: 28506316-28506316 on Build GRCh38
rs1001696462 Chr.1: 28506016-28506016 on Build GRCh38
rs761656967 Chr.1: 28506173-28506173 on Build GRCh38
rs566941831 Chr.1: 28506041-28506041 on Build GRCh38
rs397863560 Chr.1: 28506216-28506217 on Build GRCh38
rs770683956 Chr.1: 28506202-28506202 on Build GRCh38
rs996563589 Chr.1: 28506398-28506398 on Build GRCh38
rs992973993 Chr.1: 28506160-28506160 on Build GRCh38
rs1032973821 Chr.1: 28506475-28506475 on Build GRCh38
rs565751838 Chr.1: 28506060-28506060 on Build GRCh38
rs1017031446 Chr.1: 28506167-28506167 on Build GRCh38
rs777672645 Chr.1: 28506101-28506101 on Build GRCh38
rs748493338 Chr.1: 28506070-28506070 on Build GRCh38
rs1029456986 Chr.1: 28506182-28506182 on Build GRCh38
rs1014424811 Chr.1: 28506473-28506473 on Build GRCh38
rs564932856 Chr.1: 28506363-28506363 on Build GRCh38
rs34988350 Chr.1: 28506084-28506084 on Build GRCh38
rs926350144 Chr.1: 28506202-28506202 on Build GRCh38
rs6696931 Chr.1: 28506175-28506175 on Build GRCh38
rs941122126 Chr.1: 28506351-28506351 on Build GRCh38
rs879342640 Chr.1: 28506189-28506192 on Build GRCh38
rs899502184 Chr.1: 28506391-28506391 on Build GRCh38
rs536754448 Chr.1: 28506038-28506038 on Build GRCh38
rs182141011 Chr.1: 28506015-28506015 on Build GRCh38
rs187220836 Chr.1: 28506044-28506044 on Build GRCh38
rs975251547 Chr.1: 28506057-28506057 on Build GRCh38
rs41270833 Chr.1: 28506043-28506043 on Build GRCh38
rs953050075 Chr.1: 28505991-28505991 on Build GRCh38
rs574922680 Chr.1: 28506221-28506221 on Build GRCh38
rs957784065 Chr.1: 28506514-28506514 on Build GRCh38
rs1047285147 Chr.1: 28506320-28506320 on Build GRCh38
rs891070216 Chr.1: 28506410-28506410 on Build GRCh38
rs917679113 Chr.1: 28506058-28506058 on Build GRCh38
rs886887414 Chr.1: 28506148-28506148 on Build GRCh38
rs1020311040 Chr.1: 28506197-28506197 on Build GRCh38
rs776606982 Chr.1: 28506124-28506124 on Build GRCh38
rs946261132 Chr.1: 28506003-28506003 on Build GRCh38
rs1014470331 Chr.1: 28506130-28506130 on Build GRCh38
rs1050252299 Chr.1: 28506342-28506342 on Build GRCh38
rs766794789 Chr.1: 28505987-28505987 on Build GRCh38
rs759132782 Chr.1: 28506119-28506119 on Build GRCh38
rs1043310713 Chr.1: 28506014-28506014 on Build GRCh38
rs895994631 Chr.1: 28506123-28506123 on Build GRCh38
rs542995461 Chr.1: 28506434-28506434 on Build GRCh38
rs991534709 Chr.1: 28506489-28506489 on Build GRCh38
rs559647383 Chr.1: 28506313-28506313 on Build GRCh38
rs559517521 Chr.1: 28506471-28506471 on Build GRCh38
rs762371430 Chr.1: 28506215-28506215 on Build GRCh38
rs796356954 Chr.1: 28506080-28506080 on Build GRCh38
rs11810945 Chr.1: 28506074-28506074 on Build GRCh38
rs1053746143 Chr.1: 28506192-28506192 on Build GRCh38
rs6656720 Chr.1: 28506185-28506185 on Build GRCh38
rs759838536 Chr.1: 28506494-28506494 on Build GRCh38
rs967406366 Chr.1: 28506464-28506464 on Build GRCh38
rs756421744 Chr.1: 28506297-28506297 on Build GRCh38
rs905125832 Chr.1: 28506033-28506034 on Build GRCh38
rs749982349 Chr.1: 28506037-28506037 on Build GRCh38
rs537647807 Chr.1: 28506295-28506295 on Build GRCh38
rs188359143 Chr.1: 28506150-28506150 on Build GRCh38
rs781664890 Chr.1: 28506051-28506051 on Build GRCh38
rs780219314 Chr.1: 28506481-28506481 on Build GRCh38
rs535870908 Chr.1: 28506477-28506477 on Build GRCh38
rs765404213 Chr.1: 28506050-28506050 on Build GRCh38
rs567006023 Chr.1: 28506467-28506467 on Build GRCh38
rs980922991 Chr.1: 28506202-28506205 on Build GRCh38
rs781643299 Chr.1: 28506241-28506241 on Build GRCh38
rs759398942 Chr.1: 28506201-28506202 on Build GRCh38
rs989443353 Chr.1: 28506248-28506248 on Build GRCh38
rs1033836215 Chr.1: 28506508-28506508 on Build GRCh38
rs964330572 Chr.1: 28506048-28506048 on Build GRCh38
rs984305757 Chr.1: 28505994-28505994 on Build GRCh38
rs1052400752 Chr.1: 28506236-28506236 on Build GRCh38
rs56695711 Chr.1: 28506216-28506216 on Build GRCh38
rs954760777 Chr.1: 28506071-28506071 on Build GRCh38
rs566424430 Chr.1: 28506505-28506505 on Build GRCh38
rs996872894 Chr.1: 28506042-28506042 on Build GRCh38
rs764949165 Chr.1: 28506032-28506032 on Build GRCh38
rs765984083 Chr.1: 28506216-28506216 on Build GRCh38
rs922420060 Chr.1: 28506301-28506301 on Build GRCh38
rs180771311 Chr.1: 28506331-28506331 on Build GRCh38
rs745904461 Chr.1: 28506362-28506362 on Build GRCh38
rs35746493 Chr.1: 28506286-28506287 on Build GRCh38
rs2840757 Chr.1: 28506411-28506411 on Build GRCh38
rs755103623 Chr.1: 28506040-28506040 on Build GRCh38
rs539266427 Chr.1: 28506075-28506075 on Build GRCh38
rs779097106 Chr.1: 28506356-28506356 on Build GRCh38
rs148971157 Chr.1: 28506044-28506045 on Build GRCh38
rs761201143 Chr.1: 28506443-28506443 on Build GRCh38
rs1028356703 Chr.1: 28506135-28506135 on Build GRCh38
rs967458783 Chr.1: 28506273-28506273 on Build GRCh38
rs556589436 Chr.1: 28506039-28506039 on Build GRCh38
rs925960015 Chr.1: 28506289-28506289 on Build GRCh38
rs1005978149 Chr.1: 28506385-28506385 on Build GRCh38
rs987967324 Chr.1: 28506186-28506186 on Build GRCh38
rs1038008556 Chr.1: 28506372-28506372 on Build GRCh38
rs926206335 Chr.1: 28506088-28506088 on Build GRCh38
rs1022247343 Chr.1: 28506079-28506094 on Build GRCh38
rs760722803 Chr.1: 28506117-28506117 on Build GRCh38
rs763982811 Chr.1: 28506220-28506224 on Build GRCh38
rs950299586 Chr.1: 28506507-28506507 on Build GRCh38
rs373988067 Chr.1: 28506081-28506081 on Build GRCh38
rs892717216 Chr.1: 28506200-28506200 on Build GRCh38
rs558895216 Chr.1: 28506118-28506118 on Build GRCh38
rs548717763 Chr.1: 28506021-28506021 on Build GRCh38
rs771044997 Chr.1: 28506338-28506338 on Build GRCh38
rs769309515 Chr.1: 28506303-28506303 on Build GRCh38
rs1054109769 Chr.1: 28506083-28506083 on Build GRCh38
rs978761121 Chr.1: 28506283-28506283 on Build GRCh38
rs1053636612 Chr.1: 28506035-28506036 on Build GRCh38
rs772696597 Chr.1: 28506374-28506374 on Build GRCh38
rs936547317 Chr.1: 28506034-28506034 on Build GRCh38
rs752876439 Chr.1: 28506511-28506511 on Build GRCh38
rs775229064 Chr.1: 28506202-28506204 on Build GRCh38
rs917488230 Chr.1: 28506493-28506493 on Build GRCh38
rs945639023 Chr.1: 28506233-28506233 on Build GRCh38
rs1029524876 Chr.1: 28506401-28506401 on Build GRCh38
rs936316857 Chr.1: 28506220-28506220 on Build GRCh38

View Full Product Details