Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 63370644-63371122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG6
Entrez Gene ID
29929
Gene Name
ALG6, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1C
UniGene
Hs.258501
Gene Chromosome Location
Chr.1: 63367590-63438562 on Build GRCh38
Transcripts
NM_013339.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2238633 c.67C>G p.L23V Chr.1: 63371044-63371044 on Build GRCh38
7355204 c.67C>G p.L23V Chr.1: 63371044-63371044 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752858908 Chr.1: 63370989-63370989 on Build GRCh38
rs188685870 Chr.1: 63370850-63370850 on Build GRCh38
rs1009518114 Chr.1: 63370794-63370794 on Build GRCh38
rs370087611 Chr.1: 63370955-63370955 on Build GRCh38
rs771741581 Chr.1: 63370935-63370935 on Build GRCh38
rs961669633 Chr.1: 63370672-63370672 on Build GRCh38
rs751787678 Chr.1: 63371018-63371018 on Build GRCh38
rs752384068 Chr.1: 63370692-63370692 on Build GRCh38
rs893072117 Chr.1: 63370840-63370840 on Build GRCh38
rs761075072 Chr.1: 63370943-63370943 on Build GRCh38
rs776645198 Chr.1: 63371080-63371080 on Build GRCh38
rs762853715 Chr.1: 63370797-63370797 on Build GRCh38
rs758595644 Chr.1: 63370999-63370999 on Build GRCh38
rs941050814 Chr.1: 63370699-63370699 on Build GRCh38
rs933631478 Chr.1: 63370677-63370677 on Build GRCh38
rs777768835 Chr.1: 63370902-63370902 on Build GRCh38
rs750579230 Chr.1: 63371080-63371084 on Build GRCh38
rs747748799 Chr.1: 63370934-63370934 on Build GRCh38
rs573955265 Chr.1: 63370693-63370693 on Build GRCh38
rs990992107 Chr.1: 63371084-63371084 on Build GRCh38
rs904999510 Chr.1: 63370749-63370749 on Build GRCh38
rs1038621577 Chr.1: 63370744-63370744 on Build GRCh38
rs562934427 Chr.1: 63370978-63370978 on Build GRCh38
rs990960812 Chr.1: 63370750-63370750 on Build GRCh38
rs766108695 Chr.1: 63370805-63370805 on Build GRCh38
rs1018206458 Chr.1: 63370827-63370827 on Build GRCh38
rs11546562 Chr.1: 63370988-63370988 on Build GRCh38
rs375824664 Chr.1: 63370980-63370980 on Build GRCh38
rs559986470 Chr.1: 63370764-63370764 on Build GRCh38
rs373857344 Chr.1: 63371089-63371089 on Build GRCh38
rs778354011 Chr.1: 63371051-63371051 on Build GRCh38
rs924330655 Chr.1: 63370782-63370782 on Build GRCh38
rs376116153 Chr.1: 63371078-63371078 on Build GRCh38
rs772902612 Chr.1: 63371067-63371067 on Build GRCh38
rs1016079273 Chr.1: 63370858-63370858 on Build GRCh38
rs373800951 Chr.1: 63371031-63371031 on Build GRCh38
rs114299980 Chr.1: 63370674-63370674 on Build GRCh38
rs183861757 Chr.1: 63370759-63370759 on Build GRCh38
rs376601985 Chr.1: 63370942-63370942 on Build GRCh38
rs776870408 Chr.1: 63371083-63371083 on Build GRCh38
rs995929209 Chr.1: 63370871-63370871 on Build GRCh38
rs760082542 Chr.1: 63371088-63371088 on Build GRCh38
rs752876663 Chr.1: 63370836-63370836 on Build GRCh38
rs370891576 Chr.1: 63371093-63371093 on Build GRCh38
rs559834759 Chr.1: 63371079-63371080 on Build GRCh38
rs1054104046 Chr.1: 63370837-63370837 on Build GRCh38
rs908175212 Chr.1: 63370859-63370859 on Build GRCh38
rs116034504 Chr.1: 63370857-63370857 on Build GRCh38
rs754929467 Chr.1: 63371042-63371042 on Build GRCh38
rs749194292 Chr.1: 63371032-63371032 on Build GRCh38
rs368534328 Chr.1: 63371090-63371090 on Build GRCh38
rs765350120 Chr.1: 63370851-63370851 on Build GRCh38
rs746703376 Chr.1: 63371068-63371068 on Build GRCh38
rs941122209 Chr.1: 63370916-63370916 on Build GRCh38
rs759962410 Chr.1: 63370962-63370962 on Build GRCh38
rs547966478 Chr.1: 63370729-63370730 on Build GRCh38
rs775890435 Chr.1: 63370721-63370729 on Build GRCh38
rs747624965 Chr.1: 63371054-63371054 on Build GRCh38
rs968110356 Chr.1: 63370813-63370813 on Build GRCh38
rs542800575 Chr.1: 63370818-63370818 on Build GRCh38
rs755933716 Chr.1: 63371029-63371029 on Build GRCh38
rs897176330 Chr.1: 63370860-63370860 on Build GRCh38
rs34542411 Chr.1: 63370842-63370842 on Build GRCh38
rs151277583 Chr.1: 63371013-63371013 on Build GRCh38
rs764336209 Chr.1: 63371091-63371091 on Build GRCh38
rs572242645 Chr.1: 63370775-63370775 on Build GRCh38
rs773863311 Chr.1: 63370932-63370932 on Build GRCh38
rs537454260 Chr.1: 63370698-63370698 on Build GRCh38
rs974377636 Chr.1: 63370673-63370673 on Build GRCh38
rs771618945 Chr.1: 63371060-63371060 on Build GRCh38
rs545125464 Chr.1: 63370762-63370762 on Build GRCh38
rs140944848 Chr.1: 63370981-63370981 on Build GRCh38
rs555715368 Chr.1: 63370752-63370752 on Build GRCh38
rs777188080 Chr.1: 63370956-63370956 on Build GRCh38
rs113457394 Chr.1: 63370917-63370917 on Build GRCh38

View Full Product Details