Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15644929-15645325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM27
Entrez Gene ID
57393
Gene Name
transmembrane protein 27
Gene Aliases
NX-17, NX17
UniGene
Hs.129614
Gene Chromosome Location
Chr.X: 15627316-15675624 on Build GRCh38
Transcripts
NM_020665.5 XM_017029680.1 XM_017029681.1

SNP Detail

SNP ID Chromosome Location
rs3840973 Chr.X: 15645110-15645111 on Build GRCh38
rs138307428 Chr.X: 15645020-15645020 on Build GRCh38
rs768054047 Chr.X: 15645024-15645024 on Build GRCh38
rs958627244 Chr.X: 15645177-15645177 on Build GRCh38
rs780021656 Chr.X: 15645064-15645064 on Build GRCh38
rs764396185 Chr.X: 15645037-15645037 on Build GRCh38
rs917148112 Chr.X: 15645168-15645168 on Build GRCh38
rs749685946 Chr.X: 15645046-15645047 on Build GRCh38
rs1002540109 Chr.X: 15645126-15645126 on Build GRCh38
rs753192684 Chr.X: 15645029-15645029 on Build GRCh38
rs752101672 Chr.X: 15645087-15645087 on Build GRCh38
rs764565981 Chr.X: 15645084-15645084 on Build GRCh38
rs757634131 Chr.X: 15645090-15645090 on Build GRCh38
rs977568234 Chr.X: 15645269-15645269 on Build GRCh38
rs3810721 Chr.X: 15645250-15645250 on Build GRCh38
rs757908162 Chr.X: 15645056-15645056 on Build GRCh38
rs369828956 Chr.X: 15645052-15645052 on Build GRCh38
rs774427382 Chr.X: 15645006-15645006 on Build GRCh38
rs773456053 Chr.X: 15644966-15644966 on Build GRCh38
rs187219255 Chr.X: 15645054-15645054 on Build GRCh38
rs961755611 Chr.X: 15645112-15645112 on Build GRCh38
rs978504531 Chr.X: 15645080-15645080 on Build GRCh38
rs756980026 Chr.X: 15645042-15645042 on Build GRCh38
rs761067230 Chr.X: 15645034-15645034 on Build GRCh38
rs1015078734 Chr.X: 15645093-15645093 on Build GRCh38
rs35510091 Chr.X: 15645251-15645252 on Build GRCh38
rs867931283 Chr.X: 15644988-15644988 on Build GRCh38
rs936987792 Chr.X: 15645103-15645105 on Build GRCh38
rs3810722 Chr.X: 15645247-15645247 on Build GRCh38
rs991803869 Chr.X: 15645220-15645220 on Build GRCh38
rs756753318 Chr.X: 15645127-15645127 on Build GRCh38
rs770950539 Chr.X: 15644991-15644991 on Build GRCh38
rs778413092 Chr.X: 15645139-15645139 on Build GRCh38
rs753150976 Chr.X: 15644984-15644984 on Build GRCh38
rs759970785 Chr.X: 15645014-15645014 on Build GRCh38
rs781342367 Chr.X: 15645136-15645136 on Build GRCh38

View Full Product Details