Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99459940-99460495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5J2
Entrez Gene ID
9551
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2
Gene Aliases
ATP5JL
UniGene
Hs.521056
Gene Chromosome Location
Chr.7: 99458161-99466201 on Build GRCh38
Transcripts
NM_001003713.2 NM_001003714.2 NM_001039178.2 NM_004889.3 NM_001198879.1

Gene Symbol
ATP5J2-PTCD1
Entrez Gene ID
100526740
Gene Name
ATP5J2-PTCD1 readthrough
Gene Aliases
-
UniGene
Hs.632313
Gene Chromosome Location
Chr.7: 99416739-99466201 on Build GRCh38
Transcripts
NM_001003713.2 NM_001003714.2 NM_001039178.2 NM_004889.3 NM_001198879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6178836 c.137G>C p.R46T Chr.7: 99460088-99460088 on Build GRCh38
5734344 c.124G>A p.G42R Chr.7: 99460101-99460101 on Build GRCh38
5408374 c.73G>A p.E25K Chr.7: 99460152-99460152 on Build GRCh38
5031466 c.139+4A>G p.? Chr.7: 99460082-99460082 on Build GRCh38
6758067 c.129G>A p.A43A Chr.7: 99460096-99460096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11545970 Chr.7: 99460097-99460097 on Build GRCh38
rs895854524 Chr.7: 99460395-99460395 on Build GRCh38
rs764118027 Chr.7: 99460058-99460058 on Build GRCh38
rs745686013 Chr.7: 99460270-99460270 on Build GRCh38
rs766790399 Chr.7: 99460218-99460218 on Build GRCh38
rs746062789 Chr.7: 99460112-99460112 on Build GRCh38
rs1052573930 Chr.7: 99460026-99460026 on Build GRCh38
rs1014888592 Chr.7: 99460425-99460425 on Build GRCh38
rs768219447 Chr.7: 99460091-99460091 on Build GRCh38
rs183974756 Chr.7: 99460289-99460289 on Build GRCh38
rs776656692 Chr.7: 99460200-99460200 on Build GRCh38
rs931037840 Chr.7: 99460019-99460019 on Build GRCh38
rs770580648 Chr.7: 99460254-99460254 on Build GRCh38
rs750776284 Chr.7: 99460132-99460132 on Build GRCh38
rs112721137 Chr.7: 99459998-99459998 on Build GRCh38
rs765677994 Chr.7: 99460202-99460202 on Build GRCh38
rs143569736 Chr.7: 99460128-99460128 on Build GRCh38
rs947557667 Chr.7: 99459991-99459991 on Build GRCh38
rs776394137 Chr.7: 99460094-99460094 on Build GRCh38
rs760970218 Chr.7: 99460195-99460195 on Build GRCh38
rs762907766 Chr.7: 99460101-99460101 on Build GRCh38
rs374144496 Chr.7: 99460165-99460165 on Build GRCh38
rs546920519 Chr.7: 99460471-99460471 on Build GRCh38
rs370605940 Chr.7: 99460137-99460137 on Build GRCh38
rs962287483 Chr.7: 99460238-99460238 on Build GRCh38
rs973980977 Chr.7: 99460240-99460241 on Build GRCh38
rs574504620 Chr.7: 99460133-99460133 on Build GRCh38
rs746275800 Chr.7: 99460042-99460042 on Build GRCh38
rs564655443 Chr.7: 99460322-99460322 on Build GRCh38
rs769849318 Chr.7: 99460160-99460160 on Build GRCh38
rs74592598 Chr.7: 99460217-99460217 on Build GRCh38
rs767618873 Chr.7: 99460102-99460102 on Build GRCh38
rs757474349 Chr.7: 99460180-99460180 on Build GRCh38
rs943730630 Chr.7: 99460342-99460342 on Build GRCh38
rs761943310 Chr.7: 99460201-99460201 on Build GRCh38
rs993120190 Chr.7: 99460181-99460181 on Build GRCh38
rs886979382 Chr.7: 99460424-99460424 on Build GRCh38
rs191453506 Chr.7: 99460279-99460279 on Build GRCh38
rs749276404 Chr.7: 99460253-99460253 on Build GRCh38
rs750936486 Chr.7: 99460207-99460207 on Build GRCh38
rs765804602 Chr.7: 99460130-99460130 on Build GRCh38
rs149159438 Chr.7: 99460096-99460096 on Build GRCh38
rs752241591 Chr.7: 99460249-99460249 on Build GRCh38
rs973651272 Chr.7: 99460446-99460446 on Build GRCh38
rs540823353 Chr.7: 99460308-99460308 on Build GRCh38
rs2293256 Chr.7: 99460078-99460078 on Build GRCh38
rs746449773 Chr.7: 99460083-99460083 on Build GRCh38
rs559522608 Chr.7: 99460398-99460398 on Build GRCh38
rs1001645276 Chr.7: 99460373-99460373 on Build GRCh38
rs775793034 Chr.7: 99460107-99460107 on Build GRCh38
rs533020669 Chr.7: 99460423-99460423 on Build GRCh38
rs764262832 Chr.7: 99460127-99460127 on Build GRCh38
rs760602316 Chr.7: 99460126-99460126 on Build GRCh38
rs374308413 Chr.7: 99460053-99460053 on Build GRCh38
rs768806509 Chr.7: 99460196-99460196 on Build GRCh38
rs1023249264 Chr.7: 99460275-99460275 on Build GRCh38
rs546204621 Chr.7: 99460291-99460291 on Build GRCh38
rs1048751375 Chr.7: 99460406-99460406 on Build GRCh38
rs1004450303 Chr.7: 99460456-99460456 on Build GRCh38
rs960066117 Chr.7: 99460210-99460210 on Build GRCh38
rs772583509 Chr.7: 99459960-99459960 on Build GRCh38
rs891713955 Chr.7: 99460290-99460290 on Build GRCh38
rs753693595 Chr.7: 99460393-99460393 on Build GRCh38
rs935113422 Chr.7: 99460021-99460021 on Build GRCh38
rs780731999 Chr.7: 99460153-99460153 on Build GRCh38
rs904345249 Chr.7: 99460358-99460358 on Build GRCh38
rs754629343 Chr.7: 99460239-99460239 on Build GRCh38
rs751919246 Chr.7: 99460234-99460234 on Build GRCh38
rs943984583 Chr.7: 99460035-99460035 on Build GRCh38
rs773340655 Chr.7: 99460161-99460161 on Build GRCh38
rs1039334348 Chr.7: 99460378-99460378 on Build GRCh38
rs778743530 Chr.7: 99460259-99460259 on Build GRCh38
rs747701024 Chr.7: 99460156-99460156 on Build GRCh38
rs558445641 Chr.7: 99460002-99460002 on Build GRCh38
rs899482874 Chr.7: 99460391-99460391 on Build GRCh38
rs920749786 Chr.7: 99460268-99460268 on Build GRCh38
rs962626436 Chr.7: 99460431-99460431 on Build GRCh38
rs1049403266 Chr.7: 99460020-99460020 on Build GRCh38
rs528779374 Chr.7: 99460468-99460468 on Build GRCh38
rs781058090 Chr.7: 99460247-99460247 on Build GRCh38
rs754399390 Chr.7: 99460151-99460151 on Build GRCh38
rs968972496 Chr.7: 99460278-99460278 on Build GRCh38
rs945964092 Chr.7: 99460335-99460335 on Build GRCh38
rs577042985 Chr.7: 99460077-99460077 on Build GRCh38
rs749031991 Chr.7: 99460163-99460163 on Build GRCh38
rs777784470 Chr.7: 99460251-99460251 on Build GRCh38
rs763189831 Chr.7: 99460208-99460208 on Build GRCh38
rs777033268 Chr.7: 99460045-99460046 on Build GRCh38
rs916160845 Chr.7: 99459989-99459989 on Build GRCh38
rs746708460 Chr.7: 99460142-99460142 on Build GRCh38

View Full Product Details