Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 13012244-13012840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1456
Entrez Gene ID
57604
Gene Name
KIAA1456
Gene Aliases
C8orf79, TRM9L
UniGene
Hs.202521
Gene Chromosome Location
Chr.8: 12945674-13029777 on Build GRCh38
Transcripts
NM_001099677.1 NM_020844.2 XM_005273584.4 XM_005273585.4 XM_005273586.4 XM_005273587.4 XM_005273588.4 XM_005273590.4 XM_005273591.4 XM_011544596.2 XM_011544597.2 XM_011544598.2 XM_011544600.2 XM_011544601.2 XM_017013705.1 XM_017013706.1 XM_017013707.1 XM_017013708.1 XM_017013709.1 XM_017013710.1 XM_017013711.1 XM_017013712.1 XM_017013713.1 XM_017013714.1 XM_017013715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7263589 c.171A>T p.K57N Chr.8: 13012700-13012700 on Build GRCh38
4150521 c.303C>T p.G101G Chr.8: 13012832-13012832 on Build GRCh38
3316816 c.206G>T p.G69V Chr.8: 13012735-13012735 on Build GRCh38
3982392 c.155-7C>G p.? Chr.8: 13012677-13012677 on Build GRCh38
1454804 c.238G>A p.A80T Chr.8: 13012767-13012767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917095425 Chr.8: 13012782-13012782 on Build GRCh38
rs746616247 Chr.8: 13012658-13012658 on Build GRCh38
rs752365051 Chr.8: 13012749-13012749 on Build GRCh38
rs757868573 Chr.8: 13012683-13012683 on Build GRCh38
rs965994602 Chr.8: 13012457-13012457 on Build GRCh38
rs1014096713 Chr.8: 13012283-13012283 on Build GRCh38
rs1053694581 Chr.8: 13012707-13012707 on Build GRCh38
rs12156247 Chr.8: 13012630-13012630 on Build GRCh38
rs754614787 Chr.8: 13012687-13012687 on Build GRCh38
rs1027875074 Chr.8: 13012468-13012468 on Build GRCh38
rs578065264 Chr.8: 13012615-13012615 on Build GRCh38
rs536096840 Chr.8: 13012325-13012325 on Build GRCh38
rs752070881 Chr.8: 13012562-13012562 on Build GRCh38
rs368418221 Chr.8: 13012523-13012523 on Build GRCh38
rs770968807 Chr.8: 13012668-13012668 on Build GRCh38
rs190182385 Chr.8: 13012675-13012675 on Build GRCh38
rs1009759837 Chr.8: 13012559-13012559 on Build GRCh38
rs768448093 Chr.8: 13012661-13012661 on Build GRCh38
rs545539186 Chr.8: 13012731-13012731 on Build GRCh38
rs888037018 Chr.8: 13012557-13012557 on Build GRCh38
rs778716077 Chr.8: 13012815-13012815 on Build GRCh38
rs760510867 Chr.8: 13012721-13012721 on Build GRCh38
rs115574660 Chr.8: 13012448-13012448 on Build GRCh38
rs893681027 Chr.8: 13012317-13012317 on Build GRCh38
rs143563959 Chr.8: 13012494-13012494 on Build GRCh38
rs778807979 Chr.8: 13012761-13012761 on Build GRCh38
rs939725457 Chr.8: 13012425-13012425 on Build GRCh38
rs931037323 Chr.8: 13012375-13012375 on Build GRCh38
rs750335561 Chr.8: 13012505-13012505 on Build GRCh38
rs764475691 Chr.8: 13012730-13012730 on Build GRCh38
rs770344770 Chr.8: 13012780-13012780 on Build GRCh38
rs755917067 Chr.8: 13012708-13012708 on Build GRCh38
rs927652763 Chr.8: 13012430-13012430 on Build GRCh38
rs201998102 Chr.8: 13012681-13012681 on Build GRCh38
rs770866555 Chr.8: 13012715-13012715 on Build GRCh38
rs186930707 Chr.8: 13012770-13012770 on Build GRCh38
rs753545815 Chr.8: 13012805-13012805 on Build GRCh38
rs1015471443 Chr.8: 13012486-13012486 on Build GRCh38
rs1021069823 Chr.8: 13012337-13012337 on Build GRCh38
rs948633690 Chr.8: 13012529-13012529 on Build GRCh38
rs367913787 Chr.8: 13012712-13012712 on Build GRCh38
rs1040883290 Chr.8: 13012532-13012532 on Build GRCh38
rs746011537 Chr.8: 13012654-13012654 on Build GRCh38
rs939936514 Chr.8: 13012392-13012392 on Build GRCh38
rs763647389 Chr.8: 13012800-13012800 on Build GRCh38
rs185568826 Chr.8: 13012548-13012548 on Build GRCh38
rs374032501 Chr.8: 13012709-13012709 on Build GRCh38
rs117981597 Chr.8: 13012570-13012570 on Build GRCh38
rs944553689 Chr.8: 13012531-13012531 on Build GRCh38
rs988369688 Chr.8: 13012324-13012324 on Build GRCh38
rs757023992 Chr.8: 13012806-13012806 on Build GRCh38
rs1003027711 Chr.8: 13012625-13012625 on Build GRCh38
rs1019903424 Chr.8: 13012454-13012454 on Build GRCh38
rs965670281 Chr.8: 13012568-13012569 on Build GRCh38
rs757898545 Chr.8: 13012655-13012655 on Build GRCh38
rs200731092 Chr.8: 13012639-13012639 on Build GRCh38
rs931068601 Chr.8: 13012470-13012470 on Build GRCh38
rs575416849 Chr.8: 13012450-13012450 on Build GRCh38
rs2460338 Chr.8: 13012651-13012651 on Build GRCh38
rs374709869 Chr.8: 13012724-13012724 on Build GRCh38
rs368624446 Chr.8: 13012352-13012352 on Build GRCh38
rs925894438 Chr.8: 13012812-13012812 on Build GRCh38
rs1042592476 Chr.8: 13012452-13012452 on Build GRCh38
rs201071531 Chr.8: 13012794-13012794 on Build GRCh38
rs1055041357 Chr.8: 13012313-13012313 on Build GRCh38
rs1010724347 Chr.8: 13012329-13012329 on Build GRCh38
rs371196956 Chr.8: 13012401-13012401 on Build GRCh38
rs767819449 Chr.8: 13012649-13012649 on Build GRCh38
rs774447547 Chr.8: 13012672-13012672 on Build GRCh38
rs776989892 Chr.8: 13012772-13012772 on Build GRCh38
rs748312519 Chr.8: 13012277-13012277 on Build GRCh38
rs769547437 Chr.8: 13012665-13012665 on Build GRCh38
rs369005079 Chr.8: 13012774-13012774 on Build GRCh38
rs956860372 Chr.8: 13012447-13012447 on Build GRCh38
rs937878718 Chr.8: 13012312-13012312 on Build GRCh38
rs201747607 Chr.8: 13012766-13012766 on Build GRCh38
rs1023710893 Chr.8: 13012293-13012293 on Build GRCh38
rs902569000 Chr.8: 13012339-13012339 on Build GRCh38
rs760306719 Chr.8: 13012799-13012799 on Build GRCh38
rs957193403 Chr.8: 13012319-13012319 on Build GRCh38
rs997078114 Chr.8: 13012620-13012620 on Build GRCh38
rs1037048428 Chr.8: 13012752-13012752 on Build GRCh38
rs375223632 Chr.8: 13012736-13012736 on Build GRCh38
rs867648788 Chr.8: 13012526-13012526 on Build GRCh38
rs34856290 Chr.8: 13012669-13012670 on Build GRCh38
rs952274558 Chr.8: 13012471-13012471 on Build GRCh38
rs574287639 Chr.8: 13012431-13012431 on Build GRCh38
rs751131595 Chr.8: 13012685-13012685 on Build GRCh38
rs927715760 Chr.8: 13012499-13012499 on Build GRCh38
rs757768846 Chr.8: 13012606-13012606 on Build GRCh38
rs1005008137 Chr.8: 13012473-13012473 on Build GRCh38
rs143458275 Chr.8: 13012361-13012361 on Build GRCh38
rs180946026 Chr.8: 13012491-13012491 on Build GRCh38
rs529626958 Chr.8: 13012402-13012402 on Build GRCh38
rs749084984 Chr.8: 13012757-13012757 on Build GRCh38
rs370254024 Chr.8: 13012694-13012694 on Build GRCh38
rs768771870 Chr.8: 13012771-13012771 on Build GRCh38
rs775754482 Chr.8: 13012726-13012726 on Build GRCh38
rs372421670 Chr.8: 13012769-13012769 on Build GRCh38
rs1015358539 Chr.8: 13012275-13012275 on Build GRCh38
rs780990592 Chr.8: 13012692-13012692 on Build GRCh38
rs1027860324 Chr.8: 13012387-13012387 on Build GRCh38
rs759795328 Chr.8: 13012646-13012646 on Build GRCh38
rs755755518 Chr.8: 13012750-13012750 on Build GRCh38
rs760844128 Chr.8: 13012728-13012728 on Build GRCh38
rs773119260 Chr.8: 13012653-13012657 on Build GRCh38
rs1032472074 Chr.8: 13012443-13012443 on Build GRCh38
rs1023576745 Chr.8: 13012419-13012419 on Build GRCh38
rs2460339 Chr.8: 13012405-13012405 on Build GRCh38
rs751760323 Chr.8: 13012644-13012644 on Build GRCh38
rs371980778 Chr.8: 13012721-13012721 on Build GRCh38
rs900551274 Chr.8: 13012463-13012463 on Build GRCh38
rs757109794 Chr.8: 13012758-13012758 on Build GRCh38
rs996115483 Chr.8: 13012467-13012467 on Build GRCh38
rs112494740 Chr.8: 13012358-13012358 on Build GRCh38
rs771848255 Chr.8: 13012723-13012723 on Build GRCh38
rs528098440 Chr.8: 13012528-13012528 on Build GRCh38
rs754452363 Chr.8: 13012567-13012567 on Build GRCh38
rs764401021 Chr.8: 13012343-13012343 on Build GRCh38
rs111625359 Chr.8: 13012326-13012326 on Build GRCh38
rs1008919319 Chr.8: 13012351-13012351 on Build GRCh38
rs779749025 Chr.8: 13012656-13012656 on Build GRCh38
rs960701673 Chr.8: 13012280-13012280 on Build GRCh38
rs557489468 Chr.8: 13012342-13012342 on Build GRCh38
rs754335624 Chr.8: 13012682-13012682 on Build GRCh38
rs534314824 Chr.8: 13012791-13012791 on Build GRCh38
rs778338690 Chr.8: 13012652-13012652 on Build GRCh38
rs979595633 Chr.8: 13012305-13012305 on Build GRCh38
rs900566122 Chr.8: 13012547-13012547 on Build GRCh38
rs1045061732 Chr.8: 13012545-13012545 on Build GRCh38
rs558227333 Chr.8: 13012500-13012500 on Build GRCh38
rs767015114 Chr.8: 13012790-13012790 on Build GRCh38
rs902601297 Chr.8: 13012455-13012455 on Build GRCh38
rs754526740 Chr.8: 13012747-13012747 on Build GRCh38
rs932033001 Chr.8: 13012689-13012689 on Build GRCh38
rs12156420 Chr.8: 13012677-13012677 on Build GRCh38
rs1034784201 Chr.8: 13012627-13012627 on Build GRCh38
rs777692408 Chr.8: 13012632-13012632 on Build GRCh38
rs969445337 Chr.8: 13012300-13012300 on Build GRCh38
rs753276336 Chr.8: 13012517-13012517 on Build GRCh38
rs766985108 Chr.8: 13012748-13012748 on Build GRCh38
rs559636969 Chr.8: 13012426-13012426 on Build GRCh38
rs892541539 Chr.8: 13012552-13012552 on Build GRCh38
rs765935531 Chr.8: 13012684-13012684 on Build GRCh38
rs920902943 Chr.8: 13012461-13012461 on Build GRCh38
rs990459245 Chr.8: 13012501-13012501 on Build GRCh38
rs369928661 Chr.8: 13012783-13012783 on Build GRCh38
rs763595602 Chr.8: 13012788-13012788 on Build GRCh38
rs557834308 Chr.8: 13012374-13012374 on Build GRCh38
rs568817757 Chr.8: 13012561-13012561 on Build GRCh38
rs968142404 Chr.8: 13012813-13012813 on Build GRCh38
rs377631038 Chr.8: 13012664-13012664 on Build GRCh38
rs563026955 Chr.8: 13012449-13012449 on Build GRCh38
rs1055068095 Chr.8: 13012432-13012432 on Build GRCh38
rs749211078 Chr.8: 13012714-13012714 on Build GRCh38
rs778048153 Chr.8: 13012549-13012549 on Build GRCh38
rs1048271798 Chr.8: 13012386-13012386 on Build GRCh38
rs530487264 Chr.8: 13012460-13012460 on Build GRCh38
rs1040452807 Chr.8: 13012462-13012462 on Build GRCh38
rs772105016 Chr.8: 13012768-13012768 on Build GRCh38
rs1027995697 Chr.8: 13012623-13012623 on Build GRCh38
rs372978061 Chr.8: 13012804-13012804 on Build GRCh38
rs761798661 Chr.8: 13012703-13012703 on Build GRCh38
rs906453803 Chr.8: 13012310-13012310 on Build GRCh38
rs748992362 Chr.8: 13012666-13012666 on Build GRCh38
rs908304711 Chr.8: 13012411-13012411 on Build GRCh38
rs181428862 Chr.8: 13012680-13012680 on Build GRCh38
rs765091275 Chr.8: 13012597-13012597 on Build GRCh38
rs915151738 Chr.8: 13012437-13012437 on Build GRCh38

View Full Product Details