Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43406184-43406720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX101
Entrez Gene ID
83639
Gene Name
testis expressed 101
Gene Aliases
CT131, GTPR867, NYD-SP8, PRO1884, SGRG, SPATA44, TES101RP
UniGene
Hs.97978
Gene Chromosome Location
Chr.19: 43388611-43418615 on Build GRCh38
Transcripts
NM_031451.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6252244 c.15G>T p.Q5H Chr.19: 43406519-43406519 on Build GRCh38
712554 c.9G>A p.A3A Chr.19: 43406513-43406513 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs964071472 Chr.19: 43406609-43406609 on Build GRCh38
rs778496669 Chr.19: 43406456-43406456 on Build GRCh38
rs981694272 Chr.19: 43406409-43406409 on Build GRCh38
rs11669801 Chr.19: 43406521-43406521 on Build GRCh38
rs908785064 Chr.19: 43406687-43406687 on Build GRCh38
rs182286763 Chr.19: 43406440-43406440 on Build GRCh38
rs931504066 Chr.19: 43406281-43406281 on Build GRCh38
rs537945546 Chr.19: 43406577-43406577 on Build GRCh38
rs991776894 Chr.19: 43406654-43406654 on Build GRCh38
rs546539340 Chr.19: 43406363-43406363 on Build GRCh38
rs762933736 Chr.19: 43406557-43406560 on Build GRCh38
rs373254378 Chr.19: 43406640-43406640 on Build GRCh38
rs893219884 Chr.19: 43406326-43406326 on Build GRCh38
rs996120208 Chr.19: 43406324-43406324 on Build GRCh38
rs975678763 Chr.19: 43406268-43406268 on Build GRCh38
rs890332279 Chr.19: 43406598-43406598 on Build GRCh38
rs949126958 Chr.19: 43406303-43406303 on Build GRCh38
rs11673673 Chr.19: 43406497-43406497 on Build GRCh38
rs771571981 Chr.19: 43406465-43406465 on Build GRCh38
rs1008669137 Chr.19: 43406605-43406605 on Build GRCh38
rs62115683 Chr.19: 43406415-43406415 on Build GRCh38
rs201552091 Chr.19: 43406328-43406328 on Build GRCh38
rs141261739 Chr.19: 43406595-43406595 on Build GRCh38
rs201830210 Chr.19: 43406338-43406338 on Build GRCh38
rs920187013 Chr.19: 43406467-43406467 on Build GRCh38
rs762244326 Chr.19: 43406489-43406489 on Build GRCh38
rs112935810 Chr.19: 43406245-43406245 on Build GRCh38
rs187018847 Chr.19: 43406252-43406252 on Build GRCh38
rs908676972 Chr.19: 43406484-43406484 on Build GRCh38
rs8113556 Chr.19: 43406612-43406612 on Build GRCh38
rs964039172 Chr.19: 43406430-43406430 on Build GRCh38
rs1008104485 Chr.19: 43406297-43406297 on Build GRCh38
rs912526927 Chr.19: 43406260-43406260 on Build GRCh38
rs112447525 Chr.19: 43406401-43406401 on Build GRCh38
rs982531171 Chr.19: 43406680-43406680 on Build GRCh38
rs893695121 Chr.19: 43406353-43406353 on Build GRCh38
rs958933827 Chr.19: 43406652-43406652 on Build GRCh38
rs1020745724 Chr.19: 43406366-43406366 on Build GRCh38
rs1003433937 Chr.19: 43406642-43406642 on Build GRCh38
rs769055635 Chr.19: 43406471-43406471 on Build GRCh38
rs1046892759 Chr.19: 43406271-43406271 on Build GRCh38
rs763124935 Chr.19: 43406493-43406493 on Build GRCh38
rs1006655263 Chr.19: 43406225-43406225 on Build GRCh38
rs533529299 Chr.19: 43406608-43406608 on Build GRCh38
rs545095637 Chr.19: 43406312-43406312 on Build GRCh38
rs953085064 Chr.19: 43406633-43406633 on Build GRCh38
rs760297533 Chr.19: 43406470-43406470 on Build GRCh38
rs755211840 Chr.19: 43406503-43406503 on Build GRCh38
rs967463053 Chr.19: 43406209-43406209 on Build GRCh38
rs1026948052 Chr.19: 43406614-43406614 on Build GRCh38
rs528361277 Chr.19: 43406320-43406320 on Build GRCh38
rs1020799574 Chr.19: 43406626-43406626 on Build GRCh38
rs545113843 Chr.19: 43406684-43406684 on Build GRCh38
rs745326643 Chr.19: 43406524-43406524 on Build GRCh38
rs531094118 Chr.19: 43406677-43406678 on Build GRCh38
rs1009450103 Chr.19: 43406589-43406589 on Build GRCh38
rs983358562 Chr.19: 43406477-43406477 on Build GRCh38
rs1019655646 Chr.19: 43406425-43406425 on Build GRCh38
rs750456042 Chr.19: 43406491-43406491 on Build GRCh38
rs779492221 Chr.19: 43406541-43406541 on Build GRCh38
rs920161889 Chr.19: 43406277-43406277 on Build GRCh38
rs757200433 Chr.19: 43406513-43406513 on Build GRCh38
rs899748070 Chr.19: 43406582-43406582 on Build GRCh38
rs372792589 Chr.19: 43406468-43406468 on Build GRCh38
rs1032288860 Chr.19: 43406349-43406349 on Build GRCh38
rs967873815 Chr.19: 43406411-43406411 on Build GRCh38
rs557849028 Chr.19: 43406587-43406587 on Build GRCh38
rs751783249 Chr.19: 43406502-43406502 on Build GRCh38
rs771593058 Chr.19: 43406535-43406535 on Build GRCh38
rs967953657 Chr.19: 43406400-43406400 on Build GRCh38
rs958965871 Chr.19: 43406423-43406423 on Build GRCh38
rs1035888110 Chr.19: 43406644-43406644 on Build GRCh38
rs764195790 Chr.19: 43406329-43406329 on Build GRCh38
rs1009830902 Chr.19: 43406354-43406354 on Build GRCh38
rs189702326 Chr.19: 43406507-43406507 on Build GRCh38
rs952498536 Chr.19: 43406669-43406669 on Build GRCh38
rs775570159 Chr.19: 43406554-43406556 on Build GRCh38
rs986879312 Chr.19: 43406422-43406422 on Build GRCh38
rs573888169 Chr.19: 43406627-43406627 on Build GRCh38
rs17849541 Chr.19: 43406515-43406515 on Build GRCh38
rs1026619848 Chr.19: 43406655-43406655 on Build GRCh38
rs79190574 Chr.19: 43406325-43406325 on Build GRCh38
rs191755172 Chr.19: 43406432-43406432 on Build GRCh38
rs186536094 Chr.19: 43406464-43406464 on Build GRCh38
rs765747571 Chr.19: 43406490-43406490 on Build GRCh38
rs893714718 Chr.19: 43406583-43406583 on Build GRCh38
rs942693905 Chr.19: 43406262-43406262 on Build GRCh38
rs556131608 Chr.19: 43406668-43406668 on Build GRCh38
rs777049636 Chr.19: 43406488-43406488 on Build GRCh38
rs1047201866 Chr.19: 43406282-43406282 on Build GRCh38
rs886878329 Chr.19: 43406288-43406288 on Build GRCh38
rs536538632 Chr.19: 43406648-43406648 on Build GRCh38
rs73937417 Chr.19: 43406683-43406683 on Build GRCh38

View Full Product Details