Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 78633567-78634113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROBO1
Entrez Gene ID
6091
Gene Name
roundabout guidance receptor 1
Gene Aliases
DUTT1, SAX3
UniGene
Hs.744218
Gene Chromosome Location
Chr.3: 78597238-79767998 on Build GRCh38
Transcripts
NM_001145845.1 NM_002941.3 NM_133631.3 XM_006713277.2 XM_011533976.1 XM_011533977.2 XM_011533978.1 XM_011533979.1 XM_011533980.1 XM_017006982.1 XM_017006983.1 XM_017006984.1 XM_017006985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2827372 c.3329G>A p.R1110Q Chr.3: 78633952-78633952 on Build GRCh38
4120326 c.3481G>T p.G1161W Chr.3: 78633935-78633935 on Build GRCh38
731553 c.3277T>C p.Y1093H Chr.3: 78634004-78634004 on Build GRCh38
5865336 c.3331G>A p.G1111S Chr.3: 78633950-78633950 on Build GRCh38
4120324 c.3346G>T p.G1116W Chr.3: 78633935-78633935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748699403 Chr.3: 78633687-78633687 on Build GRCh38
rs758562546 Chr.3: 78633958-78633958 on Build GRCh38
rs866311711 Chr.3: 78633791-78633791 on Build GRCh38
rs543691634 Chr.3: 78633763-78633763 on Build GRCh38
rs910852309 Chr.3: 78633642-78633642 on Build GRCh38
rs754455499 Chr.3: 78633940-78633940 on Build GRCh38
rs1037830850 Chr.3: 78633677-78633677 on Build GRCh38
rs777236646 Chr.3: 78634019-78634019 on Build GRCh38
rs757383097 Chr.3: 78633967-78633967 on Build GRCh38
rs886453946 Chr.3: 78633646-78633646 on Build GRCh38
rs775718100 Chr.3: 78634042-78634042 on Build GRCh38
rs200790068 Chr.3: 78633882-78633882 on Build GRCh38
rs565172963 Chr.3: 78633825-78633826 on Build GRCh38
rs754730361 Chr.3: 78633989-78633989 on Build GRCh38
rs143714180 Chr.3: 78633651-78633651 on Build GRCh38
rs929912503 Chr.3: 78633607-78633607 on Build GRCh38
rs202246939 Chr.3: 78634013-78634013 on Build GRCh38
rs191486499 Chr.3: 78633670-78633670 on Build GRCh38
rs761406963 Chr.3: 78633898-78633898 on Build GRCh38
rs1010503133 Chr.3: 78633802-78633802 on Build GRCh38
rs1006661226 Chr.3: 78633854-78633854 on Build GRCh38
rs963770799 Chr.3: 78633872-78633872 on Build GRCh38
rs767619040 Chr.3: 78634071-78634071 on Build GRCh38
rs778705797 Chr.3: 78633990-78633990 on Build GRCh38
rs990388705 Chr.3: 78633654-78633654 on Build GRCh38
rs773965548 Chr.3: 78633891-78633891 on Build GRCh38
rs760141700 Chr.3: 78634036-78634036 on Build GRCh38
rs752914953 Chr.3: 78633954-78633954 on Build GRCh38
rs541051115 Chr.3: 78633728-78633728 on Build GRCh38
rs747778978 Chr.3: 78633996-78633996 on Build GRCh38
rs1018010396 Chr.3: 78633870-78633870 on Build GRCh38
rs779942722 Chr.3: 78633973-78633973 on Build GRCh38
rs771629310 Chr.3: 78634000-78634000 on Build GRCh38
rs572137145 Chr.3: 78633704-78633704 on Build GRCh38
rs751854747 Chr.3: 78634057-78634057 on Build GRCh38
rs772611537 Chr.3: 78633918-78633918 on Build GRCh38
rs1035865040 Chr.3: 78633752-78633752 on Build GRCh38
rs577441632 Chr.3: 78633648-78633648 on Build GRCh38
rs753729353 Chr.3: 78634081-78634081 on Build GRCh38
rs551262188 Chr.3: 78634086-78634086 on Build GRCh38
rs771566503 Chr.3: 78633899-78633899 on Build GRCh38
rs563382979 Chr.3: 78633825-78633825 on Build GRCh38
rs757299763 Chr.3: 78634070-78634070 on Build GRCh38
rs765639480 Chr.3: 78634037-78634037 on Build GRCh38
rs763439002 Chr.3: 78634049-78634049 on Build GRCh38
rs759947257 Chr.3: 78633953-78633953 on Build GRCh38
rs147941631 Chr.3: 78633638-78633638 on Build GRCh38
rs766990122 Chr.3: 78633929-78633929 on Build GRCh38
rs778702768 Chr.3: 78634088-78634088 on Build GRCh38
rs75633073 Chr.3: 78633858-78633858 on Build GRCh38
rs559809379 Chr.3: 78633881-78633881 on Build GRCh38
rs573043406 Chr.3: 78633716-78633716 on Build GRCh38
rs558629191 Chr.3: 78633673-78633673 on Build GRCh38
rs574818267 Chr.3: 78633731-78633731 on Build GRCh38
rs548204637 Chr.3: 78633964-78633964 on Build GRCh38
rs749266872 Chr.3: 78633976-78633976 on Build GRCh38
rs955197475 Chr.3: 78633982-78633982 on Build GRCh38
rs1020944927 Chr.3: 78633845-78633845 on Build GRCh38
rs922478773 Chr.3: 78633890-78633890 on Build GRCh38
rs897115800 Chr.3: 78633875-78633875 on Build GRCh38
rs1047399065 Chr.3: 78633637-78633637 on Build GRCh38
rs946201811 Chr.3: 78633711-78633711 on Build GRCh38
rs982748391 Chr.3: 78633987-78633987 on Build GRCh38
rs2660738 Chr.3: 78633730-78633730 on Build GRCh38
rs568106352 Chr.3: 78634005-78634005 on Build GRCh38
rs772985757 Chr.3: 78634002-78634002 on Build GRCh38
rs187827566 Chr.3: 78633853-78633853 on Build GRCh38
rs182559352 Chr.3: 78633798-78633798 on Build GRCh38
rs761177664 Chr.3: 78633927-78633927 on Build GRCh38
rs557315277 Chr.3: 78633587-78633587 on Build GRCh38
rs957047907 Chr.3: 78633998-78633998 on Build GRCh38
rs752071519 Chr.3: 78633876-78633876 on Build GRCh38
rs752249344 Chr.3: 78634015-78634017 on Build GRCh38

View Full Product Details