Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 22032715-22033245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHEX
Entrez Gene ID
5251
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH
UniGene
Hs.495834
Gene Chromosome Location
Chr.X: 22032327-22251310 on Build GRCh38
Transcripts
NM_000444.5 NM_001282754.1

SNP Detail

SNP ID Chromosome Location
rs752602189 Chr.X: 22032968-22032968 on Build GRCh38
rs1037250538 Chr.X: 22033058-22033058 on Build GRCh38
rs956260093 Chr.X: 22033217-22033217 on Build GRCh38
rs954554640 Chr.X: 22032937-22032937 on Build GRCh38
rs778229143 Chr.X: 22033164-22033164 on Build GRCh38
rs534550003 Chr.X: 22033130-22033130 on Build GRCh38
rs201391816 Chr.X: 22032969-22032969 on Build GRCh38
rs781744295 Chr.X: 22033149-22033149 on Build GRCh38
rs750612939 Chr.X: 22032974-22032974 on Build GRCh38
rs768927791 Chr.X: 22033144-22033144 on Build GRCh38
rs932851909 Chr.X: 22032927-22032927 on Build GRCh38
rs977996230 Chr.X: 22032822-22032822 on Build GRCh38
rs780051359 Chr.X: 22032985-22032985 on Build GRCh38
rs1016100906 Chr.X: 22032874-22032874 on Build GRCh38
rs192414303 Chr.X: 22032860-22032860 on Build GRCh38
rs147859619 Chr.X: 22033015-22033015 on Build GRCh38
rs935444170 Chr.X: 22033061-22033061 on Build GRCh38
rs923724897 Chr.X: 22032996-22032996 on Build GRCh38
rs185830047 Chr.X: 22032880-22032880 on Build GRCh38
rs1057515841 Chr.X: 22032996-22032996 on Build GRCh38
rs767546324 Chr.X: 22033083-22033083 on Build GRCh38
rs1008897142 Chr.X: 22033029-22033029 on Build GRCh38
rs1004675895 Chr.X: 22032862-22032862 on Build GRCh38
rs987230658 Chr.X: 22032948-22032948 on Build GRCh38
rs781538001 Chr.X: 22032995-22032995 on Build GRCh38
rs758549621 Chr.X: 22033134-22033134 on Build GRCh38
rs5951494 Chr.X: 22032973-22032973 on Build GRCh38
rs748950116 Chr.X: 22033165-22033165 on Build GRCh38
rs760556268 Chr.X: 22033087-22033087 on Build GRCh38
rs754991528 Chr.X: 22033145-22033145 on Build GRCh38
rs756528765 Chr.X: 22033162-22033162 on Build GRCh38
rs201251651 Chr.X: 22032988-22032988 on Build GRCh38
rs759110019 Chr.X: 22033078-22033078 on Build GRCh38
rs139862893 Chr.X: 22033086-22033086 on Build GRCh38
rs773927462 Chr.X: 22033069-22033069 on Build GRCh38
rs776842086 Chr.X: 22033185-22033185 on Build GRCh38
rs755577853 Chr.X: 22032989-22032989 on Build GRCh38
rs770448887 Chr.X: 22032752-22032752 on Build GRCh38
rs963279947 Chr.X: 22032877-22032877 on Build GRCh38
rs917868159 Chr.X: 22032738-22032738 on Build GRCh38
rs887200561 Chr.X: 22033177-22033177 on Build GRCh38
rs149541983 Chr.X: 22032898-22032898 on Build GRCh38
rs886043584 Chr.X: 22033072-22033072 on Build GRCh38
rs35301418 Chr.X: 22032885-22032886 on Build GRCh38
rs34610947 Chr.X: 22033064-22033065 on Build GRCh38
rs753106927 Chr.X: 22033157-22033157 on Build GRCh38
rs773033853 Chr.X: 22033049-22033049 on Build GRCh38
rs766584688 Chr.X: 22033143-22033143 on Build GRCh38
rs763757285 Chr.X: 22033110-22033110 on Build GRCh38
rs539005958 Chr.X: 22033109-22033109 on Build GRCh38
rs886043583 Chr.X: 22033038-22033038 on Build GRCh38
rs921596276 Chr.X: 22032903-22032903 on Build GRCh38
rs970554160 Chr.X: 22033154-22033154 on Build GRCh38
rs758504261 Chr.X: 22032983-22032983 on Build GRCh38
rs934671381 Chr.X: 22032881-22032881 on Build GRCh38
rs770026330 Chr.X: 22033023-22033023 on Build GRCh38
rs777384160 Chr.X: 22032804-22032804 on Build GRCh38
rs780387907 Chr.X: 22032798-22032798 on Build GRCh38
rs770920860 Chr.X: 22033052-22033052 on Build GRCh38
rs1003994925 Chr.X: 22033152-22033152 on Build GRCh38
rs1004644879 Chr.X: 22033187-22033187 on Build GRCh38
rs770573978 Chr.X: 22033063-22033063 on Build GRCh38
rs178710 Chr.X: 22032916-22032916 on Build GRCh38

View Full Product Details