Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6037634-6038129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLS1
Entrez Gene ID
54675
Gene Name
cardiolipin synthase 1
Gene Aliases
C20orf155, CLS, CLS1, GCD10, dJ967N21.6
UniGene
Hs.743956
Gene Chromosome Location
Chr.20: 6005938-6040053 on Build GRCh38
Transcripts
NM_001127458.1 NM_001323561.1 NM_001323562.1 NM_001323563.1 NM_001323564.1 NM_019095.5 NR_136617.1 XM_005260738.4

SNP Detail

SNP ID Chromosome Location
rs7272266 Chr.20: 6037874-6037874 on Build GRCh38
rs1047225767 Chr.20: 6038080-6038080 on Build GRCh38
rs151006316 Chr.20: 6037668-6037668 on Build GRCh38
rs928581969 Chr.20: 6037706-6037706 on Build GRCh38
rs759951023 Chr.20: 6037855-6037855 on Build GRCh38
rs560181880 Chr.20: 6037862-6037862 on Build GRCh38
rs1001379138 Chr.20: 6037822-6037822 on Build GRCh38
rs550749632 Chr.20: 6038001-6038001 on Build GRCh38
rs530573860 Chr.20: 6037754-6037754 on Build GRCh38
rs372958772 Chr.20: 6038097-6038097 on Build GRCh38
rs922868741 Chr.20: 6038062-6038062 on Build GRCh38
rs748561024 Chr.20: 6037997-6038000 on Build GRCh38
rs910045050 Chr.20: 6038081-6038081 on Build GRCh38
rs960258245 Chr.20: 6037865-6037865 on Build GRCh38
rs537912561 Chr.20: 6037841-6037841 on Build GRCh38
rs6053838 Chr.20: 6037859-6037859 on Build GRCh38
rs976440959 Chr.20: 6038008-6038008 on Build GRCh38
rs1007146382 Chr.20: 6037966-6037966 on Build GRCh38
rs2235816 Chr.20: 6038065-6038065 on Build GRCh38
rs978513066 Chr.20: 6037992-6037992 on Build GRCh38
rs888869329 Chr.20: 6037964-6037964 on Build GRCh38
rs35454173 Chr.20: 6037716-6037716 on Build GRCh38
rs968456103 Chr.20: 6037962-6037962 on Build GRCh38
rs1006090986 Chr.20: 6037804-6037804 on Build GRCh38
rs879468804 Chr.20: 6037717-6037717 on Build GRCh38
rs534129165 Chr.20: 6037705-6037705 on Build GRCh38
rs183681722 Chr.20: 6037843-6037843 on Build GRCh38
rs35553334 Chr.20: 6037716-6037717 on Build GRCh38
rs577668623 Chr.20: 6037847-6037847 on Build GRCh38
rs191462434 Chr.20: 6037716-6037716 on Build GRCh38
rs538769239 Chr.20: 6037990-6037990 on Build GRCh38
rs573632596 Chr.20: 6037807-6037807 on Build GRCh38
rs963723031 Chr.20: 6038002-6038002 on Build GRCh38
rs995520104 Chr.20: 6038037-6038037 on Build GRCh38
rs914307663 Chr.20: 6037758-6037758 on Build GRCh38
rs1020575271 Chr.20: 6037893-6037893 on Build GRCh38
rs905211190 Chr.20: 6037821-6037821 on Build GRCh38

View Full Product Details