Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37210808-37211403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEBPZ
Entrez Gene ID
10153
Gene Name
CCAAT/enhancer binding protein zeta
Gene Aliases
CBF, CBF2, HSP-CBF, NOC1
UniGene
Hs.135406
Gene Chromosome Location
Chr.2: 37201629-37231597 on Build GRCh38
Transcripts
NM_005760.2 NR_136316.1 XM_017003108.1 XM_017003109.1

Gene Symbol
CEBPZOS
Entrez Gene ID
100505876
Gene Name
CEBPZ opposite strand
Gene Aliases
CEBPZ-AS1
UniGene
Hs.666958
Gene Chromosome Location
Chr.2: 37196451-37216193 on Build GRCh38
Transcripts
NM_005760.2 NR_136316.1 XM_017003108.1 XM_017003109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1020710 c.2857G>T p.D953Y Chr.2: 37211026-37211026 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs183451635 Chr.2: 37211289-37211289 on Build GRCh38
rs149678461 Chr.2: 37211360-37211360 on Build GRCh38
rs992816035 Chr.2: 37211028-37211028 on Build GRCh38
rs140432204 Chr.2: 37211011-37211011 on Build GRCh38
rs748864469 Chr.2: 37210993-37210993 on Build GRCh38
rs574146978 Chr.2: 37211343-37211343 on Build GRCh38
rs763010341 Chr.2: 37210959-37210959 on Build GRCh38
rs371073683 Chr.2: 37211204-37211204 on Build GRCh38
rs768917155 Chr.2: 37211042-37211042 on Build GRCh38
rs927357877 Chr.2: 37210985-37210985 on Build GRCh38
rs921862807 Chr.2: 37211295-37211295 on Build GRCh38
rs1015061078 Chr.2: 37210866-37210866 on Build GRCh38
rs529135111 Chr.2: 37211124-37211124 on Build GRCh38
rs993240253 Chr.2: 37211333-37211333 on Build GRCh38
rs774292192 Chr.2: 37211111-37211111 on Build GRCh38
rs909082873 Chr.2: 37210889-37210889 on Build GRCh38
rs747503627 Chr.2: 37211038-37211038 on Build GRCh38
rs770117185 Chr.2: 37211075-37211075 on Build GRCh38
rs899370453 Chr.2: 37211244-37211244 on Build GRCh38
rs1034460979 Chr.2: 37211291-37211291 on Build GRCh38
rs770292476 Chr.2: 37211171-37211171 on Build GRCh38
rs752303203 Chr.2: 37210978-37210978 on Build GRCh38
rs954494985 Chr.2: 37211350-37211350 on Build GRCh38
rs763057403 Chr.2: 37211091-37211091 on Build GRCh38
rs116198115 Chr.2: 37211131-37211131 on Build GRCh38
rs899579788 Chr.2: 37211161-37211161 on Build GRCh38
rs139504436 Chr.2: 37211272-37211272 on Build GRCh38
rs921789126 Chr.2: 37210925-37210925 on Build GRCh38
rs1015177145 Chr.2: 37211217-37211217 on Build GRCh38
rs1005479227 Chr.2: 37210854-37210854 on Build GRCh38
rs773352325 Chr.2: 37210957-37210957 on Build GRCh38
rs958795137 Chr.2: 37211315-37211315 on Build GRCh38
rs748459356 Chr.2: 37211069-37211069 on Build GRCh38
rs984145741 Chr.2: 37210884-37210884 on Build GRCh38
rs531007625 Chr.2: 37210891-37210891 on Build GRCh38
rs767291145 Chr.2: 37210977-37210977 on Build GRCh38
rs201041713 Chr.2: 37211132-37211132 on Build GRCh38
rs781556976 Chr.2: 37211059-37211059 on Build GRCh38
rs755724384 Chr.2: 37210981-37210981 on Build GRCh38
rs142473640 Chr.2: 37210929-37210929 on Build GRCh38
rs776031565 Chr.2: 37210987-37210988 on Build GRCh38
rs760984788 Chr.2: 37211150-37211150 on Build GRCh38
rs770954341 Chr.2: 37211104-37211104 on Build GRCh38
rs563697027 Chr.2: 37210872-37210872 on Build GRCh38
rs951428928 Chr.2: 37210873-37210873 on Build GRCh38
rs777561605 Chr.2: 37210988-37210988 on Build GRCh38
rs1034514069 Chr.2: 37210855-37210855 on Build GRCh38
rs766355599 Chr.2: 37210961-37210961 on Build GRCh38
rs963462406 Chr.2: 37210867-37210867 on Build GRCh38
rs539364568 Chr.2: 37211184-37211184 on Build GRCh38
rs759457060 Chr.2: 37211121-37211121 on Build GRCh38
rs759072912 Chr.2: 37211205-37211205 on Build GRCh38
rs191323391 Chr.2: 37211201-37211201 on Build GRCh38
rs1039453817 Chr.2: 37211144-37211144 on Build GRCh38
rs756798088 Chr.2: 37211003-37211003 on Build GRCh38
rs933663180 Chr.2: 37211170-37211170 on Build GRCh38
rs761148301 Chr.2: 37211123-37211125 on Build GRCh38
rs974563684 Chr.2: 37210911-37210911 on Build GRCh38
rs201959156 Chr.2: 37210965-37210965 on Build GRCh38
rs754755465 Chr.2: 37210970-37210970 on Build GRCh38
rs765457339 Chr.2: 37211086-37211086 on Build GRCh38
rs945666053 Chr.2: 37211020-37211020 on Build GRCh38
rs1026115432 Chr.2: 37211159-37211159 on Build GRCh38
rs374450191 Chr.2: 37211114-37211114 on Build GRCh38

View Full Product Details