Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 134328-134878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMYND11
Entrez Gene ID
10771
Gene Name
zinc finger MYND-type containing 11
Gene Aliases
BRAM1, BS69, MRD30
UniGene
Hs.292265
Gene Chromosome Location
Chr.10: 134340-254637 on Build GRCh38
Transcripts
NM_001202465.1 NM_001202466.1 NM_006624.5 NM_212479.3 XM_005252359.4 XM_005252362.2 XM_006717376.2 XM_017015591.1

SNP Detail

SNP ID Chromosome Location
rs938752606 Chr.10: 134368-134368 on Build GRCh38
rs948540965 Chr.10: 134405-134405 on Build GRCh38
rs997903417 Chr.10: 134473-134473 on Build GRCh38
rs993228777 Chr.10: 134812-134812 on Build GRCh38
rs192752836 Chr.10: 134562-134562 on Build GRCh38
rs1049527442 Chr.10: 134746-134746 on Build GRCh38
rs948615019 Chr.10: 134616-134616 on Build GRCh38
rs545704944 Chr.10: 134681-134681 on Build GRCh38
rs963619757 Chr.10: 134553-134553 on Build GRCh38
rs1043277157 Chr.10: 134644-134644 on Build GRCh38
rs900867157 Chr.10: 134727-134727 on Build GRCh38
rs1026548023 Chr.10: 134779-134779 on Build GRCh38
rs182548202 Chr.10: 134389-134389 on Build GRCh38
rs1043007033 Chr.10: 134407-134407 on Build GRCh38
rs889529492 Chr.10: 134747-134747 on Build GRCh38
rs904431005 Chr.10: 134427-134427 on Build GRCh38
rs1008129822 Chr.10: 134522-134522 on Build GRCh38
rs963505584 Chr.10: 134766-134766 on Build GRCh38
rs576594485 Chr.10: 134634-134634 on Build GRCh38
rs997790734 Chr.10: 134728-134728 on Build GRCh38
rs993764040 Chr.10: 134768-134768 on Build GRCh38
rs960004390 Chr.10: 134608-134608 on Build GRCh38
rs1052936771 Chr.10: 134721-134721 on Build GRCh38
rs1057205032 Chr.10: 134384-134384 on Build GRCh38
rs376137094 Chr.10: 134366-134366 on Build GRCh38
rs782058975 Chr.10: 134647-134647 on Build GRCh38
rs199921348 Chr.10: 134624-134626 on Build GRCh38
rs1016552112 Chr.10: 134539-134539 on Build GRCh38
rs992935504 Chr.10: 134612-134612 on Build GRCh38
rs1016848358 Chr.10: 134752-134752 on Build GRCh38
rs750631936 Chr.10: 134404-134404 on Build GRCh38
rs919268783 Chr.10: 134603-134603 on Build GRCh38
rs960056361 Chr.10: 134796-134796 on Build GRCh38
rs574585332 Chr.10: 134515-134515 on Build GRCh38
rs535096365 Chr.10: 134579-134579 on Build GRCh38
rs187880522 Chr.10: 134517-134517 on Build GRCh38
rs1007765594 Chr.10: 134748-134748 on Build GRCh38
rs916037198 Chr.10: 134815-134815 on Build GRCh38
rs535896991 Chr.10: 134394-134394 on Build GRCh38
rs972271584 Chr.10: 134555-134555 on Build GRCh38
rs998514595 Chr.10: 134466-134466 on Build GRCh38
rs562398904 Chr.10: 134731-134731 on Build GRCh38
rs553706008 Chr.10: 134633-134633 on Build GRCh38
rs900811413 Chr.10: 134471-134471 on Build GRCh38
rs934585711 Chr.10: 134708-134708 on Build GRCh38
rs1052720445 Chr.10: 134467-134467 on Build GRCh38
rs915967545 Chr.10: 134613-134613 on Build GRCh38
rs756253653 Chr.10: 134559-134560 on Build GRCh38
rs953596702 Chr.10: 134521-134521 on Build GRCh38

View Full Product Details