Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 4471255-4471833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STX18
Entrez Gene ID
53407
Gene Name
syntaxin 18
Gene Aliases
Ufe1
UniGene
Hs.584913
Gene Chromosome Location
Chr.4: 4418968-4542338 on Build GRCh38
Transcripts
NM_016930.2 XM_011513480.1 XM_011513483.1 XM_011513485.2 XM_017008296.1 XM_017008297.1 XM_017008298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6792586 c.202C>T p.L68L Chr.4: 4471673-4471673 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs192556293 Chr.4: 4471645-4471645 on Build GRCh38
rs907518889 Chr.4: 4471542-4471542 on Build GRCh38
rs894379158 Chr.4: 4471316-4471316 on Build GRCh38
rs940420068 Chr.4: 4471387-4471387 on Build GRCh38
rs6816884 Chr.4: 4471489-4471489 on Build GRCh38
rs573948274 Chr.4: 4471525-4471525 on Build GRCh38
rs775390555 Chr.4: 4471631-4471631 on Build GRCh38
rs772234266 Chr.4: 4471522-4471522 on Build GRCh38
rs1048900993 Chr.4: 4471311-4471311 on Build GRCh38
rs766548701 Chr.4: 4471667-4471667 on Build GRCh38
rs755353326 Chr.4: 4471670-4471670 on Build GRCh38
rs367985235 Chr.4: 4471752-4471752 on Build GRCh38
rs528592425 Chr.4: 4471647-4471647 on Build GRCh38
rs199983128 Chr.4: 4471706-4471706 on Build GRCh38
rs776783869 Chr.4: 4471754-4471754 on Build GRCh38
rs541587766 Chr.4: 4471620-4471621 on Build GRCh38
rs561237506 Chr.4: 4471506-4471506 on Build GRCh38
rs757721713 Chr.4: 4471409-4471409 on Build GRCh38
rs865972375 Chr.4: 4471794-4471794 on Build GRCh38
rs915434478 Chr.4: 4471467-4471467 on Build GRCh38
rs776280123 Chr.4: 4471582-4471582 on Build GRCh38
rs745482364 Chr.4: 4471704-4471704 on Build GRCh38
rs780871893 Chr.4: 4471703-4471703 on Build GRCh38
rs1013279460 Chr.4: 4471324-4471324 on Build GRCh38
rs565103209 Chr.4: 4471730-4471730 on Build GRCh38
rs766021661 Chr.4: 4471621-4471621 on Build GRCh38
rs780927116 Chr.4: 4471620-4471620 on Build GRCh38
rs370143875 Chr.4: 4471741-4471741 on Build GRCh38
rs932135153 Chr.4: 4471614-4471614 on Build GRCh38
rs1009132044 Chr.4: 4471702-4471702 on Build GRCh38
rs745529801 Chr.4: 4471621-4471621 on Build GRCh38
rs373854752 Chr.4: 4471748-4471748 on Build GRCh38
rs940369340 Chr.4: 4471571-4471571 on Build GRCh38
rs538823559 Chr.4: 4471380-4471380 on Build GRCh38
rs756467163 Chr.4: 4471378-4471378 on Build GRCh38
rs1037374210 Chr.4: 4471584-4471584 on Build GRCh38
rs755393133 Chr.4: 4471313-4471313 on Build GRCh38
rs1033502410 Chr.4: 4471763-4471763 on Build GRCh38
rs996362343 Chr.4: 4471404-4471404 on Build GRCh38
rs375456906 Chr.4: 4471668-4471668 on Build GRCh38
rs770634899 Chr.4: 4471604-4471604 on Build GRCh38
rs754193877 Chr.4: 4471392-4471392 on Build GRCh38
rs773369139 Chr.4: 4471658-4471658 on Build GRCh38
rs779250632 Chr.4: 4471440-4471440 on Build GRCh38
rs28716118 Chr.4: 4471470-4471470 on Build GRCh38
rs1031393019 Chr.4: 4471417-4471417 on Build GRCh38
rs753017704 Chr.4: 4471693-4471694 on Build GRCh38
rs753261753 Chr.4: 4471681-4471681 on Build GRCh38
rs779108951 Chr.4: 4471382-4471386 on Build GRCh38
rs899296053 Chr.4: 4471395-4471395 on Build GRCh38
rs749099018 Chr.4: 4471737-4471737 on Build GRCh38
rs764413654 Chr.4: 4471390-4471390 on Build GRCh38
rs544136129 Chr.4: 4471501-4471501 on Build GRCh38
rs746503431 Chr.4: 4471603-4471603 on Build GRCh38
rs774220039 Chr.4: 4471642-4471642 on Build GRCh38
rs982012456 Chr.4: 4471535-4471535 on Build GRCh38
rs957120515 Chr.4: 4471423-4471423 on Build GRCh38
rs752845319 Chr.4: 4471377-4471377 on Build GRCh38
rs1050646596 Chr.4: 4471619-4471619 on Build GRCh38
rs370848426 Chr.4: 4471756-4471756 on Build GRCh38
rs879333814 Chr.4: 4471411-4471411 on Build GRCh38
rs749081366 Chr.4: 4471632-4471632 on Build GRCh38
rs748102771 Chr.4: 4471742-4471742 on Build GRCh38
rs144764754 Chr.4: 4471641-4471641 on Build GRCh38
rs879922298 Chr.4: 4471394-4471394 on Build GRCh38
rs1008891022 Chr.4: 4471428-4471428 on Build GRCh38
rs543048646 Chr.4: 4471572-4471572 on Build GRCh38
rs769664129 Chr.4: 4471629-4471629 on Build GRCh38
rs567997599 Chr.4: 4471790-4471790 on Build GRCh38
rs958106323 Chr.4: 4471493-4471493 on Build GRCh38
rs767683640 Chr.4: 4471307-4471307 on Build GRCh38
rs779336452 Chr.4: 4471590-4471590 on Build GRCh38
rs187633905 Chr.4: 4471286-4471286 on Build GRCh38
rs757828412 Chr.4: 4471487-4471487 on Build GRCh38
rs890748552 Chr.4: 4471425-4471425 on Build GRCh38
rs773243834 Chr.4: 4471749-4471749 on Build GRCh38
rs184815406 Chr.4: 4471803-4471803 on Build GRCh38
rs1023443515 Chr.4: 4471793-4471793 on Build GRCh38
rs760917484 Chr.4: 4471661-4471661 on Build GRCh38
rs768232939 Chr.4: 4471530-4471530 on Build GRCh38
rs551481935 Chr.4: 4471676-4471676 on Build GRCh38
rs769668740 Chr.4: 4471773-4471773 on Build GRCh38
rs768336800 Chr.4: 4471739-4471739 on Build GRCh38
rs756769803 Chr.4: 4471700-4471700 on Build GRCh38
rs550948678 Chr.4: 4471759-4471759 on Build GRCh38

View Full Product Details