Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2650584-2650769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNQ1
Entrez Gene ID
3784
Gene Name
potassium voltage-gated channel subfamily Q member 1
Gene Aliases
ATFB1, ATFB3, JLNS1, KCNA8, KCNA9, KVLQT1, Kv1.9, Kv7.1, LQT, LQT1, RWS, SQT2, WRS
UniGene
Hs.95162
Gene Chromosome Location
Chr.11: 2444991-2849110 on Build GRCh38
Transcripts
NM_000218.2 NM_181798.1 NR_002728.3

Gene Symbol
KCNQ1OT1
Entrez Gene ID
10984
Gene Name
KCNQ1 opposite strand/antisense transcript 1 (non-protein coding)
Gene Aliases
KCNQ1-AS2, KCNQ10T1, Kncq1, KvDMR1, KvLQT1-AS, LIT1, NCRNA00012
UniGene
Hs.604823
Gene Chromosome Location
Chr.11: 2608328-2699998 on Build GRCh38
Transcripts
NM_000218.2 NM_181798.1 NR_002728.3

SNP Detail

SNP ID Chromosome Location
rs1018474268 Chr.11: 2650672-2650672 on Build GRCh38
rs146309250 Chr.11: 2650648-2650648 on Build GRCh38
rs999778511 Chr.11: 2650618-2650618 on Build GRCh38
rs976268610 Chr.11: 2650719-2650719 on Build GRCh38
rs563205745 Chr.11: 2650718-2650718 on Build GRCh38
rs922956833 Chr.11: 2650732-2650732 on Build GRCh38
rs971607044 Chr.11: 2650742-2650742 on Build GRCh38
rs902546471 Chr.11: 2650615-2650615 on Build GRCh38
rs112697133 Chr.11: 2650683-2650683 on Build GRCh38
rs964440813 Chr.11: 2650690-2650690 on Build GRCh38
rs1032965219 Chr.11: 2650650-2650650 on Build GRCh38
rs55720001 Chr.11: 2650626-2650626 on Build GRCh38
rs757797337 Chr.11: 2650612-2650612 on Build GRCh38
rs1007032881 Chr.11: 2650663-2650663 on Build GRCh38
rs370469940 Chr.11: 2650639-2650639 on Build GRCh38
rs541821158 Chr.11: 2650704-2650704 on Build GRCh38
rs958283003 Chr.11: 2650660-2650660 on Build GRCh38
rs775347354 Chr.11: 2650728-2650728 on Build GRCh38

View Full Product Details