Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7504221-7504791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf45
Entrez Gene ID
374877
Gene Name
chromosome 19 open reading frame 45
Gene Aliases
-
UniGene
Hs.631862
Gene Chromosome Location
Chr.19: 7497559-7508457 on Build GRCh38
Transcripts
NM_198534.2 XM_005272473.3 XM_011527993.1 XM_011527995.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3541252 c.724G>A p.D242N Chr.19: 7504395-7504395 on Build GRCh38
5527604 c.645C>T p.S215S Chr.19: 7504316-7504316 on Build GRCh38
7197396 c.720C>T p.S240S Chr.19: 7504349-7504349 on Build GRCh38
1003237 c.695C>T p.S232L Chr.19: 7504366-7504366 on Build GRCh38
7197397 c.678C>T p.S226S Chr.19: 7504349-7504349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753272794 Chr.19: 7504272-7504273 on Build GRCh38
rs747788149 Chr.19: 7504351-7504351 on Build GRCh38
rs538696395 Chr.19: 7504446-7504446 on Build GRCh38
rs748916596 Chr.19: 7504411-7504411 on Build GRCh38
rs754499660 Chr.19: 7504341-7504341 on Build GRCh38
rs371844476 Chr.19: 7504293-7504293 on Build GRCh38
rs368329070 Chr.19: 7504542-7504542 on Build GRCh38
rs151114514 Chr.19: 7504600-7504600 on Build GRCh38
rs1016681159 Chr.19: 7504735-7504735 on Build GRCh38
rs963737541 Chr.19: 7504504-7504504 on Build GRCh38
rs768720497 Chr.19: 7504456-7504456 on Build GRCh38
rs999205919 Chr.19: 7504722-7504722 on Build GRCh38
rs969675848 Chr.19: 7504407-7504407 on Build GRCh38
rs142175123 Chr.19: 7504630-7504630 on Build GRCh38
rs746644970 Chr.19: 7504286-7504286 on Build GRCh38
rs200098882 Chr.19: 7504412-7504412 on Build GRCh38
rs368913826 Chr.19: 7504274-7504274 on Build GRCh38
rs528565917 Chr.19: 7504273-7504273 on Build GRCh38
rs914056840 Chr.19: 7504508-7504508 on Build GRCh38
rs651541 Chr.19: 7504663-7504663 on Build GRCh38
rs1005747359 Chr.19: 7504733-7504733 on Build GRCh38
rs958304794 Chr.19: 7504498-7504498 on Build GRCh38
rs542602860 Chr.19: 7504255-7504255 on Build GRCh38
rs762021719 Chr.19: 7504462-7504462 on Build GRCh38
rs745311823 Chr.19: 7504384-7504385 on Build GRCh38
rs531077068 Chr.19: 7504661-7504661 on Build GRCh38
rs769584948 Chr.19: 7504355-7504355 on Build GRCh38
rs895623140 Chr.19: 7504660-7504660 on Build GRCh38
rs776860888 Chr.19: 7504393-7504393 on Build GRCh38
rs111888890 Chr.19: 7504755-7504755 on Build GRCh38
rs558383406 Chr.19: 7504470-7504470 on Build GRCh38
rs765574039 Chr.19: 7504395-7504395 on Build GRCh38
rs762901994 Chr.19: 7504277-7504277 on Build GRCh38
rs955923713 Chr.19: 7504759-7504759 on Build GRCh38
rs757929954 Chr.19: 7504258-7504258 on Build GRCh38
rs933489880 Chr.19: 7504579-7504579 on Build GRCh38
rs750900033 Chr.19: 7504319-7504319 on Build GRCh38
rs770840066 Chr.19: 7504361-7504361 on Build GRCh38
rs886928102 Chr.19: 7504732-7504732 on Build GRCh38
rs757370816 Chr.19: 7504414-7504414 on Build GRCh38
rs867947795 Chr.19: 7504633-7504633 on Build GRCh38
rs484870 Chr.19: 7504396-7504396 on Build GRCh38
rs754217137 Chr.19: 7504244-7504244 on Build GRCh38
rs922174445 Chr.19: 7504570-7504570 on Build GRCh38
rs115122169 Chr.19: 7504756-7504756 on Build GRCh38
rs1013989335 Chr.19: 7504380-7504380 on Build GRCh38
rs913384774 Chr.19: 7504598-7504598 on Build GRCh38
rs577774313 Chr.19: 7504503-7504503 on Build GRCh38
rs576872861 Chr.19: 7504563-7504563 on Build GRCh38
rs372351047 Chr.19: 7504265-7504265 on Build GRCh38
rs755609412 Chr.19: 7504387-7504388 on Build GRCh38
rs199982930 Chr.19: 7504345-7504345 on Build GRCh38
rs749146630 Chr.19: 7504360-7504360 on Build GRCh38
rs760802665 Chr.19: 7504287-7504287 on Build GRCh38
rs760776068 Chr.19: 7504366-7504366 on Build GRCh38
rs746595606 Chr.19: 7504268-7504268 on Build GRCh38
rs757597839 Chr.19: 7504301-7504301 on Build GRCh38
rs774321062 Chr.19: 7504363-7504363 on Build GRCh38
rs377695560 Chr.19: 7504437-7504437 on Build GRCh38
rs374565357 Chr.19: 7504429-7504429 on Build GRCh38
rs770752285 Chr.19: 7504436-7504436 on Build GRCh38
rs760665139 Chr.19: 7504525-7504525 on Build GRCh38
rs397859470 Chr.19: 7504720-7504720 on Build GRCh38
rs758793040 Chr.19: 7504376-7504383 on Build GRCh38
rs146786010 Chr.19: 7504367-7504367 on Build GRCh38
rs138961688 Chr.19: 7504621-7504621 on Build GRCh38
rs543611975 Chr.19: 7504562-7504562 on Build GRCh38
rs111327568 Chr.19: 7504591-7504591 on Build GRCh38
rs776575522 Chr.19: 7504461-7504461 on Build GRCh38
rs369856876 Chr.19: 7504426-7504426 on Build GRCh38
rs749059650 Chr.19: 7504430-7504430 on Build GRCh38
rs764354714 Chr.19: 7504290-7504290 on Build GRCh38
rs780775883 Chr.19: 7504335-7504335 on Build GRCh38
rs766975635 Chr.19: 7504397-7504397 on Build GRCh38
rs145077335 Chr.19: 7504359-7504359 on Build GRCh38
rs566370488 Chr.19: 7504441-7504441 on Build GRCh38
rs933437305 Chr.19: 7504308-7504308 on Build GRCh38
rs1046902435 Chr.19: 7504729-7504729 on Build GRCh38
rs144306198 Chr.19: 7504601-7504601 on Build GRCh38
rs765539917 Chr.19: 7504313-7504313 on Build GRCh38
rs11296886 Chr.19: 7504706-7504706 on Build GRCh38
rs767517011 Chr.19: 7504280-7504280 on Build GRCh38
rs754472532 Chr.19: 7504420-7504420 on Build GRCh38
rs146389310 Chr.19: 7504631-7504631 on Build GRCh38
rs374712724 Chr.19: 7504271-7504271 on Build GRCh38
rs141277600 Chr.19: 7504320-7504320 on Build GRCh38
rs944230301 Chr.19: 7504530-7504530 on Build GRCh38
rs768354509 Chr.19: 7504269-7504269 on Build GRCh38
rs746403686 Chr.19: 7504257-7504257 on Build GRCh38
rs780760385 Chr.19: 7504381-7504382 on Build GRCh38
rs779720302 Chr.19: 7504388-7504389 on Build GRCh38
rs368627403 Chr.19: 7504254-7504254 on Build GRCh38
rs536587746 Chr.19: 7504425-7504425 on Build GRCh38

View Full Product Details