Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75832466-75832970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO6
Entrez Gene ID
4646
Gene Name
myosin VI
Gene Aliases
DFNA22, DFNB37
UniGene
Hs.149387
Gene Chromosome Location
Chr.6: 75749176-75919537 on Build GRCh38
Transcripts
NM_001300899.1 NM_004999.3 XM_005248719.3 XM_005248720.3 XM_005248721.3 XM_005248722.3 XM_005248724.3 XM_005248726.3 XM_017010899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5357311 c.497+1G>A p.? Chr.6: 75832948-75832948 on Build GRCh38
4917338 c.457T>A p.S153T Chr.6: 75832907-75832907 on Build GRCh38
5357312 c.497+1G>A p.? Chr.6: 75832948-75832948 on Build GRCh38
5405858 c.406C>T p.R136* Chr.6: 75832856-75832856 on Build GRCh38
1239531 c.432T>C p.S144S Chr.6: 75832882-75832882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941721200 Chr.6: 75832877-75832877 on Build GRCh38
rs773676437 Chr.6: 75832933-75832933 on Build GRCh38
rs923151446 Chr.6: 75832774-75832774 on Build GRCh38
rs565770950 Chr.6: 75832891-75832891 on Build GRCh38
rs956746324 Chr.6: 75832742-75832742 on Build GRCh38
rs921552820 Chr.6: 75832491-75832491 on Build GRCh38
rs778814452 Chr.6: 75832845-75832845 on Build GRCh38
rs547428643 Chr.6: 75832726-75832726 on Build GRCh38
rs760763107 Chr.6: 75832754-75832754 on Build GRCh38
rs934819356 Chr.6: 75832583-75832583 on Build GRCh38
rs1044413272 Chr.6: 75832678-75832678 on Build GRCh38
rs868572055 Chr.6: 75832917-75832917 on Build GRCh38
rs905821087 Chr.6: 75832708-75832708 on Build GRCh38
rs868310653 Chr.6: 75832679-75832679 on Build GRCh38
rs749136566 Chr.6: 75832924-75832924 on Build GRCh38
rs995168878 Chr.6: 75832756-75832756 on Build GRCh38
rs979387069 Chr.6: 75832694-75832694 on Build GRCh38
rs748103963 Chr.6: 75832793-75832793 on Build GRCh38
rs950063619 Chr.6: 75832664-75832664 on Build GRCh38
rs759060536 Chr.6: 75832812-75832812 on Build GRCh38
rs769916641 Chr.6: 75832796-75832796 on Build GRCh38
rs876657710 Chr.6: 75832908-75832908 on Build GRCh38
rs969386764 Chr.6: 75832632-75832632 on Build GRCh38
rs765333476 Chr.6: 75832852-75832852 on Build GRCh38
rs1013878948 Chr.6: 75832546-75832546 on Build GRCh38
rs764635911 Chr.6: 75832826-75832826 on Build GRCh38
rs780422125 Chr.6: 75832868-75832868 on Build GRCh38
rs926122484 Chr.6: 75832562-75832562 on Build GRCh38
rs758630450 Chr.6: 75832856-75832856 on Build GRCh38
rs752991210 Chr.6: 75832818-75832818 on Build GRCh38
rs397517053 Chr.6: 75832920-75832920 on Build GRCh38
rs917321698 Chr.6: 75832627-75832627 on Build GRCh38
rs112278668 Chr.6: 75832797-75832797 on Build GRCh38
rs1004700630 Chr.6: 75832798-75832798 on Build GRCh38
rs201507590 Chr.6: 75832925-75832925 on Build GRCh38
rs187196436 Chr.6: 75832519-75832519 on Build GRCh38
rs767881103 Chr.6: 75832814-75832814 on Build GRCh38
rs111829089 Chr.6: 75832611-75832611 on Build GRCh38
rs746119627 Chr.6: 75832850-75832850 on Build GRCh38
rs200963569 Chr.6: 75832915-75832915 on Build GRCh38
rs557373025 Chr.6: 75832720-75832720 on Build GRCh38
rs754270544 Chr.6: 75832832-75832832 on Build GRCh38
rs762689660 Chr.6: 75832555-75832555 on Build GRCh38
rs757389132 Chr.6: 75832837-75832837 on Build GRCh38
rs1048941658 Chr.6: 75832768-75832768 on Build GRCh38
rs933144845 Chr.6: 75832805-75832805 on Build GRCh38
rs774084674 Chr.6: 75832811-75832811 on Build GRCh38
rs978998265 Chr.6: 75832554-75832554 on Build GRCh38
rs182235369 Chr.6: 75832506-75832506 on Build GRCh38
rs886289512 Chr.6: 75832785-75832785 on Build GRCh38
rs748011960 Chr.6: 75832889-75832889 on Build GRCh38
rs865934913 Chr.6: 75832531-75832531 on Build GRCh38
rs575716193 Chr.6: 75832781-75832781 on Build GRCh38
rs73751027 Chr.6: 75832592-75832592 on Build GRCh38
rs756460203 Chr.6: 75832822-75832822 on Build GRCh38

View Full Product Details