Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28506110-28506600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCC1
Entrez Gene ID
1104
Gene Name
regulator of chromosome condensation 1
Gene Aliases
CHC1, RCC1-I, SNHG3-RCC1
UniGene
Hs.469723
Gene Chromosome Location
Chr.1: 28505943-28539196 on Build GRCh38
Transcripts
NM_001048199.2 NR_030725.1 NR_030726.1 NR_002909.2 NR_036473.1

Gene Symbol
SNHG3
Entrez Gene ID
8420
Gene Name
small nucleolar RNA host gene 3
Gene Aliases
NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
UniGene
Hs.469723
Gene Chromosome Location
Chr.1: 28505943-28510892 on Build GRCh38
Transcripts
NM_001048199.2 NR_030725.1 NR_030726.1 NR_002909.2 NR_036473.1

SNP Detail

SNP ID Chromosome Location
rs142712867 Chr.1: 28506294-28506294 on Build GRCh38
rs546184610 Chr.1: 28506487-28506487 on Build GRCh38
rs961578387 Chr.1: 28506153-28506153 on Build GRCh38
rs1046839957 Chr.1: 28506145-28506145 on Build GRCh38
rs1038523700 Chr.1: 28506560-28506560 on Build GRCh38
rs899612693 Chr.1: 28506562-28506562 on Build GRCh38
rs774203093 Chr.1: 28506202-28506203 on Build GRCh38
rs970630114 Chr.1: 28506498-28506498 on Build GRCh38
rs751024327 Chr.1: 28506217-28506217 on Build GRCh38
rs765371028 Chr.1: 28506509-28506509 on Build GRCh38
rs373823348 Chr.1: 28506278-28506278 on Build GRCh38
rs565714290 Chr.1: 28506169-28506169 on Build GRCh38
rs997177221 Chr.1: 28506179-28506179 on Build GRCh38
rs762410440 Chr.1: 28506200-28506201 on Build GRCh38
rs557662811 Chr.1: 28506316-28506316 on Build GRCh38
rs761656967 Chr.1: 28506173-28506173 on Build GRCh38
rs397863560 Chr.1: 28506216-28506217 on Build GRCh38
rs770683956 Chr.1: 28506202-28506202 on Build GRCh38
rs996563589 Chr.1: 28506398-28506398 on Build GRCh38
rs992973993 Chr.1: 28506160-28506160 on Build GRCh38
rs1032973821 Chr.1: 28506475-28506475 on Build GRCh38
rs1017031446 Chr.1: 28506167-28506167 on Build GRCh38
rs758815332 Chr.1: 28506542-28506543 on Build GRCh38
rs6699748 Chr.1: 28506558-28506558 on Build GRCh38
rs1029456986 Chr.1: 28506182-28506182 on Build GRCh38
rs1014424811 Chr.1: 28506473-28506473 on Build GRCh38
rs564932856 Chr.1: 28506363-28506363 on Build GRCh38
rs926350144 Chr.1: 28506202-28506202 on Build GRCh38
rs941122126 Chr.1: 28506351-28506351 on Build GRCh38
rs6696931 Chr.1: 28506175-28506175 on Build GRCh38
rs879342640 Chr.1: 28506189-28506192 on Build GRCh38
rs899502184 Chr.1: 28506391-28506391 on Build GRCh38
rs574922680 Chr.1: 28506221-28506221 on Build GRCh38
rs957784065 Chr.1: 28506514-28506514 on Build GRCh38
rs1047285147 Chr.1: 28506320-28506320 on Build GRCh38
rs891070216 Chr.1: 28506410-28506410 on Build GRCh38
rs886887414 Chr.1: 28506148-28506148 on Build GRCh38
rs1020311040 Chr.1: 28506197-28506197 on Build GRCh38
rs1050252299 Chr.1: 28506342-28506342 on Build GRCh38
rs559176556 Chr.1: 28506523-28506523 on Build GRCh38
rs764091943 Chr.1: 28506529-28506529 on Build GRCh38
rs999602603 Chr.1: 28506539-28506548 on Build GRCh38
rs542995461 Chr.1: 28506434-28506434 on Build GRCh38
rs991534709 Chr.1: 28506489-28506489 on Build GRCh38
rs559647383 Chr.1: 28506313-28506313 on Build GRCh38
rs559517521 Chr.1: 28506471-28506471 on Build GRCh38
rs762371430 Chr.1: 28506215-28506215 on Build GRCh38
rs1053746143 Chr.1: 28506192-28506192 on Build GRCh38
rs6656720 Chr.1: 28506185-28506185 on Build GRCh38
rs759838536 Chr.1: 28506494-28506494 on Build GRCh38
rs967406366 Chr.1: 28506464-28506464 on Build GRCh38
rs756421744 Chr.1: 28506297-28506297 on Build GRCh38
rs537647807 Chr.1: 28506295-28506295 on Build GRCh38
rs188359143 Chr.1: 28506150-28506150 on Build GRCh38
rs535870908 Chr.1: 28506477-28506477 on Build GRCh38
rs780219314 Chr.1: 28506481-28506481 on Build GRCh38
rs781643299 Chr.1: 28506241-28506241 on Build GRCh38
rs567006023 Chr.1: 28506467-28506467 on Build GRCh38
rs980922991 Chr.1: 28506202-28506205 on Build GRCh38
rs989443353 Chr.1: 28506248-28506248 on Build GRCh38
rs1033836215 Chr.1: 28506508-28506508 on Build GRCh38
rs759398942 Chr.1: 28506201-28506202 on Build GRCh38
rs1052400752 Chr.1: 28506236-28506236 on Build GRCh38
rs976661308 Chr.1: 28506545-28506545 on Build GRCh38
rs56695711 Chr.1: 28506216-28506216 on Build GRCh38
rs566424430 Chr.1: 28506505-28506505 on Build GRCh38
rs147351130 Chr.1: 28506542-28506542 on Build GRCh38
rs115068088 Chr.1: 28506519-28506519 on Build GRCh38
rs765984083 Chr.1: 28506216-28506216 on Build GRCh38
rs922420060 Chr.1: 28506301-28506301 on Build GRCh38
rs745904461 Chr.1: 28506362-28506362 on Build GRCh38
rs180771311 Chr.1: 28506331-28506331 on Build GRCh38
rs762954319 Chr.1: 28506520-28506520 on Build GRCh38
rs35746493 Chr.1: 28506286-28506287 on Build GRCh38
rs2840757 Chr.1: 28506411-28506411 on Build GRCh38
rs779097106 Chr.1: 28506356-28506356 on Build GRCh38
rs761201143 Chr.1: 28506443-28506443 on Build GRCh38
rs1028356703 Chr.1: 28506135-28506135 on Build GRCh38
rs967458783 Chr.1: 28506273-28506273 on Build GRCh38
rs925960015 Chr.1: 28506289-28506289 on Build GRCh38
rs1005978149 Chr.1: 28506385-28506385 on Build GRCh38
rs987967324 Chr.1: 28506186-28506186 on Build GRCh38
rs922483472 Chr.1: 28506546-28506546 on Build GRCh38
rs1038008556 Chr.1: 28506372-28506372 on Build GRCh38
rs763982811 Chr.1: 28506220-28506224 on Build GRCh38
rs950299586 Chr.1: 28506507-28506507 on Build GRCh38
rs892717216 Chr.1: 28506200-28506200 on Build GRCh38
rs771044997 Chr.1: 28506338-28506338 on Build GRCh38
rs769309515 Chr.1: 28506303-28506303 on Build GRCh38
rs978761121 Chr.1: 28506283-28506283 on Build GRCh38
rs752876439 Chr.1: 28506511-28506511 on Build GRCh38
rs772696597 Chr.1: 28506374-28506374 on Build GRCh38
rs775229064 Chr.1: 28506202-28506204 on Build GRCh38
rs917488230 Chr.1: 28506493-28506493 on Build GRCh38
rs945639023 Chr.1: 28506233-28506233 on Build GRCh38
rs1029524876 Chr.1: 28506401-28506401 on Build GRCh38
rs936316857 Chr.1: 28506220-28506220 on Build GRCh38

View Full Product Details