Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2761026-2761521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC283440
Entrez Gene ID
283440
Gene Name
uncharacterized LOC283440
Gene Aliases
-
UniGene
Hs.658536
Gene Chromosome Location
Chr.12: 2761200-2771669 on Build GRCh38
Transcripts
NR_033958.1

SNP Detail

SNP ID Chromosome Location
rs1015294145 Chr.12: 2761351-2761351 on Build GRCh38
rs879825577 Chr.12: 2761489-2761489 on Build GRCh38
rs763788696 Chr.12: 2761182-2761182 on Build GRCh38
rs924657459 Chr.12: 2761468-2761468 on Build GRCh38
rs983102744 Chr.12: 2761312-2761312 on Build GRCh38
rs558019028 Chr.12: 2761450-2761450 on Build GRCh38
rs547259833 Chr.12: 2761196-2761196 on Build GRCh38
rs374695009 Chr.12: 2761106-2761106 on Build GRCh38
rs908164697 Chr.12: 2761105-2761105 on Build GRCh38
rs779935974 Chr.12: 2761314-2761314 on Build GRCh38
rs750678441 Chr.12: 2761321-2761321 on Build GRCh38
rs537093605 Chr.12: 2761302-2761302 on Build GRCh38
rs369094473 Chr.12: 2761273-2761273 on Build GRCh38
rs955276717 Chr.12: 2761284-2761284 on Build GRCh38
rs557212417 Chr.12: 2761383-2761383 on Build GRCh38
rs758702827 Chr.12: 2761354-2761354 on Build GRCh38
rs879157815 Chr.12: 2761225-2761225 on Build GRCh38
rs1042346982 Chr.12: 2761467-2761467 on Build GRCh38
rs74054736 Chr.12: 2761087-2761087 on Build GRCh38
rs1012331741 Chr.12: 2761163-2761163 on Build GRCh38
rs896087554 Chr.12: 2761201-2761201 on Build GRCh38
rs548420806 Chr.12: 2761076-2761076 on Build GRCh38
rs772595932 Chr.12: 2761246-2761246 on Build GRCh38
rs765316046 Chr.12: 2761317-2761317 on Build GRCh38
rs906516561 Chr.12: 2761203-2761203 on Build GRCh38
rs776037572 Chr.12: 2761247-2761247 on Build GRCh38
rs559574792 Chr.12: 2761487-2761487 on Build GRCh38
rs753351347 Chr.12: 2761315-2761315 on Build GRCh38
rs920792345 Chr.12: 2761138-2761138 on Build GRCh38
rs886663211 Chr.12: 2761123-2761123 on Build GRCh38
rs539760509 Chr.12: 2761152-2761152 on Build GRCh38
rs936017743 Chr.12: 2761472-2761472 on Build GRCh38
rs371535836 Chr.12: 2761391-2761391 on Build GRCh38
rs940860870 Chr.12: 2761126-2761126 on Build GRCh38
rs1038310786 Chr.12: 2761438-2761438 on Build GRCh38
rs975081089 Chr.12: 2761050-2761050 on Build GRCh38
rs565571065 Chr.12: 2761235-2761235 on Build GRCh38
rs368046444 Chr.12: 2761355-2761355 on Build GRCh38
rs903172706 Chr.12: 2761234-2761234 on Build GRCh38
rs753552374 Chr.12: 2761336-2761336 on Build GRCh38
rs933668532 Chr.12: 2761322-2761322 on Build GRCh38
rs974222651 Chr.12: 2761118-2761118 on Build GRCh38
rs937734587 Chr.12: 2761408-2761408 on Build GRCh38
rs765209195 Chr.12: 2761325-2761328 on Build GRCh38
rs991628234 Chr.12: 2761412-2761412 on Build GRCh38
rs917378224 Chr.12: 2761449-2761449 on Build GRCh38
rs908121814 Chr.12: 2761378-2761378 on Build GRCh38
rs554406314 Chr.12: 2761248-2761248 on Build GRCh38
rs982477164 Chr.12: 2761091-2761091 on Build GRCh38
rs950316195 Chr.12: 2761215-2761215 on Build GRCh38
rs189727062 Chr.12: 2761103-2761103 on Build GRCh38
rs1033074048 Chr.12: 2761237-2761237 on Build GRCh38
rs937493926 Chr.12: 2761143-2761143 on Build GRCh38
rs116819690 Chr.12: 2761240-2761240 on Build GRCh38
rs926395542 Chr.12: 2761404-2761404 on Build GRCh38
rs561842039 Chr.12: 2761086-2761086 on Build GRCh38
rs975113611 Chr.12: 2761294-2761294 on Build GRCh38

View Full Product Details