Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41297982-41298489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OIP5-AS1
Entrez Gene ID
729082
Gene Name
OIP5 antisense RNA 1
Gene Aliases
cyrano
UniGene
Hs.380164
Gene Chromosome Location
Chr.15: 41284003-41299597 on Build GRCh38
Transcripts
NR_026757.1

SNP Detail

SNP ID Chromosome Location
rs1000173812 Chr.15: 41298295-41298295 on Build GRCh38
rs34536592 Chr.15: 41298100-41298101 on Build GRCh38
rs1047384665 Chr.15: 41298077-41298077 on Build GRCh38
rs571005027 Chr.15: 41298076-41298076 on Build GRCh38
rs941685710 Chr.15: 41298108-41298108 on Build GRCh38
rs930110983 Chr.15: 41298021-41298023 on Build GRCh38
rs541858786 Chr.15: 41298266-41298266 on Build GRCh38
rs200876596 Chr.15: 41298101-41298101 on Build GRCh38
rs141262189 Chr.15: 41298067-41298067 on Build GRCh38
rs533604368 Chr.15: 41298020-41298020 on Build GRCh38
rs993348458 Chr.15: 41298305-41298305 on Build GRCh38
rs746910551 Chr.15: 41298454-41298455 on Build GRCh38
rs540005084 Chr.15: 41298080-41298080 on Build GRCh38
rs890325125 Chr.15: 41298165-41298169 on Build GRCh38
rs371439141 Chr.15: 41298296-41298296 on Build GRCh38
rs748332406 Chr.15: 41298174-41298174 on Build GRCh38
rs951559561 Chr.15: 41298394-41298394 on Build GRCh38
rs530823453 Chr.15: 41298052-41298052 on Build GRCh38
rs1020165322 Chr.15: 41298231-41298231 on Build GRCh38
rs576643167 Chr.15: 41298219-41298219 on Build GRCh38
rs910139909 Chr.15: 41298419-41298420 on Build GRCh38
rs187068629 Chr.15: 41298365-41298365 on Build GRCh38
rs968565196 Chr.15: 41298265-41298265 on Build GRCh38
rs201843751 Chr.15: 41298102-41298102 on Build GRCh38
rs370123479 Chr.15: 41298302-41298302 on Build GRCh38
rs1037748491 Chr.15: 41298118-41298118 on Build GRCh38
rs1010014487 Chr.15: 41298182-41298182 on Build GRCh38
rs1024391215 Chr.15: 41298335-41298335 on Build GRCh38
rs183138535 Chr.15: 41298162-41298162 on Build GRCh38
rs982915057 Chr.15: 41298399-41298399 on Build GRCh38
rs900261188 Chr.15: 41298160-41298160 on Build GRCh38
rs941563175 Chr.15: 41298426-41298426 on Build GRCh38

View Full Product Details