Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60523653-60524154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PART1
Entrez Gene ID
25859
Gene Name
prostate androgen-regulated transcript 1 (non-protein coding)
Gene Aliases
NCRNA00206
UniGene
Hs.146312
Gene Chromosome Location
Chr.5: 60487713-60547657 on Build GRCh38
Transcripts
NR_024617.1 NR_028509.1

SNP Detail

SNP ID Chromosome Location
rs918942570 Chr.5: 60523885-60523885 on Build GRCh38
rs1011675619 Chr.5: 60523762-60523762 on Build GRCh38
rs920789078 Chr.5: 60523696-60523696 on Build GRCh38
rs747972138 Chr.5: 60523869-60523869 on Build GRCh38
rs188694332 Chr.5: 60523719-60523719 on Build GRCh38
rs987957935 Chr.5: 60523925-60523925 on Build GRCh38
rs1026380363 Chr.5: 60524119-60524119 on Build GRCh38
rs1014384102 Chr.5: 60523759-60523759 on Build GRCh38
rs984579356 Chr.5: 60523902-60523902 on Build GRCh38
rs543447565 Chr.5: 60524030-60524030 on Build GRCh38
rs1007021489 Chr.5: 60524076-60524076 on Build GRCh38
rs930859523 Chr.5: 60524010-60524010 on Build GRCh38
rs10516153 Chr.5: 60524034-60524034 on Build GRCh38
rs1019799164 Chr.5: 60524089-60524089 on Build GRCh38
rs7710967 Chr.5: 60524113-60524113 on Build GRCh38
rs192809701 Chr.5: 60524014-60524014 on Build GRCh38
rs746975061 Chr.5: 60523706-60523706 on Build GRCh38
rs141644855 Chr.5: 60523866-60523866 on Build GRCh38
rs1028540123 Chr.5: 60523911-60523911 on Build GRCh38
rs909027123 Chr.5: 60523904-60523904 on Build GRCh38
rs7728865 Chr.5: 60524114-60524114 on Build GRCh38
rs573328783 Chr.5: 60523710-60523710 on Build GRCh38
rs1007139185 Chr.5: 60523850-60523850 on Build GRCh38
rs1022189252 Chr.5: 60523827-60523827 on Build GRCh38
rs1053503356 Chr.5: 60523996-60523996 on Build GRCh38
rs950494712 Chr.5: 60523901-60523901 on Build GRCh38
rs911926900 Chr.5: 60523927-60523927 on Build GRCh38
rs532293739 Chr.5: 60524080-60524080 on Build GRCh38
rs745355414 Chr.5: 60523915-60523915 on Build GRCh38
rs996706401 Chr.5: 60523906-60523906 on Build GRCh38
rs569485805 Chr.5: 60523951-60523951 on Build GRCh38
rs891814442 Chr.5: 60524000-60524000 on Build GRCh38
rs540081942 Chr.5: 60523964-60523964 on Build GRCh38
rs1032910857 Chr.5: 60523876-60523876 on Build GRCh38
rs979986451 Chr.5: 60523916-60523916 on Build GRCh38
rs954264455 Chr.5: 60523847-60523847 on Build GRCh38
rs773436341 Chr.5: 60523933-60523933 on Build GRCh38
rs560994273 Chr.5: 60524009-60524009 on Build GRCh38
rs386688444 Chr.5: 60524113-60524114 on Build GRCh38
rs972309739 Chr.5: 60523989-60523989 on Build GRCh38
rs769533378 Chr.5: 60523874-60523874 on Build GRCh38
rs975712384 Chr.5: 60523877-60523877 on Build GRCh38

View Full Product Details