Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60526210-60526753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PART1
Entrez Gene ID
25859
Gene Name
prostate androgen-regulated transcript 1 (non-protein coding)
Gene Aliases
NCRNA00206
UniGene
Hs.146312
Gene Chromosome Location
Chr.5: 60487713-60547657 on Build GRCh38
Transcripts
NR_024617.1 NR_028509.1

SNP Detail

SNP ID Chromosome Location
rs986594135 Chr.5: 60526515-60526515 on Build GRCh38
rs904851520 Chr.5: 60526564-60526564 on Build GRCh38
rs1032522831 Chr.5: 60526696-60526696 on Build GRCh38
rs190619496 Chr.5: 60526587-60526587 on Build GRCh38
rs752183703 Chr.5: 60526485-60526485 on Build GRCh38
rs1055055769 Chr.5: 60526305-60526305 on Build GRCh38
rs1001518690 Chr.5: 60526371-60526371 on Build GRCh38
rs138530976 Chr.5: 60526722-60526722 on Build GRCh38
rs913331297 Chr.5: 60526414-60526414 on Build GRCh38
rs941152110 Chr.5: 60526724-60526725 on Build GRCh38
rs567727104 Chr.5: 60526657-60526657 on Build GRCh38
rs776966390 Chr.5: 60526267-60526267 on Build GRCh38
rs939908138 Chr.5: 60526580-60526580 on Build GRCh38
rs764113578 Chr.5: 60526518-60526518 on Build GRCh38
rs555334121 Chr.5: 60526556-60526556 on Build GRCh38
rs1041270162 Chr.5: 60526478-60526478 on Build GRCh38
rs184062122 Chr.5: 60526604-60526604 on Build GRCh38
rs911211503 Chr.5: 60526465-60526465 on Build GRCh38
rs758669320 Chr.5: 60526653-60526653 on Build GRCh38
rs888208182 Chr.5: 60526504-60526504 on Build GRCh38
rs576143171 Chr.5: 60526231-60526231 on Build GRCh38
rs76849498 Chr.5: 60526581-60526581 on Build GRCh38
rs144955992 Chr.5: 60526334-60526335 on Build GRCh38
rs765465868 Chr.5: 60526317-60526317 on Build GRCh38
rs1003489686 Chr.5: 60526565-60526565 on Build GRCh38
rs896546416 Chr.5: 60526306-60526306 on Build GRCh38
rs562520234 Chr.5: 60526673-60526673 on Build GRCh38
rs539160817 Chr.5: 60526399-60526399 on Build GRCh38
rs566467491 Chr.5: 60526430-60526430 on Build GRCh38
rs963996772 Chr.5: 60526661-60526661 on Build GRCh38
rs888168566 Chr.5: 60526252-60526252 on Build GRCh38
rs1023688518 Chr.5: 60526282-60526282 on Build GRCh38
rs1021523637 Chr.5: 60526340-60526340 on Build GRCh38
rs753613338 Chr.5: 60526713-60526714 on Build GRCh38
rs776966195 Chr.5: 60526611-60526611 on Build GRCh38
rs1047643682 Chr.5: 60526492-60526492 on Build GRCh38
rs551622892 Chr.5: 60526721-60526721 on Build GRCh38
rs969175789 Chr.5: 60526445-60526445 on Build GRCh38
rs1022509698 Chr.5: 60526621-60526621 on Build GRCh38
rs143006783 Chr.5: 60526668-60526668 on Build GRCh38
rs753660076 Chr.5: 60526523-60526523 on Build GRCh38
rs768298035 Chr.5: 60526461-60526461 on Build GRCh38
rs540722629 Chr.5: 60526634-60526634 on Build GRCh38
rs546338242 Chr.5: 60526416-60526416 on Build GRCh38
rs762264293 Chr.5: 60526275-60526275 on Build GRCh38
rs759185868 Chr.5: 60526413-60526413 on Build GRCh38
rs186143617 Chr.5: 60526490-60526490 on Build GRCh38
rs900729921 Chr.5: 60526706-60526706 on Build GRCh38
rs541822761 Chr.5: 60526639-60526639 on Build GRCh38
rs987950677 Chr.5: 60526378-60526378 on Build GRCh38
rs975044400 Chr.5: 60526466-60526466 on Build GRCh38
rs75051760 Chr.5: 60526623-60526623 on Build GRCh38
rs898497989 Chr.5: 60526253-60526253 on Build GRCh38
rs149031320 Chr.5: 60526658-60526658 on Build GRCh38
rs1005672171 Chr.5: 60526552-60526552 on Build GRCh38
rs867513572 Chr.5: 60526679-60526679 on Build GRCh38
rs73093122 Chr.5: 60526366-60526366 on Build GRCh38
rs760110643 Chr.5: 60526695-60526695 on Build GRCh38
rs188937510 Chr.5: 60526629-60526629 on Build GRCh38
rs193238624 Chr.5: 60526716-60526716 on Build GRCh38

View Full Product Details