Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 88745583-88746087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR4289
Entrez Gene ID
100423015
Gene Name
microRNA 4289
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.9: 88745836-88745905 on Build GRCh38
Transcripts
NR_036252.1

SNP Detail

SNP ID Chromosome Location
rs905500718 Chr.9: 88745919-88745919 on Build GRCh38
rs910494568 Chr.9: 88745947-88745947 on Build GRCh38
rs1015734204 Chr.9: 88746056-88746056 on Build GRCh38
rs1035409142 Chr.9: 88745698-88745698 on Build GRCh38
rs957312917 Chr.9: 88745744-88745744 on Build GRCh38
rs977111768 Chr.9: 88745732-88745732 on Build GRCh38
rs751214149 Chr.9: 88746064-88746064 on Build GRCh38
rs529451723 Chr.9: 88745980-88745980 on Build GRCh38
rs765058439 Chr.9: 88745890-88745890 on Build GRCh38
rs867629128 Chr.9: 88745819-88745819 on Build GRCh38
rs540869781 Chr.9: 88745861-88745861 on Build GRCh38
rs1035357269 Chr.9: 88746024-88746024 on Build GRCh38
rs1033856225 Chr.9: 88745670-88745670 on Build GRCh38
rs562779451 Chr.9: 88745876-88745876 on Build GRCh38
rs955982990 Chr.9: 88745644-88745644 on Build GRCh38
rs138610348 Chr.9: 88745638-88745638 on Build GRCh38
rs187933884 Chr.9: 88746044-88746044 on Build GRCh38
rs975975264 Chr.9: 88745999-88746000 on Build GRCh38
rs1001147413 Chr.9: 88746020-88746020 on Build GRCh38
rs890926219 Chr.9: 88745632-88745632 on Build GRCh38
rs28624420 Chr.9: 88745697-88745697 on Build GRCh38
rs547905765 Chr.9: 88746049-88746049 on Build GRCh38
rs545792088 Chr.9: 88745631-88745631 on Build GRCh38
rs544732693 Chr.9: 88745790-88745790 on Build GRCh38
rs776763931 Chr.9: 88745899-88745899 on Build GRCh38
rs909274017 Chr.9: 88746048-88746049 on Build GRCh38
rs998602240 Chr.9: 88745880-88745880 on Build GRCh38
rs761951355 Chr.9: 88745729-88745729 on Build GRCh38
rs992335898 Chr.9: 88745728-88745728 on Build GRCh38
rs143714958 Chr.9: 88745841-88745841 on Build GRCh38
rs906328320 Chr.9: 88745741-88745741 on Build GRCh38
rs1025549101 Chr.9: 88745875-88745875 on Build GRCh38
rs767916433 Chr.9: 88745763-88745763 on Build GRCh38
rs777204928 Chr.9: 88745901-88745901 on Build GRCh38
rs910426204 Chr.9: 88745768-88745768 on Build GRCh38
rs908727644 Chr.9: 88745782-88745782 on Build GRCh38
rs929105294 Chr.9: 88746034-88746034 on Build GRCh38
rs148056946 Chr.9: 88745862-88745862 on Build GRCh38
rs142917264 Chr.9: 88745829-88745829 on Build GRCh38
rs77620062 Chr.9: 88745690-88745690 on Build GRCh38
rs183749712 Chr.9: 88745842-88745842 on Build GRCh38
rs1006428373 Chr.9: 88745662-88745662 on Build GRCh38
rs544986803 Chr.9: 88745801-88745801 on Build GRCh38
rs757763595 Chr.9: 88745793-88745793 on Build GRCh38
rs1011934261 Chr.9: 88745655-88745655 on Build GRCh38
rs1021259674 Chr.9: 88745681-88745681 on Build GRCh38
rs773583997 Chr.9: 88745795-88745795 on Build GRCh38
rs752273861 Chr.9: 88745893-88745893 on Build GRCh38
rs567672580 Chr.9: 88746063-88746063 on Build GRCh38
rs990068775 Chr.9: 88745764-88745764 on Build GRCh38
rs190954348 Chr.9: 88745814-88745814 on Build GRCh38
rs1006145483 Chr.9: 88745849-88745849 on Build GRCh38
rs964973457 Chr.9: 88745963-88745963 on Build GRCh38
rs147422691 Chr.9: 88745851-88745851 on Build GRCh38
rs531273900 Chr.9: 88746017-88746017 on Build GRCh38
rs989784312 Chr.9: 88745920-88745920 on Build GRCh38
rs565008571 Chr.9: 88745812-88745812 on Build GRCh38
rs1031958788 Chr.9: 88745639-88745639 on Build GRCh38
rs959815449 Chr.9: 88745701-88745701 on Build GRCh38
rs940272659 Chr.9: 88745789-88745789 on Build GRCh38

View Full Product Details