Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58243413-58243969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP4R1L
Entrez Gene ID
55370
Gene Name
protein phosphatase 4 regulatory subunit 1 like (pseudogene)
Gene Aliases
C20orf192, PPP4R1P1, PRO1085
UniGene
Hs.473179
Gene Chromosome Location
Chr.20: 58232777-58309439 on Build GRCh38
Transcripts
NR_003505.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4848042 c.79G>A p.E27K Chr.20: 58243649-58243649 on Build GRCh38
1632364 c.131C>A p.P44H Chr.20: 58243597-58243597 on Build GRCh38
4137071 c.43G>A p.A15T Chr.20: 58243685-58243685 on Build GRCh38
5942892 c.42C>T p.S14S Chr.20: 58243686-58243686 on Build GRCh38
5942893 c.41C>T p.S14F Chr.20: 58243687-58243687 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767686555 Chr.20: 58243886-58243886 on Build GRCh38
rs945586485 Chr.20: 58243853-58243853 on Build GRCh38
rs61744799 Chr.20: 58243686-58243686 on Build GRCh38
rs941670909 Chr.20: 58243579-58243579 on Build GRCh38
rs977485102 Chr.20: 58243486-58243486 on Build GRCh38
rs777057789 Chr.20: 58243656-58243656 on Build GRCh38
rs765655330 Chr.20: 58243681-58243681 on Build GRCh38
rs577029747 Chr.20: 58243727-58243727 on Build GRCh38
rs970539019 Chr.20: 58243779-58243779 on Build GRCh38
rs756029579 Chr.20: 58243875-58243875 on Build GRCh38
rs556694321 Chr.20: 58243564-58243564 on Build GRCh38
rs1054439273 Chr.20: 58243563-58243563 on Build GRCh38
rs745604613 Chr.20: 58243553-58243553 on Build GRCh38
rs934208904 Chr.20: 58243893-58243893 on Build GRCh38
rs1033804611 Chr.20: 58243790-58243790 on Build GRCh38
rs56321006 Chr.20: 58243483-58243483 on Build GRCh38
rs147504432 Chr.20: 58243496-58243496 on Build GRCh38
rs914593941 Chr.20: 58243454-58243454 on Build GRCh38
rs545111992 Chr.20: 58243832-58243832 on Build GRCh38
rs1036154655 Chr.20: 58243655-58243655 on Build GRCh38
rs371638814 Chr.20: 58243670-58243670 on Build GRCh38
rs909127813 Chr.20: 58243873-58243873 on Build GRCh38
rs573942388 Chr.20: 58243690-58243690 on Build GRCh38
rs986079270 Chr.20: 58243870-58243870 on Build GRCh38
rs938466106 Chr.20: 58243906-58243906 on Build GRCh38
rs977215608 Chr.20: 58243815-58243815 on Build GRCh38
rs534653345 Chr.20: 58243449-58243449 on Build GRCh38
rs915483679 Chr.20: 58243833-58243833 on Build GRCh38
rs898855784 Chr.20: 58243943-58243943 on Build GRCh38
rs562546362 Chr.20: 58243752-58243752 on Build GRCh38
rs767528332 Chr.20: 58243800-58243800 on Build GRCh38
rs965503233 Chr.20: 58243802-58243802 on Build GRCh38
rs1026083232 Chr.20: 58243755-58243755 on Build GRCh38
rs977373837 Chr.20: 58243773-58243773 on Build GRCh38
rs181233790 Chr.20: 58243482-58243482 on Build GRCh38
rs769183755 Chr.20: 58243599-58243599 on Build GRCh38
rs970759566 Chr.20: 58243515-58243515 on Build GRCh38
rs527520383 Chr.20: 58243499-58243499 on Build GRCh38
rs963291217 Chr.20: 58243705-58243705 on Build GRCh38
rs199949983 Chr.20: 58243498-58243498 on Build GRCh38
rs751371214 Chr.20: 58243572-58243572 on Build GRCh38
rs982572917 Chr.20: 58243787-58243787 on Build GRCh38
rs746455993 Chr.20: 58243587-58243587 on Build GRCh38
rs968729039 Chr.20: 58243735-58243735 on Build GRCh38
rs1021799098 Chr.20: 58243697-58243697 on Build GRCh38
rs184876443 Chr.20: 58243687-58243687 on Build GRCh38
rs1036019386 Chr.20: 58243885-58243885 on Build GRCh38
rs762181072 Chr.20: 58243661-58243661 on Build GRCh38
rs888892999 Chr.20: 58243900-58243900 on Build GRCh38
rs533224001 Chr.20: 58243831-58243831 on Build GRCh38
rs751687302 Chr.20: 58243739-58243739 on Build GRCh38
rs879684505 Chr.20: 58243830-58243830 on Build GRCh38
rs1037339139 Chr.20: 58243925-58243925 on Build GRCh38
rs759785242 Chr.20: 58243514-58243514 on Build GRCh38
rs897466711 Chr.20: 58243894-58243894 on Build GRCh38
rs923068517 Chr.20: 58243546-58243546 on Build GRCh38
rs922730669 Chr.20: 58243881-58243881 on Build GRCh38
rs959634753 Chr.20: 58243810-58243810 on Build GRCh38
rs771596786 Chr.20: 58243555-58243555 on Build GRCh38
rs901359708 Chr.20: 58243598-58243598 on Build GRCh38
rs573109127 Chr.20: 58243685-58243685 on Build GRCh38
rs367903306 Chr.20: 58243669-58243669 on Build GRCh38
rs931933977 Chr.20: 58243558-58243558 on Build GRCh38
rs558155560 Chr.20: 58243588-58243588 on Build GRCh38
rs568928098 Chr.20: 58243571-58243571 on Build GRCh38
rs761501182 Chr.20: 58243715-58243715 on Build GRCh38
rs938036086 Chr.20: 58243557-58243557 on Build GRCh38

View Full Product Details