Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150942450-150943006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF300P1
Entrez Gene ID
134466
Gene Name
zinc finger protein 300 pseudogene 1
Gene Aliases
-
UniGene
Hs.661876
Gene Chromosome Location
Chr.5: 150930436-150946584 on Build GRCh38
Transcripts
NR_026867.1

SNP Detail

SNP ID Chromosome Location
rs928992343 Chr.5: 150942541-150942541 on Build GRCh38
rs759229004 Chr.5: 150942597-150942598 on Build GRCh38
rs957536794 Chr.5: 150942763-150942763 on Build GRCh38
rs528265925 Chr.5: 150942503-150942503 on Build GRCh38
rs746703639 Chr.5: 150942624-150942624 on Build GRCh38
rs902411979 Chr.5: 150942741-150942741 on Build GRCh38
rs964332820 Chr.5: 150942888-150942888 on Build GRCh38
rs953006615 Chr.5: 150942869-150942869 on Build GRCh38
rs751253698 Chr.5: 150942768-150942768 on Build GRCh38
rs17658318 Chr.5: 150942708-150942708 on Build GRCh38
rs749303665 Chr.5: 150942745-150942745 on Build GRCh38
rs777188378 Chr.5: 150942519-150942519 on Build GRCh38
rs970706228 Chr.5: 150942794-150942794 on Build GRCh38
rs544762154 Chr.5: 150942591-150942591 on Build GRCh38
rs191675275 Chr.5: 150942808-150942808 on Build GRCh38
rs199619349 Chr.5: 150942645-150942645 on Build GRCh38
rs563061857 Chr.5: 150942665-150942665 on Build GRCh38
rs1025107359 Chr.5: 150942475-150942475 on Build GRCh38
rs750859248 Chr.5: 150942549-150942549 on Build GRCh38
rs531304088 Chr.5: 150942893-150942893 on Build GRCh38
rs752675577 Chr.5: 150942709-150942709 on Build GRCh38
rs908883972 Chr.5: 150942886-150942886 on Build GRCh38
rs112258423 Chr.5: 150942537-150942537 on Build GRCh38
rs115531107 Chr.5: 150942943-150942943 on Build GRCh38
rs75906401 Chr.5: 150942648-150942648 on Build GRCh38
rs563980784 Chr.5: 150942832-150942832 on Build GRCh38
rs577268629 Chr.5: 150942585-150942585 on Build GRCh38
rs1046901056 Chr.5: 150942579-150942579 on Build GRCh38
rs909266099 Chr.5: 150942894-150942894 on Build GRCh38
rs150207116 Chr.5: 150942734-150942734 on Build GRCh38
rs1053728683 Chr.5: 150942761-150942761 on Build GRCh38
rs763442704 Chr.5: 150942554-150942554 on Build GRCh38
rs17800981 Chr.5: 150942509-150942509 on Build GRCh38
rs984341801 Chr.5: 150942884-150942884 on Build GRCh38
rs766661989 Chr.5: 150942555-150942555 on Build GRCh38
rs773880286 Chr.5: 150942658-150942658 on Build GRCh38
rs974525910 Chr.5: 150942916-150942916 on Build GRCh38
rs1005312727 Chr.5: 150942620-150942620 on Build GRCh38
rs745611706 Chr.5: 150942593-150942593 on Build GRCh38
rs757178807 Chr.5: 150942586-150942586 on Build GRCh38
rs750407812 Chr.5: 150942953-150942953 on Build GRCh38
rs1026339754 Chr.5: 150942846-150942846 on Build GRCh38
rs936600908 Chr.5: 150942978-150942978 on Build GRCh38
rs933175920 Chr.5: 150942936-150942936 on Build GRCh38
rs115445748 Chr.5: 150942766-150942766 on Build GRCh38
rs765611600 Chr.5: 150942546-150942546 on Build GRCh38
rs751847649 Chr.5: 150942563-150942563 on Build GRCh38
rs932544470 Chr.5: 150942962-150942962 on Build GRCh38
rs971337058 Chr.5: 150942821-150942821 on Build GRCh38
rs756236573 Chr.5: 150942711-150942711 on Build GRCh38
rs1000713979 Chr.5: 150942753-150942753 on Build GRCh38
rs908852883 Chr.5: 150942623-150942623 on Build GRCh38
rs771864188 Chr.5: 150942594-150942594 on Build GRCh38
rs763354104 Chr.5: 150942659-150942659 on Build GRCh38
rs751805501 Chr.5: 150942677-150942677 on Build GRCh38
rs17111493 Chr.5: 150942686-150942686 on Build GRCh38
rs768199050 Chr.5: 150942627-150942627 on Build GRCh38
rs781200006 Chr.5: 150942577-150942577 on Build GRCh38
rs549507939 Chr.5: 150942923-150942923 on Build GRCh38
rs779613392 Chr.5: 150942596-150942596 on Build GRCh38
rs776053133 Chr.5: 150942636-150942636 on Build GRCh38
rs763934602 Chr.5: 150942736-150942736 on Build GRCh38
rs755021061 Chr.5: 150942571-150942571 on Build GRCh38

View Full Product Details