Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42287281-42287439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIC
Entrez Gene ID
23152
Gene Name
capicua transcriptional repressor
Gene Aliases
-
UniGene
Hs.388236
Gene Chromosome Location
Chr.19: 42268537-42295796 on Build GRCh38
Transcripts
NM_001304815.1 NM_015125.4 XM_005258673.2 XM_005258674.2 XM_005258675.2 XM_011526660.2 XM_011526661.2 XM_011526662.2 XM_011526663.2 XM_011526664.2 XM_011526665.2 XM_011526666.2 XM_011526667.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4984444 c.470C>G p.S157C Chr.19: 42287337-42287337 on Build GRCh38
6915185 c.500G>A p.G167E Chr.19: 42287367-42287367 on Build GRCh38
107328 c.518C>T p.S173F Chr.19: 42287385-42287385 on Build GRCh38
374313 c.473C>T p.P158L Chr.19: 42287340-42287340 on Build GRCh38
6429864 c.509G>T p.R170L Chr.19: 42287376-42287376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202120202 Chr.19: 42287419-42287419 on Build GRCh38
rs28469564 Chr.19: 42287310-42287310 on Build GRCh38
rs751520566 Chr.19: 42287332-42287332 on Build GRCh38
rs769059501 Chr.19: 42287413-42287413 on Build GRCh38
rs771545626 Chr.19: 42287314-42287314 on Build GRCh38
rs969199047 Chr.19: 42287409-42287409 on Build GRCh38
rs147753753 Chr.19: 42287385-42287385 on Build GRCh38
rs771041360 Chr.19: 42287306-42287306 on Build GRCh38
rs368616680 Chr.19: 42287367-42287367 on Build GRCh38
rs772904133 Chr.19: 42287324-42287324 on Build GRCh38
rs772636832 Chr.19: 42287366-42287366 on Build GRCh38
rs34324376 Chr.19: 42287385-42287385 on Build GRCh38
rs747679652 Chr.19: 42287311-42287311 on Build GRCh38
rs1041259071 Chr.19: 42287301-42287301 on Build GRCh38
rs199958449 Chr.19: 42287330-42287330 on Build GRCh38
rs757335104 Chr.19: 42287362-42287362 on Build GRCh38
rs775953544 Chr.19: 42287312-42287312 on Build GRCh38
rs932288253 Chr.19: 42287303-42287303 on Build GRCh38
rs1011626127 Chr.19: 42287408-42287408 on Build GRCh38
rs587778209 Chr.19: 42287418-42287418 on Build GRCh38
rs755927294 Chr.19: 42287359-42287359 on Build GRCh38
rs77843943 Chr.19: 42287365-42287365 on Build GRCh38
rs375676767 Chr.19: 42287356-42287356 on Build GRCh38
rs769270589 Chr.19: 42287316-42287316 on Build GRCh38
rs371107104 Chr.19: 42287315-42287315 on Build GRCh38
rs747433677 Chr.19: 42287372-42287372 on Build GRCh38
rs562265768 Chr.19: 42287313-42287313 on Build GRCh38
rs754063822 Chr.19: 42287361-42287361 on Build GRCh38
rs759429513 Chr.19: 42287340-42287340 on Build GRCh38
rs777927145 Chr.19: 42287360-42287360 on Build GRCh38
rs573476602 Chr.19: 42287305-42287305 on Build GRCh38
rs900099983 Chr.19: 42287302-42287302 on Build GRCh38
rs751030414 Chr.19: 42287308-42287312 on Build GRCh38
rs200317945 Chr.19: 42287353-42287353 on Build GRCh38

View Full Product Details