Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101131694-101131868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLX1
Entrez Gene ID
3195
Gene Name
T-cell leukemia homeobox 1
Gene Aliases
HOX11, TCL3
UniGene
Hs.89583
Gene Chromosome Location
Chr.10: 101130773-101137789 on Build GRCh38
Transcripts
NM_001195517.1 NM_005521.3 XM_011539744.2 XM_011539745.2 NR_130724.1

Gene Symbol
TLX1NB
Entrez Gene ID
100038246
Gene Name
TLX1 neighbor
Gene Aliases
APT-B7, TD1, TDI
UniGene
Hs.368784
Gene Chromosome Location
Chr.10: 101089321-101141266 on Build GRCh38
Transcripts
NM_001195517.1 NM_005521.3 XM_011539744.2 XM_011539745.2 NR_130724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6551254 c.259G>T p.G87C Chr.10: 101131800-101131800 on Build GRCh38
6551253 c.175G>T p.G59C Chr.10: 101131716-101131716 on Build GRCh38
5999356 c.241G>T p.G81C Chr.10: 101131782-101131782 on Build GRCh38
6518210 c.267C>A p.G89G Chr.10: 101131808-101131808 on Build GRCh38
1602882 c.322G>C p.A108P Chr.10: 101131863-101131863 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984454722 Chr.10: 101131768-101131768 on Build GRCh38
rs780244330 Chr.10: 101131734-101131734 on Build GRCh38
rs536772130 Chr.10: 101131835-101131835 on Build GRCh38
rs972033453 Chr.10: 101131812-101131812 on Build GRCh38
rs756282883 Chr.10: 101131731-101131731 on Build GRCh38
rs992155351 Chr.10: 101131753-101131753 on Build GRCh38
rs569394912 Chr.10: 101131807-101131807 on Build GRCh38
rs975804059 Chr.10: 101131795-101131795 on Build GRCh38
rs747467520 Chr.10: 101131808-101131808 on Build GRCh38
rs748613492 Chr.10: 101131781-101131781 on Build GRCh38
rs999899225 Chr.10: 101131740-101131740 on Build GRCh38
rs778994450 Chr.10: 101131763-101131763 on Build GRCh38
rs867078365 Chr.10: 101131806-101131806 on Build GRCh38
rs755280568 Chr.10: 101131741-101131741 on Build GRCh38
rs557943782 Chr.10: 101131757-101131757 on Build GRCh38
rs907653930 Chr.10: 101131771-101131771 on Build GRCh38
rs768417758 Chr.10: 101131836-101131836 on Build GRCh38
rs2742015 Chr.10: 101131752-101131752 on Build GRCh38
rs940435257 Chr.10: 101131782-101131782 on Build GRCh38
rs775029487 Chr.10: 101131828-101131828 on Build GRCh38
rs758625391 Chr.10: 101131784-101131784 on Build GRCh38
rs750325726 Chr.10: 101131729-101131729 on Build GRCh38
rs771871097 Chr.10: 101131714-101131715 on Build GRCh38
rs773093460 Chr.10: 101131799-101131800 on Build GRCh38
rs917107167 Chr.10: 101131769-101131769 on Build GRCh38
rs1027204962 Chr.10: 101131754-101131754 on Build GRCh38
rs771395705 Chr.10: 101131826-101131826 on Build GRCh38
rs1032683928 Chr.10: 101131756-101131756 on Build GRCh38
rs753834322 Chr.10: 101131735-101131735 on Build GRCh38
rs967604146 Chr.10: 101131800-101131802 on Build GRCh38

View Full Product Details