Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 111707940-111708203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLCXD2
Entrez Gene ID
257068
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 2
Gene Aliases
-
UniGene
Hs.477114
Gene Chromosome Location
Chr.3: 111674676-111846447 on Build GRCh38
Transcripts
NM_001185106.1 NM_153268.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3585221 c.393C>T p.I131I Chr.3: 111708155-111708155 on Build GRCh38
6748474 c.401delT p.F135fs*9 Chr.3: 111708163-111708163 on Build GRCh38
4317674 c.411C>G p.I137M Chr.3: 111708173-111708173 on Build GRCh38
1036224 c.351G>A p.V117V Chr.3: 111708113-111708113 on Build GRCh38
6896416 c.196G>A p.E66K Chr.3: 111707958-111707958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984857069 Chr.3: 111708158-111708158 on Build GRCh38
rs373550772 Chr.3: 111708129-111708129 on Build GRCh38
rs866667223 Chr.3: 111708133-111708133 on Build GRCh38
rs748891173 Chr.3: 111708006-111708006 on Build GRCh38
rs954267073 Chr.3: 111708059-111708059 on Build GRCh38
rs751841138 Chr.3: 111708094-111708094 on Build GRCh38
rs138285901 Chr.3: 111708001-111708001 on Build GRCh38
rs774099382 Chr.3: 111708150-111708150 on Build GRCh38
rs986928817 Chr.3: 111708073-111708073 on Build GRCh38
rs779661439 Chr.3: 111708134-111708134 on Build GRCh38
rs761706804 Chr.3: 111708041-111708041 on Build GRCh38
rs571921225 Chr.3: 111708069-111708069 on Build GRCh38
rs35531992 Chr.3: 111708129-111708129 on Build GRCh38
rs199703580 Chr.3: 111708093-111708093 on Build GRCh38
rs747884333 Chr.3: 111708151-111708151 on Build GRCh38
rs191366528 Chr.3: 111708053-111708053 on Build GRCh38
rs149301928 Chr.3: 111708168-111708168 on Build GRCh38
rs748024981 Chr.3: 111708114-111708114 on Build GRCh38
rs555338951 Chr.3: 111708109-111708109 on Build GRCh38
rs144009553 Chr.3: 111708068-111708068 on Build GRCh38
rs760549244 Chr.3: 111708161-111708161 on Build GRCh38
rs752984974 Chr.3: 111707967-111707967 on Build GRCh38
rs1047499914 Chr.3: 111708007-111708007 on Build GRCh38
rs201638622 Chr.3: 111708108-111708108 on Build GRCh38
rs746307203 Chr.3: 111708128-111708128 on Build GRCh38
rs149378405 Chr.3: 111708011-111708011 on Build GRCh38
rs767336717 Chr.3: 111708043-111708043 on Build GRCh38
rs147048469 Chr.3: 111708000-111708000 on Build GRCh38
rs759609181 Chr.3: 111708060-111708060 on Build GRCh38
rs1018721117 Chr.3: 111708075-111708075 on Build GRCh38
rs368917773 Chr.3: 111707965-111707965 on Build GRCh38
rs747426362 Chr.3: 111707987-111707987 on Build GRCh38
rs775510663 Chr.3: 111708175-111708175 on Build GRCh38
rs377101481 Chr.3: 111708070-111708070 on Build GRCh38
rs771971223 Chr.3: 111708154-111708154 on Build GRCh38
rs758655679 Chr.3: 111707970-111707970 on Build GRCh38
rs767646237 Chr.3: 111707979-111707979 on Build GRCh38
rs138778348 Chr.3: 111708141-111708141 on Build GRCh38
rs772996504 Chr.3: 111708157-111708157 on Build GRCh38
rs777301432 Chr.3: 111708029-111708029 on Build GRCh38
rs1057014395 Chr.3: 111707975-111707975 on Build GRCh38
rs201874936 Chr.3: 111707992-111707992 on Build GRCh38
rs768160911 Chr.3: 111708135-111708135 on Build GRCh38
rs370877205 Chr.3: 111708100-111708100 on Build GRCh38
rs143931101 Chr.3: 111708005-111708005 on Build GRCh38
rs549501624 Chr.3: 111707998-111707998 on Build GRCh38

View Full Product Details