Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24407827-24408074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NYNRIN
Entrez Gene ID
57523
Gene Name
NYN domain and retroviral integrase containing
Gene Aliases
CGIN1, KIAA1305
UniGene
Hs.288348
Gene Chromosome Location
Chr.14: 24398786-24419288 on Build GRCh38
Transcripts
NM_025081.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3782833 c.363C>T p.G121G Chr.14: 24408033-24408033 on Build GRCh38
6226169 c.263A>G p.H88R Chr.14: 24407933-24407933 on Build GRCh38
394429 c.319C>T p.L107F Chr.14: 24407989-24407989 on Build GRCh38
6732034 c.338G>C p.G113A Chr.14: 24408008-24408008 on Build GRCh38
6216097 c.270C>T p.A90A Chr.14: 24407940-24407940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753798964 Chr.14: 24408052-24408052 on Build GRCh38
rs750685237 Chr.14: 24408000-24408000 on Build GRCh38
rs77367832 Chr.14: 24407858-24407858 on Build GRCh38
rs116410758 Chr.14: 24407856-24407856 on Build GRCh38
rs972257676 Chr.14: 24407997-24407997 on Build GRCh38
rs1024917051 Chr.14: 24407861-24407861 on Build GRCh38
rs764539965 Chr.14: 24407845-24407845 on Build GRCh38
rs756142193 Chr.14: 24407914-24407914 on Build GRCh38
rs762144552 Chr.14: 24407979-24407979 on Build GRCh38
rs758632924 Chr.14: 24407908-24407908 on Build GRCh38
rs780283437 Chr.14: 24407911-24407911 on Build GRCh38
rs778485525 Chr.14: 24407933-24407933 on Build GRCh38
rs1016527220 Chr.14: 24407954-24407954 on Build GRCh38
rs777192126 Chr.14: 24407971-24407971 on Build GRCh38
rs751733364 Chr.14: 24408020-24408020 on Build GRCh38
rs750645928 Chr.14: 24407900-24407900 on Build GRCh38
rs10148781 Chr.14: 24408009-24408009 on Build GRCh38
rs1009271726 Chr.14: 24407888-24407888 on Build GRCh38
rs146351794 Chr.14: 24407955-24407955 on Build GRCh38
rs769191962 Chr.14: 24407959-24407959 on Build GRCh38
rs771675951 Chr.14: 24407946-24407946 on Build GRCh38
rs189369960 Chr.14: 24408033-24408033 on Build GRCh38
rs986563058 Chr.14: 24408047-24408047 on Build GRCh38
rs368797604 Chr.14: 24407925-24407925 on Build GRCh38
rs765646101 Chr.14: 24407988-24407988 on Build GRCh38
rs139716312 Chr.14: 24407967-24407967 on Build GRCh38
rs557266750 Chr.14: 24408013-24408013 on Build GRCh38
rs200331786 Chr.14: 24407864-24407864 on Build GRCh38
rs745526277 Chr.14: 24407945-24407945 on Build GRCh38
rs761205623 Chr.14: 24407844-24407844 on Build GRCh38
rs571250742 Chr.14: 24408011-24408011 on Build GRCh38
rs777739171 Chr.14: 24407922-24407922 on Build GRCh38
rs1052732727 Chr.14: 24407846-24407846 on Build GRCh38
rs777636022 Chr.14: 24408041-24408041 on Build GRCh38
rs112226335 Chr.14: 24407886-24407886 on Build GRCh38

View Full Product Details